Assay Details
| Gene Symbol: XYLT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_022167.3 | NP_071450.2 | 6 - 7 | 1412 | 91 | |
| NR_110010.1 | 6 - 7 | 1412 | 91 | |||
| XM_005257572.4 | XP_005257629.1 | 6 - 7 | 1726 | 91 | ||
| GenBank mRNA | AJ277442.1 | - | 6 - 7 | 1412 | 91 | |
| AK299999.1 | - | 6 - 7 | 1309 | 91 | ||
| AY358090.1 | - | 6 - 7 | 1308 | 91 | ||
| BC052262.1 | - | 6 - 7 | 1332 | 6380013 | 91 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC17000663.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
glycosaminoglycan biosynthetic process
heparan sulfate proteoglycan biosynthetic process
chondroitin sulfate biosynthetic process
heparin biosynthetic process
chondroitin sulfate proteoglycan biosynthetic process
acetylglucosaminyltransferase activity
protein xylosyltransferase activity
glycosaminoglycan biosynthetic process
heparan sulfate proteoglycan biosynthetic process
chondroitin sulfate biosynthetic process
heparin biosynthetic process
chondroitin sulfate proteoglycan biosynthetic process
acetylglucosaminyltransferase activity
protein xylosyltransferase activity
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1700008250.hg.1 | ENST00000017003.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008250.hg.1 | ENST00000017003.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008250.hg.1 | ENST00000017003.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008250.hg.1 | ENST00000017003.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008250.hg.1 | ENST00000017003.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008250.hg.1 | ENST00000017003.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008250.hg.1 | ENST00000017003.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008250.hg.1 | ENST00000017003.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008250.hg.1 | ENST00000017003.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008250.hg.1 | ENST00000017003.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008250.hg.1 | ENST00000017003.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008250.hg.1 | ENST00000017003.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008250.hg.1 | ENST00000017003.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008250.hg.1 | ENST00000017003.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008250.hg.1 | ENST00000017003.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008250.hg.1 | ENST00000017003.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008250.hg.1 | ENST00000017003.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008250.hg.1 | ENST00000017003.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008250.hg.1 | ENST00000376550.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008250.hg.1 | ENST00000376550.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008250.hg.1 | ENST00000376550.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008250.hg.1 | ENST00000376550.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008250.hg.1 | ENST00000376550.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008250.hg.1 | ENST00000376550.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008250.hg.1 | ENST00000376550.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008250.hg.1 | ENST00000376550.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008250.hg.1 | ENST00000376550.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008250.hg.1 | ENST00000376550.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008250.hg.1 | ENST00000376550.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008250.hg.1 | ENST00000376550.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008250.hg.1 | ENST00000376550.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008250.hg.1 | ENST00000376550.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008250.hg.1 | ENST00000376550.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008250.hg.1 | ENST00000507602.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008250.hg.1 | ENST00000507602.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008250.hg.1 | ENST00000507602.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008250.hg.1 | ENST00000507602.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008250.hg.1 | ENST00000507602.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008250.hg.1 | ENST00000507602.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008250.hg.1 | ENST00000507602.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008250.hg.1 | ENST00000507602.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008250.hg.1 | ENST00000507602.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008250.hg.1 | ENST00000507602.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008250.hg.1 | ENST00000507602.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008250.hg.1 | ENST00000507602.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008250.hg.1 | ENST00000507602.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008250.hg.1 | ENST00000507602.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008250.hg.1 | ENST00000507602.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008250.hg.1 | ENST00000509778.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008250.hg.1 | ENST00000509778.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008250.hg.1 | ENST00000511654.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008250.hg.1 | ENST00000511654.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008250.hg.1 | ENST00000511654.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008250.hg.1 | ENST00000511654.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008250.hg.1 | ENST00000511654.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008250.hg.1 | ENST00000571021.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008250.hg.1 | ENST00000571021.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008250.hg.1 | ENST00000571021.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008250.hg.1 | ENST00000574840.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008250.hg.1 | ENST00000574840.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008250.hg.1 | NM_022167.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008250.hg.1 | NM_022167.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008250.hg.1 | NM_022167.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008250.hg.1 | NM_022167.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008250.hg.1 | NM_022167.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008250.hg.1 | NM_022167.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008250.hg.1 | NM_022167.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008250.hg.1 | NM_022167.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008250.hg.1 | NM_022167.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008250.hg.1 | NM_022167.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008250.hg.1 | NM_022167.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008250.hg.1 | NM_022167.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008250.hg.1 | NM_022167.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008250.hg.1 | NM_022167.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008250.hg.1 | NM_022167.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008250.hg.1 | NM_022167.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008250.hg.1 | NM_022167.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008250.hg.1 | NM_022167.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008250.hg.1 | NM_022167.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008250.hg.1 | NM_022167.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008250.hg.1 | NR_110010.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008250.hg.1 | NR_110010.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008250.hg.1 | NR_110010.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008250.hg.1 | NR_110010.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008250.hg.1 | NR_110010.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008250.hg.1 | NR_110010.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008250.hg.1 | NR_110010.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008250.hg.1 | NR_110010.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008250.hg.1 | NR_110010.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008250.hg.1 | NR_110010.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008250.hg.1 | NR_110010.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008250.hg.1 | NR_110010.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008250.hg.1 | NR_110010.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008250.hg.1 | NR_110010.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008250.hg.1 | NR_110010.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008250.hg.1 | NR_110010.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008250.hg.1 | NR_110010.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17008334.hg.1 | ENST00000017003.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17008334.hg.1 | ENST00000507602.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17008334.hg.1 | NM_022167.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17008334.hg.1 | TC17000663.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1700161087.hg.1 | ENST00000017003.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700161087.hg.1 | ENST00000376550.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700161087.hg.1 | ENST00000507602.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700161087.hg.1 | NM_022167.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700161087.hg.1 | NR_110010.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700161087.hg.1 | ENST00000017003.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700161087.hg.1 | ENST00000376550.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700161087.hg.1 | ENST00000507602.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700161087.hg.1 | NM_022167.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700161087.hg.1 | NR_110010.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700161087.hg.1 | TC1700008250.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1700008250.hg.1 | ENST00000017003.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008250.hg.1 | ENST00000017003.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008250.hg.1 | ENST00000017003.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008250.hg.1 | ENST00000017003.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008250.hg.1 | ENST00000017003.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008250.hg.1 | ENST00000017003.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008250.hg.1 | ENST00000017003.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008250.hg.1 | ENST00000017003.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008250.hg.1 | ENST00000017003.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008250.hg.1 | ENST00000017003.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008250.hg.1 | ENST00000017003.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008250.hg.1 | ENST00000017003.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008250.hg.1 | ENST00000017003.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008250.hg.1 | ENST00000017003.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008250.hg.1 | ENST00000017003.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008250.hg.1 | ENST00000017003.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008250.hg.1 | ENST00000017003.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008250.hg.1 | ENST00000017003.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008250.hg.1 | ENST00000376550.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008250.hg.1 | ENST00000376550.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008250.hg.1 | ENST00000376550.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008250.hg.1 | ENST00000376550.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008250.hg.1 | ENST00000376550.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008250.hg.1 | ENST00000376550.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008250.hg.1 | ENST00000376550.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008250.hg.1 | ENST00000376550.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008250.hg.1 | ENST00000376550.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008250.hg.1 | ENST00000376550.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008250.hg.1 | ENST00000376550.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008250.hg.1 | ENST00000376550.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008250.hg.1 | ENST00000376550.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008250.hg.1 | ENST00000376550.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008250.hg.1 | ENST00000376550.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008250.hg.1 | ENST00000507602.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008250.hg.1 | ENST00000507602.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008250.hg.1 | ENST00000507602.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008250.hg.1 | ENST00000507602.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008250.hg.1 | ENST00000507602.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008250.hg.1 | ENST00000507602.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008250.hg.1 | ENST00000507602.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008250.hg.1 | ENST00000507602.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008250.hg.1 | ENST00000507602.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008250.hg.1 | ENST00000507602.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008250.hg.1 | ENST00000507602.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008250.hg.1 | ENST00000507602.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008250.hg.1 | ENST00000507602.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008250.hg.1 | ENST00000507602.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008250.hg.1 | ENST00000507602.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008250.hg.1 | ENST00000509778.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008250.hg.1 | ENST00000509778.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008250.hg.1 | ENST00000511654.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008250.hg.1 | ENST00000511654.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008250.hg.1 | ENST00000511654.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008250.hg.1 | ENST00000511654.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008250.hg.1 | ENST00000511654.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008250.hg.1 | ENST00000571021.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008250.hg.1 | ENST00000571021.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008250.hg.1 | ENST00000571021.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008250.hg.1 | ENST00000574840.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008250.hg.1 | ENST00000574840.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008250.hg.1 | NM_022167.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008250.hg.1 | NM_022167.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008250.hg.1 | NM_022167.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008250.hg.1 | NM_022167.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008250.hg.1 | NM_022167.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008250.hg.1 | NM_022167.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008250.hg.1 | NM_022167.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008250.hg.1 | NM_022167.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008250.hg.1 | NM_022167.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008250.hg.1 | NM_022167.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008250.hg.1 | NM_022167.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008250.hg.1 | NM_022167.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008250.hg.1 | NM_022167.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008250.hg.1 | NM_022167.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008250.hg.1 | NM_022167.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008250.hg.1 | NM_022167.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008250.hg.1 | NM_022167.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008250.hg.1 | NM_022167.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008250.hg.1 | NM_022167.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008250.hg.1 | NM_022167.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008250.hg.1 | NR_110010.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008250.hg.1 | NR_110010.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008250.hg.1 | NR_110010.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008250.hg.1 | NR_110010.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008250.hg.1 | NR_110010.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008250.hg.1 | NR_110010.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008250.hg.1 | NR_110010.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008250.hg.1 | NR_110010.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008250.hg.1 | NR_110010.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008250.hg.1 | NR_110010.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008250.hg.1 | NR_110010.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008250.hg.1 | NR_110010.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008250.hg.1 | NR_110010.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008250.hg.1 | NR_110010.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008250.hg.1 | NR_110010.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008250.hg.1 | NR_110010.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008250.hg.1 | NR_110010.1 | Refseq |

XYLT2
Genomic Map