Assay Details
| Gene Symbol: ANXA6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005268432.4 | XP_005268489.1 | 20 - 21 | 1801 | 93 | |
| GenBank mRNA | KU177904.1 | - | 19 - 20 | 1573 | 93 | |
More Information
Set Membership: |
|

ANXA6
Genomic Map