Assay Details
| Gene Symbol: TCEB3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_003198.2 | NP_003189.2 | 8 - 9 | 2112 | 61 | |
| GenBank mRNA | AF116729.1 | - | 8 - 9 | 1961 | 61 | |
| AK310229.1 | - | 8 - 9 | 2098 | 61 | ||
| AK313079.1 | - | 8 - 9 | 1984 | 61 | ||
| BC002883.2 | - | 8 - 9 | 1996 | 3941728 | 61 | |
| BC020448.1 | - | 5 - 6 | 724 | 61 | ||
| L47345.1 | - | 8 - 9 | 2006 | 61 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01000300.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR01004559.hg.1 | TC01000300.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01004559.hg.1 | ENST00000418390.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01004559.hg.1 | NM_003198.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007331.hg.1 | ENST00000418390.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007331.hg.1 | ENST00000418390.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007331.hg.1 | ENST00000418390.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007331.hg.1 | ENST00000418390.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007331.hg.1 | ENST00000418390.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007331.hg.1 | ENST00000418390.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007331.hg.1 | ENST00000418390.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007331.hg.1 | ENST00000418390.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007331.hg.1 | ENST00000418390.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007331.hg.1 | ENST00000418390.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007331.hg.1 | ENST00000418390.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007331.hg.1 | ENST00000418390.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007331.hg.1 | ENST00000418390.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007331.hg.1 | ENST00000418390.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007331.hg.1 | ENST00000418390.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007331.hg.1 | ENST00000418390.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007331.hg.1 | ENST00000418390.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007331.hg.1 | ENST00000487554.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007331.hg.1 | ENST00000487554.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007331.hg.1 | ENST00000487554.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007331.hg.1 | ENST00000487554.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007331.hg.1 | ENST00000609199.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007331.hg.1 | ENST00000609199.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007331.hg.1 | ENST00000609199.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007331.hg.1 | ENST00000609199.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007331.hg.1 | ENST00000609199.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007331.hg.1 | ENST00000609199.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007331.hg.1 | ENST00000609199.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007331.hg.1 | ENST00000609199.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007331.hg.1 | ENST00000609199.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007331.hg.1 | ENST00000609199.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007331.hg.1 | ENST00000609199.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007331.hg.1 | ENST00000609199.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007331.hg.1 | ENST00000609199.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007331.hg.1 | ENST00000613537.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007331.hg.1 | ENST00000613537.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007331.hg.1 | ENST00000613537.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007331.hg.1 | ENST00000613537.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007331.hg.1 | ENST00000613537.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007331.hg.1 | ENST00000613537.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007331.hg.1 | ENST00000613537.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007331.hg.1 | ENST00000613537.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007331.hg.1 | ENST00000613537.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007331.hg.1 | ENST00000613537.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007331.hg.1 | ENST00000613537.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007331.hg.1 | ENST00000613537.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007331.hg.1 | ENST00000613537.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007331.hg.1 | ENST00000613537.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007331.hg.1 | ENST00000613537.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007331.hg.1 | ENST00000613537.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007331.hg.1 | NM_003198.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007331.hg.1 | NM_003198.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007331.hg.1 | NM_003198.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007331.hg.1 | NM_003198.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007331.hg.1 | NM_003198.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007331.hg.1 | NM_003198.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007331.hg.1 | NM_003198.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007331.hg.1 | NM_003198.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007331.hg.1 | NM_003198.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007331.hg.1 | NM_003198.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007331.hg.1 | NM_003198.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007331.hg.1 | NM_003198.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007331.hg.1 | NM_003198.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007331.hg.1 | NM_003198.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007331.hg.1 | NM_003198.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007331.hg.1 | NM_003198.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100151042.hg.1 | ENST00000418390.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100151042.hg.1 | ENST00000609199.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100151042.hg.1 | ENST00000613537.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100151042.hg.1 | NM_003198.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100151042.hg.1 | ENST00000418390.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100151042.hg.1 | ENST00000609199.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100151042.hg.1 | ENST00000613537.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100151042.hg.1 | NM_003198.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100151042.hg.1 | TC0100007331.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0100007331.hg.1 | ENST00000418390.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007331.hg.1 | ENST00000418390.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007331.hg.1 | ENST00000418390.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007331.hg.1 | ENST00000418390.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007331.hg.1 | ENST00000418390.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007331.hg.1 | ENST00000418390.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007331.hg.1 | ENST00000418390.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007331.hg.1 | ENST00000418390.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007331.hg.1 | ENST00000418390.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007331.hg.1 | ENST00000418390.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007331.hg.1 | ENST00000418390.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007331.hg.1 | ENST00000418390.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007331.hg.1 | ENST00000418390.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007331.hg.1 | ENST00000418390.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007331.hg.1 | ENST00000418390.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007331.hg.1 | ENST00000418390.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007331.hg.1 | ENST00000418390.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007331.hg.1 | ENST00000487554.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007331.hg.1 | ENST00000487554.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007331.hg.1 | ENST00000487554.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007331.hg.1 | ENST00000487554.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007331.hg.1 | ENST00000609199.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007331.hg.1 | ENST00000609199.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007331.hg.1 | ENST00000609199.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007331.hg.1 | ENST00000609199.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007331.hg.1 | ENST00000609199.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007331.hg.1 | ENST00000609199.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007331.hg.1 | ENST00000609199.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007331.hg.1 | ENST00000609199.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007331.hg.1 | ENST00000609199.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007331.hg.1 | ENST00000609199.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007331.hg.1 | ENST00000609199.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007331.hg.1 | ENST00000609199.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007331.hg.1 | ENST00000609199.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007331.hg.1 | ENST00000613537.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007331.hg.1 | ENST00000613537.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007331.hg.1 | ENST00000613537.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007331.hg.1 | ENST00000613537.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007331.hg.1 | ENST00000613537.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007331.hg.1 | ENST00000613537.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007331.hg.1 | ENST00000613537.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007331.hg.1 | ENST00000613537.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007331.hg.1 | ENST00000613537.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007331.hg.1 | ENST00000613537.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007331.hg.1 | ENST00000613537.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007331.hg.1 | ENST00000613537.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007331.hg.1 | ENST00000613537.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007331.hg.1 | ENST00000613537.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007331.hg.1 | ENST00000613537.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007331.hg.1 | ENST00000613537.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007331.hg.1 | NM_003198.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007331.hg.1 | NM_003198.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007331.hg.1 | NM_003198.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007331.hg.1 | NM_003198.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007331.hg.1 | NM_003198.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007331.hg.1 | NM_003198.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007331.hg.1 | NM_003198.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007331.hg.1 | NM_003198.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007331.hg.1 | NM_003198.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007331.hg.1 | NM_003198.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007331.hg.1 | NM_003198.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007331.hg.1 | NM_003198.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007331.hg.1 | NM_003198.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007331.hg.1 | NM_003198.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007331.hg.1 | NM_003198.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007331.hg.1 | NM_003198.2 | Refseq |

TCEB3
Genomic Map