Assay Details
| Gene Symbol: CA5B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007220.3 | NP_009151.1 | 3 - 4 | 456 | 103 | |
| XM_005274442.4 | XP_005274499.1 | 4 - 5 | 544 | 103 | ||
| XM_017029247.1 | XP_016884736.1 | 2 - 3 | 242 | 103 | ||
| GenBank mRNA | AB021660.1 | - | 3 - 4 | 476 | 103 | |
| AK291050.1 | - | 3 - 4 | 475 | 103 | ||
| BC028142.2 | - | 2 - 3 | 393 | 5214534 | 103 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC0X002313.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR0X000982.hg.1 | TC0X002313.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X000982.hg.1 | ENST00000318636.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X000982.hg.1 | ENST00000454127.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X000982.hg.1 | NM_007220.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011277.hg.1 | ENST00000318636.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011277.hg.1 | ENST00000318636.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011277.hg.1 | ENST00000318636.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011277.hg.1 | ENST00000318636.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011277.hg.1 | ENST00000318636.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011277.hg.1 | ENST00000318636.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011277.hg.1 | ENST00000318636.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011277.hg.1 | ENST00000318636.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011277.hg.1 | ENST00000318636.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011277.hg.1 | ENST00000318636.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011277.hg.1 | ENST00000318636.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011277.hg.1 | ENST00000318636.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011277.hg.1 | ENST00000318636.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011277.hg.1 | ENST00000318636.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011277.hg.1 | ENST00000318636.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011277.hg.1 | ENST00000380313.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011277.hg.1 | ENST00000380313.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011277.hg.1 | ENST00000380313.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011277.hg.1 | ENST00000380313.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011277.hg.1 | ENST00000380319.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011277.hg.1 | ENST00000380319.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011277.hg.1 | ENST00000380319.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011277.hg.1 | ENST00000380319.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011277.hg.1 | ENST00000380319.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011277.hg.1 | ENST00000380319.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011277.hg.1 | ENST00000454127.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011277.hg.1 | ENST00000454127.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011277.hg.1 | ENST00000454127.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011277.hg.1 | ENST00000454127.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011277.hg.1 | ENST00000454127.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011277.hg.1 | ENST00000454127.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011277.hg.1 | ENST00000454127.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011277.hg.1 | ENST00000454127.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011277.hg.1 | ENST00000454127.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011277.hg.1 | ENST00000454127.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011277.hg.1 | ENST00000454127.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011277.hg.1 | ENST00000474624.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011277.hg.1 | ENST00000474624.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011277.hg.1 | ENST00000474624.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011277.hg.1 | ENST00000474624.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011277.hg.1 | ENST00000474624.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011277.hg.1 | ENST00000478341.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011277.hg.1 | ENST00000478341.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011277.hg.1 | ENST00000478341.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011277.hg.1 | ENST00000478923.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011277.hg.1 | ENST00000478923.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011277.hg.1 | ENST00000478923.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011277.hg.1 | ENST00000478923.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011277.hg.1 | ENST00000478923.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011277.hg.1 | ENST00000479740.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011277.hg.1 | ENST00000479740.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011277.hg.1 | ENST00000479740.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011277.hg.1 | ENST00000479740.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011277.hg.1 | ENST00000479740.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011277.hg.1 | ENST00000496188.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011277.hg.1 | ENST00000496188.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011277.hg.1 | ENST00000496188.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011277.hg.1 | ENST00000496188.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011277.hg.1 | NM_007220.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011277.hg.1 | NM_007220.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011277.hg.1 | NM_007220.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011277.hg.1 | NM_007220.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011277.hg.1 | NM_007220.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011277.hg.1 | NM_007220.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011277.hg.1 | NM_007220.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011277.hg.1 | NM_007220.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011277.hg.1 | NM_007220.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011277.hg.1 | NM_007220.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011277.hg.1 | NM_007220.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011277.hg.1 | NM_007220.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011277.hg.1 | NM_007220.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011277.hg.1 | NM_007220.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00011277.hg.1 | ENST00000318636.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011277.hg.1 | ENST00000318636.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011277.hg.1 | ENST00000318636.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011277.hg.1 | ENST00000318636.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011277.hg.1 | ENST00000318636.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011277.hg.1 | ENST00000318636.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011277.hg.1 | ENST00000318636.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011277.hg.1 | ENST00000318636.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011277.hg.1 | ENST00000318636.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011277.hg.1 | ENST00000318636.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011277.hg.1 | ENST00000318636.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011277.hg.1 | ENST00000318636.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011277.hg.1 | ENST00000318636.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011277.hg.1 | ENST00000318636.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011277.hg.1 | ENST00000318636.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011277.hg.1 | ENST00000380313.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011277.hg.1 | ENST00000380313.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011277.hg.1 | ENST00000380313.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011277.hg.1 | ENST00000380313.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011277.hg.1 | ENST00000380319.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011277.hg.1 | ENST00000380319.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011277.hg.1 | ENST00000380319.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011277.hg.1 | ENST00000380319.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011277.hg.1 | ENST00000380319.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011277.hg.1 | ENST00000380319.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011277.hg.1 | ENST00000454127.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011277.hg.1 | ENST00000454127.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011277.hg.1 | ENST00000454127.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011277.hg.1 | ENST00000454127.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011277.hg.1 | ENST00000454127.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011277.hg.1 | ENST00000454127.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011277.hg.1 | ENST00000454127.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011277.hg.1 | ENST00000454127.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011277.hg.1 | ENST00000454127.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011277.hg.1 | ENST00000454127.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011277.hg.1 | ENST00000454127.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011277.hg.1 | ENST00000474624.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011277.hg.1 | ENST00000474624.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011277.hg.1 | ENST00000474624.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011277.hg.1 | ENST00000474624.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011277.hg.1 | ENST00000474624.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011277.hg.1 | ENST00000478341.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011277.hg.1 | ENST00000478341.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011277.hg.1 | ENST00000478341.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011277.hg.1 | ENST00000478923.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011277.hg.1 | ENST00000478923.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011277.hg.1 | ENST00000478923.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011277.hg.1 | ENST00000478923.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011277.hg.1 | ENST00000478923.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011277.hg.1 | ENST00000479740.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011277.hg.1 | ENST00000479740.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011277.hg.1 | ENST00000479740.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011277.hg.1 | ENST00000479740.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011277.hg.1 | ENST00000479740.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011277.hg.1 | ENST00000496188.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011277.hg.1 | ENST00000496188.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011277.hg.1 | ENST00000496188.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011277.hg.1 | ENST00000496188.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011277.hg.1 | NM_007220.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00011277.hg.1 | NM_007220.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00011277.hg.1 | NM_007220.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00011277.hg.1 | NM_007220.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00011277.hg.1 | NM_007220.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00011277.hg.1 | NM_007220.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00011277.hg.1 | NM_007220.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00011277.hg.1 | NM_007220.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00011277.hg.1 | NM_007220.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00011277.hg.1 | NM_007220.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00011277.hg.1 | NM_007220.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00011277.hg.1 | NM_007220.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00011277.hg.1 | NM_007220.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00011277.hg.1 | NM_007220.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00177290.hg.1 | ENST00000318636.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00177290.hg.1 | ENST00000454127.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00177290.hg.1 | ENST00000479740.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00177290.hg.1 | NM_007220.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00177290.hg.1 | ENST00000318636.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00177290.hg.1 | ENST00000454127.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00177290.hg.1 | ENST00000479740.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00177290.hg.1 | NM_007220.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00177290.hg.1 | TC0X00011277.hg.1 | Clariom D TC |

CA5B
Genomic Map