| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix HT HG-U133+ PM Array Plate |
1559352_PM_a_at |
- |
- |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17012385.hg.1 |
ENST00000500627.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17012385.hg.1 |
NR_015454.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17012385.hg.1 |
TC17000940.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17012383.hg.1 |
ENST00000500627.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17012383.hg.1 |
NR_015454.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17012383.hg.1 |
TC17000940.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17012384.hg.1 |
ENST00000500627.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17012384.hg.1 |
NR_015454.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17012384.hg.1 |
TC17000940.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17012386.hg.1 |
ENST00000500627.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17012386.hg.1 |
NR_015454.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17012386.hg.1 |
TC17000940.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17012387.hg.1 |
ENST00000500627.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17012387.hg.1 |
NR_015454.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17012387.hg.1 |
TC17000940.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700009210.hg.1 |
ENST00000582106.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700009210.hg.1 |
ENST00000582106.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700009210.hg.1 |
ENST00000582106.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700009210.hg.1 |
ENST00000582106.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700009210.hg.1 |
ENST00000582106.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700009210.hg.1 |
ENST00000582106.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700009210.hg.1 |
ENST00000582106.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700009210.hg.1 |
ENST00000582106.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700009210.hg.1 |
ENST00000583492.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700009210.hg.1 |
ENST00000583492.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700009210.hg.1 |
ENST00000583492.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700009210.hg.1 |
NR_015454.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700009210.hg.1 |
NR_015454.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700009210.hg.1 |
NR_015454.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700009210.hg.1 |
NR_015454.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700009210.hg.1 |
NR_015454.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700009210.hg.1 |
NR_015454.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700009210.hg.1 |
NR_015454.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700009210.hg.1 |
NR_015454.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700169877.hg.1 |
ENST00000582106.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700169877.hg.1 |
NR_015454.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700169877.hg.1 |
ENST00000582106.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700169877.hg.1 |
NR_015454.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700169877.hg.1 |
TC1700009210.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1700169882.hg.1 |
ENST00000582106.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700169882.hg.1 |
NR_015454.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700169882.hg.1 |
ENST00000582106.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700169882.hg.1 |
NR_015454.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700169882.hg.1 |
TC1700009210.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC1700009210.hg.1 |
ENST00000582106.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700009210.hg.1 |
ENST00000582106.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700009210.hg.1 |
ENST00000582106.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700009210.hg.1 |
ENST00000582106.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700009210.hg.1 |
ENST00000582106.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700009210.hg.1 |
ENST00000582106.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700009210.hg.1 |
ENST00000582106.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700009210.hg.1 |
ENST00000582106.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700009210.hg.1 |
ENST00000583492.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700009210.hg.1 |
ENST00000583492.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700009210.hg.1 |
ENST00000583492.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700009210.hg.1 |
NR_015454.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700009210.hg.1 |
NR_015454.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700009210.hg.1 |
NR_015454.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700009210.hg.1 |
NR_015454.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700009210.hg.1 |
NR_015454.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700009210.hg.1 |
NR_015454.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700009210.hg.1 |
NR_015454.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700009210.hg.1 |
NR_015454.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700169878.hg.1 |
ENST00000582106.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700169878.hg.1 |
NR_015454.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700169878.hg.1 |
ENST00000582106.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700169878.hg.1 |
NR_015454.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700169878.hg.1 |
TC1700009210.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1700169881.hg.1 |
ENST00000582106.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700169881.hg.1 |
ENST00000583492.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700169881.hg.1 |
NR_015454.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700169881.hg.1 |
ENST00000582106.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700169881.hg.1 |
ENST00000583492.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700169881.hg.1 |
NR_015454.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700169881.hg.1 |
TC1700009210.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1700169879.hg.1 |
ENST00000582106.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700169879.hg.1 |
ENST00000583492.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700169879.hg.1 |
NR_015454.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700169879.hg.1 |
ENST00000582106.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700169879.hg.1 |
ENST00000583492.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700169879.hg.1 |
NR_015454.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700169879.hg.1 |
TC1700009210.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1700169884.hg.1 |
ENST00000582106.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700169884.hg.1 |
NR_015454.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700169884.hg.1 |
ENST00000582106.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700169884.hg.1 |
NR_015454.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700169884.hg.1 |
TC1700009210.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1700169876.hg.1 |
ENST00000582106.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700169876.hg.1 |
NR_015454.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700169876.hg.1 |
ENST00000582106.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700169876.hg.1 |
NR_015454.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700169876.hg.1 |
TC1700009210.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1700169883.hg.1 |
ENST00000582106.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700169883.hg.1 |
ENST00000583492.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700169883.hg.1 |
NR_015454.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700169883.hg.1 |
ENST00000582106.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700169883.hg.1 |
ENST00000583492.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700169883.hg.1 |
NR_015454.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700169883.hg.1 |
TC1700009210.hg.1 |
Clariom D TC |
| Human Genome U133 Plus 2.0 Array |
1559352_a_at |
- |
- |
| Human Genome U133 Plus 2.0 Array |
1559352_PM_a_at |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1559352_a_at7 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1559352_a_at11 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1559352_a_at6 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1559352_a_at10 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1559352_a_at3 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1559352_a_at4 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1559352_a_at5 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1559352_a_at2 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1559352_a_at8 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1559352_a_at1 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1559352_a_at9 |
- |
- |