Assay Details
| Gene Symbol: ERAL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_134328.1 | 7 - 8 | 844 | 100 | ||
| GenBank mRNA | AF082658.1 | - | 7 - 8 | 785 | 100 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

ERAL1
Genomic Map