Assay Details
| Gene Symbol: FAM150B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002919.2 | NP_001002919.2 | 2 - 3 | 602 | 138 | |
| XM_005264689.4 | XP_005264746.1 | 2 - 3 | 615 | 138 | ||
| XM_006711879.3 | XP_006711942.1 | 2 - 3 | 616 | 138 | ||
| XM_011510342.2 | XP_011508644.1 | 2 - 3 | 627 | 138 | ||
| GenBank mRNA | BC033490.1 | - | 2 - 3 | 476 | 138 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC02001496.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
positive regulation of neuron projection development
positive regulation of protein tyrosine kinase activity
positive regulation of ERK1 and ERK2 cascade
positive regulation of ERK5 cascade
protein binding
receptor signaling protein tyrosine kinase activator activity
receptor tyrosine kinase binding
positive regulation of neuron projection development
positive regulation of protein tyrosine kinase activity
positive regulation of ERK1 and ERK2 cascade
positive regulation of ERK5 cascade
protein binding
receptor signaling protein tyrosine kinase activator activity
receptor tyrosine kinase binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR02024553.hg.1 | TC02001496.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02024553.hg.1 | ENST00000403610.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02024553.hg.1 | NM_001002919.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02024557.hg.1 | TC02001496.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02024557.hg.1 | ENST00000403610.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02024557.hg.1 | NM_001002919.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02024547.hg.1 | TC02001496.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02024547.hg.1 | ENST00000344414.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02024547.hg.1 | ENST00000401503.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02024547.hg.1 | ENST00000403610.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02024547.hg.1 | ENST00000405290.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02024547.hg.1 | ENST00000436353.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02024547.hg.1 | NM_001002919.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02024554.hg.1 | TC02001496.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02024554.hg.1 | ENST00000403610.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02024554.hg.1 | NM_001002919.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02024555.hg.1 | TC02001496.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02024555.hg.1 | ENST00000403610.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02024555.hg.1 | ENST00000436353.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02024555.hg.1 | NM_001002919.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02024558.hg.1 | TC02001496.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02024558.hg.1 | ENST00000403610.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02024558.hg.1 | NM_001002919.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011450.hg.1 | ENST00000344414.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011450.hg.1 | ENST00000344414.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011450.hg.1 | ENST00000344414.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011450.hg.1 | ENST00000344414.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011450.hg.1 | ENST00000344414.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011450.hg.1 | ENST00000401489.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011450.hg.1 | ENST00000401489.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011450.hg.1 | ENST00000401489.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011450.hg.1 | ENST00000401489.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011450.hg.1 | ENST00000401489.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011450.hg.1 | ENST00000401503.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011450.hg.1 | ENST00000401503.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011450.hg.1 | ENST00000401503.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011450.hg.1 | ENST00000401503.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011450.hg.1 | ENST00000401503.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011450.hg.1 | ENST00000401503.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011450.hg.1 | ENST00000403610.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011450.hg.1 | ENST00000403610.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011450.hg.1 | ENST00000403610.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011450.hg.1 | ENST00000403610.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011450.hg.1 | ENST00000403610.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011450.hg.1 | ENST00000403610.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011450.hg.1 | ENST00000403610.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011450.hg.1 | ENST00000403610.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011450.hg.1 | ENST00000403610.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011450.hg.1 | ENST00000403610.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011450.hg.1 | ENST00000403610.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011450.hg.1 | ENST00000405290.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011450.hg.1 | ENST00000405290.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011450.hg.1 | ENST00000405290.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011450.hg.1 | ENST00000405290.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011450.hg.1 | ENST00000405290.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011450.hg.1 | ENST00000452023.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011450.hg.1 | ENST00000452023.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011450.hg.1 | ENST00000452023.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011450.hg.1 | ENST00000452023.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011450.hg.1 | ENST00000452023.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011450.hg.1 | ENST00000452023.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011450.hg.1 | ENST00000452023.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011450.hg.1 | ENST00000452023.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011450.hg.1 | ENST00000463919.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011450.hg.1 | ENST00000463919.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011450.hg.1 | ENST00000463919.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011450.hg.1 | ENST00000463919.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011450.hg.1 | ENST00000463919.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011450.hg.1 | ENST00000619265.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011450.hg.1 | ENST00000619265.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011450.hg.1 | ENST00000619265.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011450.hg.1 | ENST00000619265.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011450.hg.1 | ENST00000619265.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011450.hg.1 | NM_001002919.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011450.hg.1 | NM_001002919.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011450.hg.1 | NM_001002919.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011450.hg.1 | NM_001002919.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011450.hg.1 | NM_001002919.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011450.hg.1 | NM_001002919.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011450.hg.1 | NM_001002919.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011450.hg.1 | NM_001002919.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011450.hg.1 | NM_001002919.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011450.hg.1 | NM_001002919.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011450.hg.1 | NM_001002919.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200183190.hg.1 | ENST00000344414.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200183190.hg.1 | ENST00000401489.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200183190.hg.1 | ENST00000401503.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200183190.hg.1 | ENST00000403610.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200183190.hg.1 | ENST00000405290.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200183190.hg.1 | ENST00000452023.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200183190.hg.1 | ENST00000463919.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200183190.hg.1 | ENST00000619265.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200183190.hg.1 | NM_001002919.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200183190.hg.1 | ENST00000344414.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200183190.hg.1 | ENST00000401489.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200183190.hg.1 | ENST00000401503.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200183190.hg.1 | ENST00000403610.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200183190.hg.1 | ENST00000405290.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200183190.hg.1 | ENST00000452023.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200183190.hg.1 | ENST00000463919.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200183190.hg.1 | ENST00000619265.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200183190.hg.1 | NM_001002919.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200183190.hg.1 | TC0200011450.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0200183197.hg.1 | ENST00000401489.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200183197.hg.1 | ENST00000403610.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200183197.hg.1 | ENST00000452023.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200183197.hg.1 | NM_001002919.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200183197.hg.1 | ENST00000401489.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200183197.hg.1 | ENST00000403610.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200183197.hg.1 | ENST00000452023.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200183197.hg.1 | NM_001002919.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200183197.hg.1 | TC0200011450.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0200183203.hg.1 | ENST00000403610.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200183203.hg.1 | ENST00000452023.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200183203.hg.1 | NM_001002919.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200183203.hg.1 | ENST00000403610.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200183203.hg.1 | ENST00000452023.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200183203.hg.1 | NM_001002919.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200183203.hg.1 | TC0200011450.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0200183200.hg.1 | ENST00000403610.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200183200.hg.1 | ENST00000452023.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200183200.hg.1 | ENST00000619265.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200183200.hg.1 | NM_001002919.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200183200.hg.1 | ENST00000403610.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200183200.hg.1 | ENST00000452023.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200183200.hg.1 | ENST00000619265.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200183200.hg.1 | NM_001002919.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200183200.hg.1 | TC0200011450.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0200011450.hg.1 | ENST00000344414.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011450.hg.1 | ENST00000344414.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011450.hg.1 | ENST00000344414.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011450.hg.1 | ENST00000344414.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011450.hg.1 | ENST00000344414.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011450.hg.1 | ENST00000401489.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011450.hg.1 | ENST00000401489.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011450.hg.1 | ENST00000401489.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011450.hg.1 | ENST00000401489.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011450.hg.1 | ENST00000401489.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011450.hg.1 | ENST00000401503.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011450.hg.1 | ENST00000401503.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011450.hg.1 | ENST00000401503.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011450.hg.1 | ENST00000401503.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011450.hg.1 | ENST00000401503.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011450.hg.1 | ENST00000401503.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011450.hg.1 | ENST00000403610.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011450.hg.1 | ENST00000403610.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011450.hg.1 | ENST00000403610.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011450.hg.1 | ENST00000403610.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011450.hg.1 | ENST00000403610.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011450.hg.1 | ENST00000403610.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011450.hg.1 | ENST00000403610.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011450.hg.1 | ENST00000403610.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011450.hg.1 | ENST00000403610.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011450.hg.1 | ENST00000403610.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011450.hg.1 | ENST00000403610.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011450.hg.1 | ENST00000405290.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011450.hg.1 | ENST00000405290.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011450.hg.1 | ENST00000405290.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011450.hg.1 | ENST00000405290.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011450.hg.1 | ENST00000405290.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011450.hg.1 | ENST00000452023.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011450.hg.1 | ENST00000452023.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011450.hg.1 | ENST00000452023.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011450.hg.1 | ENST00000452023.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011450.hg.1 | ENST00000452023.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011450.hg.1 | ENST00000452023.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011450.hg.1 | ENST00000452023.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011450.hg.1 | ENST00000452023.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011450.hg.1 | ENST00000463919.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011450.hg.1 | ENST00000463919.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011450.hg.1 | ENST00000463919.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011450.hg.1 | ENST00000463919.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011450.hg.1 | ENST00000463919.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011450.hg.1 | ENST00000619265.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011450.hg.1 | ENST00000619265.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011450.hg.1 | ENST00000619265.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011450.hg.1 | ENST00000619265.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011450.hg.1 | ENST00000619265.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011450.hg.1 | NM_001002919.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200011450.hg.1 | NM_001002919.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200011450.hg.1 | NM_001002919.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200011450.hg.1 | NM_001002919.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200011450.hg.1 | NM_001002919.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200011450.hg.1 | NM_001002919.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200011450.hg.1 | NM_001002919.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200011450.hg.1 | NM_001002919.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200011450.hg.1 | NM_001002919.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200011450.hg.1 | NM_001002919.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200011450.hg.1 | NM_001002919.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200183198.hg.1 | ENST00000403610.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200183198.hg.1 | ENST00000452023.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200183198.hg.1 | NM_001002919.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200183198.hg.1 | ENST00000403610.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200183198.hg.1 | ENST00000452023.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200183198.hg.1 | NM_001002919.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200183198.hg.1 | TC0200011450.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0200183204.hg.1 | ENST00000403610.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200183204.hg.1 | NM_001002919.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200183204.hg.1 | ENST00000403610.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200183204.hg.1 | NM_001002919.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200183204.hg.1 | TC0200011450.hg.1 | Clariom D TC |

FAM150B
Genomic Map