| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Clariom D Assay |
PSR1700147726.hg.1 |
ENST00000330494.11 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700147726.hg.1 |
ENST00000358181.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700147726.hg.1 |
ENST00000380358.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700147726.hg.1 |
ENST00000470531.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700147726.hg.1 |
ENST00000572750.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700147726.hg.1 |
NM_001005271.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700147726.hg.1 |
NM_001005273.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700147726.hg.1 |
NM_005852.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700147726.hg.1 |
ENST00000330494.11 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700147726.hg.1 |
ENST00000358181.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700147726.hg.1 |
ENST00000380358.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700147726.hg.1 |
ENST00000470531.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700147726.hg.1 |
ENST00000572750.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700147726.hg.1 |
NM_001005271.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700147726.hg.1 |
NM_001005273.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700147726.hg.1 |
NM_005852.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700147726.hg.1 |
TC1700006775.hg.1 |
Clariom D TC |