Assay Details
| Gene Symbol: ARL6IP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001313858.1 | NP_001300787.1 | 5 - 6 | 625 | 71 | |
| NM_015161.2 | NP_055976.1 | 5 - 6 | 701 | 71 | ||
| GenBank mRNA | AK291137.1 | - | 5 - 6 | 567 | 71 | |
| AK300825.1 | - | 6 - 7 | 509 | 71 | ||
| AK301636.1 | - | 5 - 6 | 625 | 71 | ||
| AK307315.1 | - | 4 - 5 | 452 | 71 | ||
| AK308092.1 | - | 6 - 7 | 485 | 71 | ||
| AK308744.1 | - | 2 - 3 | 147 | 71 | ||
| BC010281.1 | - | 5 - 6 | 558 | 3048642 | 71 | |
| BX648446.1 | - | 6 - 7 | 488 | 71 | ||
| D31885.1 | - | 5 - 6 | 558 | 71 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
positive regulation of L-glutamate transport
cotranslational protein targeting to membrane
apoptotic process
negative regulation of apoptotic process
negative regulation of cysteine-type endopeptidase activity involved in apoptotic process
endoplasmic reticulum tubular network assembly
endoplasmic reticulum tubular network membrane organization
protein binding
positive regulation of L-glutamate transport
cotranslational protein targeting to membrane
apoptotic process
negative regulation of apoptotic process
negative regulation of cysteine-type endopeptidase activity involved in apoptotic process
endoplasmic reticulum tubular network assembly
endoplasmic reticulum tubular network membrane organization
protein binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1600011494.hg.1 | ENST00000304414.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011494.hg.1 | ENST00000304414.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011494.hg.1 | ENST00000304414.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011494.hg.1 | ENST00000304414.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011494.hg.1 | ENST00000304414.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011494.hg.1 | ENST00000304414.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011494.hg.1 | ENST00000304414.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011494.hg.1 | ENST00000304414.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011494.hg.1 | ENST00000304414.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011494.hg.1 | ENST00000304414.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011494.hg.1 | ENST00000304414.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011494.hg.1 | ENST00000304414.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011494.hg.1 | ENST00000304414.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011494.hg.1 | ENST00000546206.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011494.hg.1 | ENST00000546206.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011494.hg.1 | ENST00000546206.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011494.hg.1 | ENST00000546206.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011494.hg.1 | ENST00000546206.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011494.hg.1 | ENST00000546206.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011494.hg.1 | ENST00000546206.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011494.hg.1 | ENST00000546206.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011494.hg.1 | ENST00000546206.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011494.hg.1 | ENST00000562234.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011494.hg.1 | ENST00000562234.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011494.hg.1 | ENST00000562234.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011494.hg.1 | ENST00000562234.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011494.hg.1 | ENST00000562234.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011494.hg.1 | ENST00000562819.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011494.hg.1 | ENST00000562819.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011494.hg.1 | ENST00000562819.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011494.hg.1 | ENST00000562819.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011494.hg.1 | ENST00000562819.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011494.hg.1 | ENST00000563861.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011494.hg.1 | ENST00000563861.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011494.hg.1 | ENST00000563861.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011494.hg.1 | ENST00000563861.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011494.hg.1 | ENST00000563861.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011494.hg.1 | ENST00000563861.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011494.hg.1 | ENST00000563861.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011494.hg.1 | ENST00000563861.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011494.hg.1 | ENST00000563861.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011494.hg.1 | ENST00000563861.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011494.hg.1 | ENST00000566391.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011494.hg.1 | ENST00000566391.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011494.hg.1 | ENST00000566391.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011494.hg.1 | ENST00000566391.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011494.hg.1 | ENST00000567969.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011494.hg.1 | ENST00000567969.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011494.hg.1 | ENST00000567969.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011494.hg.1 | ENST00000567969.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011494.hg.1 | ENST00000567969.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011494.hg.1 | ENST00000567969.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011494.hg.1 | ENST00000569976.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011494.hg.1 | ENST00000569976.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011494.hg.1 | ENST00000569976.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011494.hg.1 | ENST00000569976.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011494.hg.1 | ENST00000569976.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011494.hg.1 | ENST00000569976.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011494.hg.1 | ENST00000569976.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011494.hg.1 | NM_015161.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011494.hg.1 | NM_015161.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011494.hg.1 | NM_015161.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011494.hg.1 | NM_015161.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011494.hg.1 | NM_015161.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011494.hg.1 | NM_015161.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011494.hg.1 | NM_015161.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011494.hg.1 | NM_015161.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011494.hg.1 | NM_015161.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011494.hg.1 | NM_015161.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011494.hg.1 | NM_015161.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011494.hg.1 | NM_015161.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600192927.hg.1 | ENST00000304414.11 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600192927.hg.1 | ENST00000546206.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600192927.hg.1 | ENST00000562819.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600192927.hg.1 | ENST00000563861.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600192927.hg.1 | NM_015161.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600192927.hg.1 | ENST00000304414.11 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600192927.hg.1 | ENST00000546206.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600192927.hg.1 | ENST00000562819.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600192927.hg.1 | ENST00000563861.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600192927.hg.1 | NM_015161.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600192927.hg.1 | TC1600011494.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1600011494.hg.1 | ENST00000304414.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011494.hg.1 | ENST00000304414.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011494.hg.1 | ENST00000304414.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011494.hg.1 | ENST00000304414.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011494.hg.1 | ENST00000304414.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011494.hg.1 | ENST00000304414.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011494.hg.1 | ENST00000304414.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011494.hg.1 | ENST00000304414.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011494.hg.1 | ENST00000304414.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011494.hg.1 | ENST00000304414.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011494.hg.1 | ENST00000304414.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011494.hg.1 | ENST00000304414.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011494.hg.1 | ENST00000304414.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011494.hg.1 | ENST00000546206.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011494.hg.1 | ENST00000546206.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011494.hg.1 | ENST00000546206.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011494.hg.1 | ENST00000546206.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011494.hg.1 | ENST00000546206.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011494.hg.1 | ENST00000546206.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011494.hg.1 | ENST00000546206.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011494.hg.1 | ENST00000546206.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011494.hg.1 | ENST00000546206.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011494.hg.1 | ENST00000562234.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011494.hg.1 | ENST00000562234.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011494.hg.1 | ENST00000562234.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011494.hg.1 | ENST00000562234.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011494.hg.1 | ENST00000562234.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011494.hg.1 | ENST00000562819.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011494.hg.1 | ENST00000562819.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011494.hg.1 | ENST00000562819.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011494.hg.1 | ENST00000562819.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011494.hg.1 | ENST00000562819.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011494.hg.1 | ENST00000563861.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011494.hg.1 | ENST00000563861.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011494.hg.1 | ENST00000563861.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011494.hg.1 | ENST00000563861.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011494.hg.1 | ENST00000563861.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011494.hg.1 | ENST00000563861.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011494.hg.1 | ENST00000563861.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011494.hg.1 | ENST00000563861.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011494.hg.1 | ENST00000563861.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011494.hg.1 | ENST00000563861.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011494.hg.1 | ENST00000566391.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011494.hg.1 | ENST00000566391.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011494.hg.1 | ENST00000566391.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011494.hg.1 | ENST00000566391.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011494.hg.1 | ENST00000567969.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011494.hg.1 | ENST00000567969.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011494.hg.1 | ENST00000567969.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011494.hg.1 | ENST00000567969.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011494.hg.1 | ENST00000567969.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011494.hg.1 | ENST00000567969.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011494.hg.1 | ENST00000569976.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011494.hg.1 | ENST00000569976.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011494.hg.1 | ENST00000569976.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011494.hg.1 | ENST00000569976.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011494.hg.1 | ENST00000569976.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011494.hg.1 | ENST00000569976.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011494.hg.1 | ENST00000569976.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011494.hg.1 | NM_015161.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011494.hg.1 | NM_015161.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011494.hg.1 | NM_015161.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011494.hg.1 | NM_015161.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011494.hg.1 | NM_015161.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011494.hg.1 | NM_015161.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011494.hg.1 | NM_015161.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011494.hg.1 | NM_015161.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011494.hg.1 | NM_015161.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011494.hg.1 | NM_015161.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011494.hg.1 | NM_015161.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011494.hg.1 | NM_015161.2 | Refseq |

ARL6IP1
Genomic Map