Assay Details
| Gene Symbol: PVR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001135769.2 | NP_001129241.1 | 5 - 6 | 1289 | 79 | |
| GenBank mRNA | DA849917.1 | - | 1 - 2 | 81 | 79 | |
More Information
Set Membership: |
|

PVR
Genomic Map