Assay Details
| Gene Symbol: SHC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001130040.1 | NP_001123512.1 | 9 - 10 | 1471 | 87 | |
| NM_003029.4 | NP_003020.2 | 10 - 11 | 1159 | 87 | ||
| XM_011509892.2 | XP_011508194.1 | 9 - 10 | 2711 | 87 | ||
| XM_011509894.2 | XP_011508196.1 | 8 - 9 | 2661 | 87 | ||
| XM_011509897.1 | XP_011508199.1 | 9 - 10 | 1096 | 87 | ||
| XM_017002081.1 | XP_016857570.1 | 8 - 9 | 2629 | 87 | ||
| GenBank mRNA | AK096169.1 | - | 9 - 10 | 1454 | 87 | |
| AK130014.1 | - | 8 - 9 | 804 | 87 | ||
| AK292143.1 | - | 10 - 11 | 1159 | 87 | ||
| AK296786.1 | - | 10 - 11 | 1282 | 87 | ||
| AK315842.1 | - | 10 - 11 | 1070 | 87 | ||
| BC014158.2 | - | 10 - 11 | 1040 | 4541216 | 87 | |
| BX647149.1 | - | 9 - 10 | 1471 | 87 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

SHC1
Genomic Map