Assay Details
| Gene Symbol: ZKSCAN7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001288590.1 | NP_001275519.1 | 3 - 4 | 1019 | 74 | |
| NM_001288591.1 | NP_001275520.1 | 3 - 4 | 1019 | 74 | ||
| NM_001288592.1 | NP_001275521.1 | 2 - 3 | 478 | 74 | ||
| NM_018651.3 | NP_061121.2 | 3 - 4 | 1056 | 74 | ||
| NM_025169.2 | NP_079445.1 | 3 - 4 | 1056 | 74 | ||
| GenBank mRNA | AF154846.1 | - | 2 - 3 | 672 | 74 | |
| AK291285.1 | - | 3 - 4 | 1001 | 74 | ||
| AY280798.1 | - | 3 - 4 | 1010 | 74 | ||
| BC010555.1 | - | 3 - 4 | 974 | 74 | ||
| BC063020.1 | - | 3 - 4 | 1008 | 74 | ||
| BC120982.1 | - | 3 - 4 | 748 | 74 | ||
| BC120983.1 | - | 3 - 4 | 787 | 74 | ||
| BC127639.1 | - | 3 - 4 | 787 | 74 | ||
| BC127640.1 | - | 3 - 4 | 787 | 74 | ||
| BC148260.1 | - | 2 - 3 | 209 | 74 | ||
More Information
Set Membership: |
|

ZKSCAN7
Genomic Map