| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02026752.hg.1 |
TC02001667.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02026752.hg.1 |
ENST00000272371.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02026752.hg.1 |
ENST00000338581.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02026752.hg.1 |
ENST00000339598.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02026752.hg.1 |
ENST00000402415.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02026752.hg.1 |
ENST00000403946.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02026752.hg.1 |
NM_004802.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02026752.hg.1 |
NM_194248.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02026752.hg.1 |
NM_194322.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02026752.hg.1 |
NM_194323.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200187640.hg.1 |
ENST00000272371.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200187640.hg.1 |
ENST00000338581.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200187640.hg.1 |
ENST00000339598.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200187640.hg.1 |
ENST00000402415.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200187640.hg.1 |
ENST00000403946.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200187640.hg.1 |
ENST00000426958.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200187640.hg.1 |
NM_001287489.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200187640.hg.1 |
NM_004802.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200187640.hg.1 |
NM_194248.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200187640.hg.1 |
NM_194322.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200187640.hg.1 |
NM_194323.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200187640.hg.1 |
ENST00000272371.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200187640.hg.1 |
ENST00000338581.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200187640.hg.1 |
ENST00000339598.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200187640.hg.1 |
ENST00000402415.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200187640.hg.1 |
ENST00000403946.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200187640.hg.1 |
ENST00000426958.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200187640.hg.1 |
NM_001287489.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200187640.hg.1 |
NM_004802.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200187640.hg.1 |
NM_194248.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200187640.hg.1 |
NM_194322.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200187640.hg.1 |
NM_194323.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200187640.hg.1 |
TC0200012038.hg.1 |
Clariom D TC |