Assay Details
| Gene Symbol: OSMR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001323504.1 | NP_001310433.1 | 1 - 2 | 386 | 104 | |
| NM_003999.2 | NP_003990.1 | 1 - 2 | 386 | 104 | ||
| XM_005248384.1 | XP_005248441.1 | 1 - 2 | 373 | 104 | ||
| XM_005248386.2 | XP_005248443.1 | 1 - 2 | 373 | 104 | ||
| XM_005248387.2 | XP_005248444.1 | 1 - 2 | 373 | 104 | ||
| XM_011514161.2 | XP_011512463.1 | 1 - 2 | 373 | 104 | ||
| XM_017010019.1 | XP_016865508.1 | 1 - 2 | 373 | 104 | ||
| GenBank mRNA | AK304016.1 | - | 1 - 2 | 340 | 104 | |
| BC125209.1 | - | 1 - 2 | 156 | 40123501 | 104 | |
| BC125210.1 | - | 1 - 2 | 156 | 40123502 | 104 | |
| DR006275.1 | - | 1 - 2 | 393 | 104 | ||
| U60805.1 | - | 1 - 2 | 351 | 104 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC05000179.hg.1
Additional Information:
This assay is in the 5'UTR region of the AK304016.1,BC125209.1,BC125210.1,NM_001323504.1,NM_003999.2,U60805.1,XM_005248384.1,XM_005248386.2,XM_005248387.2,XM_011514161.2,XM_017010019.1 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR05002093.hg.1 | TC05000179.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR05002093.hg.1 | ENST00000274276.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05002093.hg.1 | NM_003999.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR05002092.hg.1 | TC05000179.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR05002092.hg.1 | ENST00000274276.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05002092.hg.1 | ENST00000502536.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05002092.hg.1 | NM_001168355.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR05002092.hg.1 | NM_003999.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR05002094.hg.1 | TC05000179.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR05002094.hg.1 | ENST00000274276.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05002094.hg.1 | NM_003999.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007201.hg.1 | ENST00000274276.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007201.hg.1 | ENST00000274276.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007201.hg.1 | ENST00000274276.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007201.hg.1 | ENST00000274276.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007201.hg.1 | ENST00000274276.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007201.hg.1 | ENST00000274276.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007201.hg.1 | ENST00000274276.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007201.hg.1 | ENST00000274276.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007201.hg.1 | ENST00000274276.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007201.hg.1 | ENST00000274276.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007201.hg.1 | ENST00000274276.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007201.hg.1 | ENST00000274276.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007201.hg.1 | ENST00000274276.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007201.hg.1 | ENST00000274276.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007201.hg.1 | ENST00000274276.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007201.hg.1 | ENST00000274276.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007201.hg.1 | ENST00000274276.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007201.hg.1 | ENST00000274276.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007201.hg.1 | ENST00000274276.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007201.hg.1 | ENST00000274276.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007201.hg.1 | ENST00000274276.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007201.hg.1 | ENST00000274276.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007201.hg.1 | ENST00000274276.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007201.hg.1 | ENST00000502536.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007201.hg.1 | ENST00000502536.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007201.hg.1 | ENST00000502536.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007201.hg.1 | ENST00000502536.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007201.hg.1 | ENST00000502536.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007201.hg.1 | ENST00000502536.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007201.hg.1 | ENST00000502536.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007201.hg.1 | ENST00000502536.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007201.hg.1 | ENST00000502536.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007201.hg.1 | ENST00000502536.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007201.hg.1 | ENST00000508882.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007201.hg.1 | ENST00000508882.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007201.hg.1 | ENST00000508882.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007201.hg.1 | ENST00000509237.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007201.hg.1 | ENST00000509237.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007201.hg.1 | ENST00000509237.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007201.hg.1 | ENST00000509237.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007201.hg.1 | ENST00000513831.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007201.hg.1 | ENST00000513831.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007201.hg.1 | ENST00000513831.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007201.hg.1 | ENST00000513831.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007201.hg.1 | NM_001168355.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007201.hg.1 | NM_001168355.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007201.hg.1 | NM_001168355.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007201.hg.1 | NM_001168355.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007201.hg.1 | NM_001168355.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007201.hg.1 | NM_001168355.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007201.hg.1 | NM_001168355.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007201.hg.1 | NM_001168355.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007201.hg.1 | NM_001168355.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007201.hg.1 | NM_001168355.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007201.hg.1 | NM_003999.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007201.hg.1 | NM_003999.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007201.hg.1 | NM_003999.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007201.hg.1 | NM_003999.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007201.hg.1 | NM_003999.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007201.hg.1 | NM_003999.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007201.hg.1 | NM_003999.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007201.hg.1 | NM_003999.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007201.hg.1 | NM_003999.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007201.hg.1 | NM_003999.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007201.hg.1 | NM_003999.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007201.hg.1 | NM_003999.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007201.hg.1 | NM_003999.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007201.hg.1 | NM_003999.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007201.hg.1 | NM_003999.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007201.hg.1 | NM_003999.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007201.hg.1 | NM_003999.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007201.hg.1 | NM_003999.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007201.hg.1 | NM_003999.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007201.hg.1 | NM_003999.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007201.hg.1 | NM_003999.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007201.hg.1 | NM_003999.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007201.hg.1 | NM_003999.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR05002095.hg.1 | TC05000179.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR05002095.hg.1 | ENST00000274276.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05002095.hg.1 | ENST00000502536.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05002095.hg.1 | NM_001168355.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR05002095.hg.1 | NM_003999.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007201.hg.1 | ENST00000274276.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007201.hg.1 | ENST00000274276.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007201.hg.1 | ENST00000274276.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007201.hg.1 | ENST00000274276.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007201.hg.1 | ENST00000274276.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007201.hg.1 | ENST00000274276.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007201.hg.1 | ENST00000274276.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007201.hg.1 | ENST00000274276.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007201.hg.1 | ENST00000274276.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007201.hg.1 | ENST00000274276.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007201.hg.1 | ENST00000274276.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007201.hg.1 | ENST00000274276.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007201.hg.1 | ENST00000274276.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007201.hg.1 | ENST00000274276.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007201.hg.1 | ENST00000274276.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007201.hg.1 | ENST00000274276.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007201.hg.1 | ENST00000274276.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007201.hg.1 | ENST00000274276.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007201.hg.1 | ENST00000274276.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007201.hg.1 | ENST00000274276.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007201.hg.1 | ENST00000274276.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007201.hg.1 | ENST00000274276.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007201.hg.1 | ENST00000274276.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007201.hg.1 | ENST00000502536.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007201.hg.1 | ENST00000502536.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007201.hg.1 | ENST00000502536.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007201.hg.1 | ENST00000502536.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007201.hg.1 | ENST00000502536.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007201.hg.1 | ENST00000502536.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007201.hg.1 | ENST00000502536.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007201.hg.1 | ENST00000502536.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007201.hg.1 | ENST00000502536.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007201.hg.1 | ENST00000502536.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007201.hg.1 | ENST00000508882.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007201.hg.1 | ENST00000508882.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007201.hg.1 | ENST00000508882.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007201.hg.1 | ENST00000509237.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007201.hg.1 | ENST00000509237.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007201.hg.1 | ENST00000509237.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007201.hg.1 | ENST00000509237.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007201.hg.1 | ENST00000513831.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007201.hg.1 | ENST00000513831.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007201.hg.1 | ENST00000513831.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007201.hg.1 | ENST00000513831.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007201.hg.1 | NM_001168355.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007201.hg.1 | NM_001168355.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007201.hg.1 | NM_001168355.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007201.hg.1 | NM_001168355.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007201.hg.1 | NM_001168355.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007201.hg.1 | NM_001168355.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007201.hg.1 | NM_001168355.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007201.hg.1 | NM_001168355.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007201.hg.1 | NM_001168355.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007201.hg.1 | NM_001168355.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007201.hg.1 | NM_003999.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007201.hg.1 | NM_003999.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007201.hg.1 | NM_003999.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007201.hg.1 | NM_003999.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007201.hg.1 | NM_003999.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007201.hg.1 | NM_003999.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007201.hg.1 | NM_003999.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007201.hg.1 | NM_003999.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007201.hg.1 | NM_003999.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007201.hg.1 | NM_003999.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007201.hg.1 | NM_003999.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007201.hg.1 | NM_003999.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007201.hg.1 | NM_003999.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007201.hg.1 | NM_003999.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007201.hg.1 | NM_003999.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007201.hg.1 | NM_003999.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007201.hg.1 | NM_003999.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007201.hg.1 | NM_003999.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007201.hg.1 | NM_003999.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007201.hg.1 | NM_003999.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007201.hg.1 | NM_003999.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007201.hg.1 | NM_003999.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007201.hg.1 | NM_003999.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500148720.hg.1 | ENST00000274276.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500148720.hg.1 | NM_003999.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500148720.hg.1 | ENST00000274276.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500148720.hg.1 | NM_003999.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500148720.hg.1 | TC0500007201.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0500148719.hg.1 | ENST00000274276.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500148719.hg.1 | NM_003999.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500148719.hg.1 | ENST00000274276.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500148719.hg.1 | NM_003999.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500148719.hg.1 | TC0500007201.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0500148722.hg.1 | ENST00000274276.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500148722.hg.1 | ENST00000502536.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500148722.hg.1 | NM_001168355.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500148722.hg.1 | NM_003999.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500148722.hg.1 | ENST00000274276.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500148722.hg.1 | ENST00000502536.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500148722.hg.1 | NM_001168355.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500148722.hg.1 | NM_003999.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500148722.hg.1 | TC0500007201.hg.1 | Clariom D TC |

OSMR
Genomic Map