Assay Details
| Gene Symbol: MYH14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077186.1 | NP_001070654.1 | 5 - 6 | 747 | 70 | |
| NM_001145809.1 | NP_001139281.1 | 5 - 6 | 747 | 70 | ||
| XM_006723386.3 | XP_006723449.1 | 5 - 6 | 808 | 70 | ||
| XM_011527320.2 | XP_011525622.1 | 5 - 6 | 825 | 70 | ||
| XM_011527323.2 | XP_011525625.1 | 5 - 6 | 815 | 70 | ||
| GenBank mRNA | AB290169.1 | - | 4 - 5 | 717 | 70 | |
| AK125082.1 | - | 5 - 6 | 826 | 70 | ||
| AK307783.1 | - | 5 - 6 | 730 | 70 | ||
| L29142.1 | - | 1 - 2 | 85 | 70 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK125082.1 transcripts.
Set Membership: |
|

MYH14
Genomic Map