Assay Details
| Gene Symbol: PLEKHA5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001256470.1 | NP_001243399.1 | 24 - 25 | 3008 | 143 | |
| NM_001256787.1 | NP_001243716.1 | 18 - 19 | 2564 | 143 | ||
| XM_005253400.1 | XP_005253457.1 | 19 - 20 | 2659 | 143 | ||
| XM_006719091.1 | XP_006719154.1 | 20 - 21 | 2713 | 143 | ||
| XM_006719093.1 | XP_006719156.1 | 18 - 19 | 2575 | 143 | ||
| XM_006719096.1 | XP_006719159.1 | 23 - 24 | 2950 | 143 | ||
| XM_011520714.1 | XP_011519016.1 | 22 - 23 | 2914 | 143 | ||
| XM_011520715.1 | XP_011519017.1 | 21 - 22 | 2833 | 143 | ||
| XM_017019495.1 | XP_016874984.1 | 23 - 24 | 2935 | 143 | ||
| XM_017019497.1 | XP_016874986.1 | 21 - 22 | 2764 | 143 | ||
| GenBank mRNA | AB642244.1 | - | 24 - 25 | 2974 | 143 | |
| AK056054.1 | - | 12 - 13 | 1525 | 143 | ||
| AK294739.1 | - | 19 - 20 | 2565 | 143 | ||
| AK296277.1 | - | 17 - 18 | 2476 | 143 | ||
| AL834259.1 | - | 5 - 6 | 557 | 143 | ||
More Information
Set Membership: |
|

PLEKHA5
Genomic Map