Assay Details
Gene Symbol: KLRF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_120305.1 | 2 - 3 | 248 | 110 | ||
GenBank mRNA | AF267244.1 | - | 2 - 3 | 248 | 110 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1200006773.hg.1 | ENST00000279545.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006773.hg.1 | ENST00000279545.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006773.hg.1 | ENST00000279545.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006773.hg.1 | ENST00000279545.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006773.hg.1 | ENST00000279545.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006773.hg.1 | ENST00000279545.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006773.hg.1 | ENST00000279545.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006773.hg.1 | ENST00000279545.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006773.hg.1 | ENST00000354855.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006773.hg.1 | ENST00000354855.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006773.hg.1 | ENST00000354855.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006773.hg.1 | ENST00000354855.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006773.hg.1 | ENST00000354855.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006773.hg.1 | ENST00000354855.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006773.hg.1 | ENST00000537723.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006773.hg.1 | ENST00000537723.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006773.hg.1 | ENST00000537723.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006773.hg.1 | ENST00000537723.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006773.hg.1 | ENST00000537723.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006773.hg.1 | ENST00000545196.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006773.hg.1 | ENST00000545196.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006773.hg.1 | ENST00000545196.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006773.hg.1 | ENST00000545196.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006773.hg.1 | ENST00000545196.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006773.hg.1 | ENST00000545196.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006773.hg.1 | ENST00000612321.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006773.hg.1 | ENST00000612321.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006773.hg.1 | ENST00000612321.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006773.hg.1 | ENST00000612321.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006773.hg.1 | ENST00000612321.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006773.hg.1 | ENST00000612321.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006773.hg.1 | ENST00000612321.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006773.hg.1 | ENST00000616259.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006773.hg.1 | ENST00000616259.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006773.hg.1 | ENST00000616259.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006773.hg.1 | ENST00000616259.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006773.hg.1 | ENST00000616259.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006773.hg.1 | ENST00000617793.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006773.hg.1 | ENST00000617793.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006773.hg.1 | ENST00000617793.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006773.hg.1 | ENST00000617793.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006773.hg.1 | ENST00000617793.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006773.hg.1 | ENST00000617793.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006773.hg.1 | ENST00000617793.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006773.hg.1 | ENST00000617793.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006773.hg.1 | ENST00000617889.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006773.hg.1 | ENST00000617889.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006773.hg.1 | ENST00000617889.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006773.hg.1 | ENST00000617889.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006773.hg.1 | ENST00000617889.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006773.hg.1 | ENST00000617889.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006773.hg.1 | ENST00000617889.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006773.hg.1 | ENST00000617889.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006773.hg.1 | ENST00000617889.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006773.hg.1 | NM_001291822.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200006773.hg.1 | NM_001291822.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200006773.hg.1 | NM_001291822.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200006773.hg.1 | NM_001291822.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200006773.hg.1 | NM_001291822.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200006773.hg.1 | NM_001291822.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200006773.hg.1 | NM_001291822.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200006773.hg.1 | NM_001291822.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200006773.hg.1 | NM_001291823.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200006773.hg.1 | NM_001291823.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200006773.hg.1 | NM_001291823.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200006773.hg.1 | NM_001291823.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200006773.hg.1 | NM_001291823.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200006773.hg.1 | NM_001291823.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200006773.hg.1 | NM_016523.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200006773.hg.1 | NM_016523.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200006773.hg.1 | NM_016523.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200006773.hg.1 | NM_016523.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200006773.hg.1 | NM_016523.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200006773.hg.1 | NM_016523.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200006773.hg.1 | NM_016523.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200006773.hg.1 | NM_016523.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200006773.hg.1 | NM_016523.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200006773.hg.1 | NR_120305.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200006773.hg.1 | NR_120305.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200006773.hg.1 | NR_120305.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200006773.hg.1 | NR_120305.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200006773.hg.1 | NR_120305.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200006773.hg.1 | NR_120305.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200006773.hg.1 | NR_120305.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200147603.hg.1 | ENST00000279545.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200147603.hg.1 | ENST00000545196.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200147603.hg.1 | ENST00000612321.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200147603.hg.1 | ENST00000616259.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200147603.hg.1 | ENST00000617793.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200147603.hg.1 | ENST00000617889.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200147603.hg.1 | NM_001291822.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200147603.hg.1 | NM_016523.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200147603.hg.1 | NR_120305.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200147603.hg.1 | ENST00000279545.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200147603.hg.1 | ENST00000545196.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200147603.hg.1 | ENST00000612321.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200147603.hg.1 | ENST00000616259.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200147603.hg.1 | ENST00000617793.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200147603.hg.1 | ENST00000617889.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200147603.hg.1 | NM_001291822.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200147603.hg.1 | NM_016523.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200147603.hg.1 | NR_120305.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200147603.hg.1 | TC1200006773.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC1200006773.hg.1 | ENST00000279545.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006773.hg.1 | ENST00000279545.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006773.hg.1 | ENST00000279545.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006773.hg.1 | ENST00000279545.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006773.hg.1 | ENST00000279545.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006773.hg.1 | ENST00000279545.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006773.hg.1 | ENST00000279545.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006773.hg.1 | ENST00000279545.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006773.hg.1 | ENST00000354855.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006773.hg.1 | ENST00000354855.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006773.hg.1 | ENST00000354855.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006773.hg.1 | ENST00000354855.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006773.hg.1 | ENST00000354855.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006773.hg.1 | ENST00000354855.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006773.hg.1 | ENST00000537723.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006773.hg.1 | ENST00000537723.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006773.hg.1 | ENST00000537723.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006773.hg.1 | ENST00000537723.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006773.hg.1 | ENST00000537723.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006773.hg.1 | ENST00000545196.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006773.hg.1 | ENST00000545196.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006773.hg.1 | ENST00000545196.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006773.hg.1 | ENST00000545196.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006773.hg.1 | ENST00000545196.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006773.hg.1 | ENST00000545196.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006773.hg.1 | ENST00000612321.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006773.hg.1 | ENST00000612321.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006773.hg.1 | ENST00000612321.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006773.hg.1 | ENST00000612321.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006773.hg.1 | ENST00000612321.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006773.hg.1 | ENST00000612321.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006773.hg.1 | ENST00000612321.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006773.hg.1 | ENST00000616259.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006773.hg.1 | ENST00000616259.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006773.hg.1 | ENST00000616259.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006773.hg.1 | ENST00000616259.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006773.hg.1 | ENST00000616259.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006773.hg.1 | ENST00000617793.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006773.hg.1 | ENST00000617793.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006773.hg.1 | ENST00000617793.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006773.hg.1 | ENST00000617793.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006773.hg.1 | ENST00000617793.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006773.hg.1 | ENST00000617793.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006773.hg.1 | ENST00000617793.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006773.hg.1 | ENST00000617793.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006773.hg.1 | ENST00000617889.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006773.hg.1 | ENST00000617889.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006773.hg.1 | ENST00000617889.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006773.hg.1 | ENST00000617889.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006773.hg.1 | ENST00000617889.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006773.hg.1 | ENST00000617889.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006773.hg.1 | ENST00000617889.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006773.hg.1 | ENST00000617889.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006773.hg.1 | ENST00000617889.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006773.hg.1 | NM_001291822.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200006773.hg.1 | NM_001291822.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200006773.hg.1 | NM_001291822.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200006773.hg.1 | NM_001291822.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200006773.hg.1 | NM_001291822.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200006773.hg.1 | NM_001291822.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200006773.hg.1 | NM_001291822.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200006773.hg.1 | NM_001291822.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200006773.hg.1 | NM_001291823.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200006773.hg.1 | NM_001291823.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200006773.hg.1 | NM_001291823.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200006773.hg.1 | NM_001291823.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200006773.hg.1 | NM_001291823.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200006773.hg.1 | NM_001291823.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200006773.hg.1 | NM_016523.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1200006773.hg.1 | NM_016523.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1200006773.hg.1 | NM_016523.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1200006773.hg.1 | NM_016523.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1200006773.hg.1 | NM_016523.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1200006773.hg.1 | NM_016523.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1200006773.hg.1 | NM_016523.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1200006773.hg.1 | NM_016523.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1200006773.hg.1 | NM_016523.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1200006773.hg.1 | NR_120305.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200006773.hg.1 | NR_120305.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200006773.hg.1 | NR_120305.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200006773.hg.1 | NR_120305.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200006773.hg.1 | NR_120305.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200006773.hg.1 | NR_120305.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200006773.hg.1 | NR_120305.1 | Refseq |
