Assay Details
| Gene Symbol: TRMT13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019083.2 | NP_061956.2 | 4 - 5 | 344 | 112 | |
| XM_017001504.1 | XP_016856993.1 | 4 - 5 | 331 | 112 | ||
| GenBank mRNA | AK001149.1 | - | 4 - 5 | 329 | 112 | |
| AK002081.1 | - | 4 - 5 | 331 | 112 | ||
| AK289404.1 | - | 4 - 5 | 331 | 112 | ||
| AK294738.1 | - | 4 - 5 | 329 | 112 | ||
| BC057226.1 | - | 4 - 5 | 344 | 112 | ||
| BC075811.1 | - | 4 - 5 | 341 | 5104751 | 112 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01000899.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0100009204.hg.1 | ENST00000370139.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009204.hg.1 | ENST00000370139.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009204.hg.1 | ENST00000370139.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009204.hg.1 | ENST00000370139.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009204.hg.1 | ENST00000370139.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009204.hg.1 | ENST00000370139.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009204.hg.1 | ENST00000370139.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009204.hg.1 | ENST00000370139.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009204.hg.1 | ENST00000370141.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009204.hg.1 | ENST00000370141.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009204.hg.1 | ENST00000370141.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009204.hg.1 | ENST00000370141.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009204.hg.1 | ENST00000370141.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009204.hg.1 | ENST00000370141.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009204.hg.1 | ENST00000370141.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009204.hg.1 | ENST00000370141.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009204.hg.1 | ENST00000370141.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009204.hg.1 | ENST00000370141.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009204.hg.1 | ENST00000370141.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009204.hg.1 | ENST00000370141.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009204.hg.1 | ENST00000370141.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009204.hg.1 | ENST00000370141.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009204.hg.1 | ENST00000370143.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009204.hg.1 | ENST00000370143.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009204.hg.1 | ENST00000370143.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009204.hg.1 | ENST00000370143.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009204.hg.1 | ENST00000370143.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009204.hg.1 | ENST00000370143.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009204.hg.1 | ENST00000370143.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009204.hg.1 | ENST00000370143.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009204.hg.1 | ENST00000482437.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009204.hg.1 | ENST00000482437.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009204.hg.1 | ENST00000482437.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009204.hg.1 | ENST00000482437.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009204.hg.1 | ENST00000482437.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009204.hg.1 | ENST00000482437.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009204.hg.1 | ENST00000482437.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009204.hg.1 | ENST00000482437.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009204.hg.1 | ENST00000482437.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009204.hg.1 | ENST00000482437.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009204.hg.1 | ENST00000482437.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009204.hg.1 | ENST00000482437.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009204.hg.1 | ENST00000482437.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009204.hg.1 | ENST00000482437.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009204.hg.1 | ENST00000482437.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009204.hg.1 | ENST00000482437.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009204.hg.1 | ENST00000493651.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009204.hg.1 | ENST00000493651.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009204.hg.1 | ENST00000493651.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009204.hg.1 | ENST00000493651.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009204.hg.1 | NM_019083.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009204.hg.1 | NM_019083.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009204.hg.1 | NM_019083.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009204.hg.1 | NM_019083.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009204.hg.1 | NM_019083.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009204.hg.1 | NM_019083.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009204.hg.1 | NM_019083.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009204.hg.1 | NM_019083.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009204.hg.1 | NM_019083.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009204.hg.1 | NM_019083.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009204.hg.1 | NM_019083.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009204.hg.1 | NM_019083.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009204.hg.1 | NM_019083.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009204.hg.1 | NM_019083.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01014155.hg.1 | TC01000899.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01014155.hg.1 | ENST00000370139.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01014155.hg.1 | ENST00000370141.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01014155.hg.1 | NM_019083.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100165567.hg.1 | ENST00000370139.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100165567.hg.1 | ENST00000370141.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100165567.hg.1 | ENST00000482437.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100165567.hg.1 | NM_019083.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100165567.hg.1 | ENST00000370139.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100165567.hg.1 | ENST00000370141.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100165567.hg.1 | ENST00000482437.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100165567.hg.1 | NM_019083.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100165567.hg.1 | TC0100009204.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0100009204.hg.1 | ENST00000370139.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009204.hg.1 | ENST00000370139.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009204.hg.1 | ENST00000370139.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009204.hg.1 | ENST00000370139.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009204.hg.1 | ENST00000370139.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009204.hg.1 | ENST00000370139.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009204.hg.1 | ENST00000370139.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009204.hg.1 | ENST00000370139.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009204.hg.1 | ENST00000370141.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009204.hg.1 | ENST00000370141.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009204.hg.1 | ENST00000370141.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009204.hg.1 | ENST00000370141.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009204.hg.1 | ENST00000370141.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009204.hg.1 | ENST00000370141.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009204.hg.1 | ENST00000370141.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009204.hg.1 | ENST00000370141.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009204.hg.1 | ENST00000370141.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009204.hg.1 | ENST00000370141.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009204.hg.1 | ENST00000370141.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009204.hg.1 | ENST00000370141.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009204.hg.1 | ENST00000370141.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009204.hg.1 | ENST00000370141.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009204.hg.1 | ENST00000370143.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009204.hg.1 | ENST00000370143.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009204.hg.1 | ENST00000370143.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009204.hg.1 | ENST00000370143.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009204.hg.1 | ENST00000370143.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009204.hg.1 | ENST00000370143.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009204.hg.1 | ENST00000370143.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009204.hg.1 | ENST00000370143.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009204.hg.1 | ENST00000482437.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009204.hg.1 | ENST00000482437.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009204.hg.1 | ENST00000482437.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009204.hg.1 | ENST00000482437.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009204.hg.1 | ENST00000482437.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009204.hg.1 | ENST00000482437.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009204.hg.1 | ENST00000482437.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009204.hg.1 | ENST00000482437.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009204.hg.1 | ENST00000482437.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009204.hg.1 | ENST00000482437.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009204.hg.1 | ENST00000482437.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009204.hg.1 | ENST00000482437.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009204.hg.1 | ENST00000482437.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009204.hg.1 | ENST00000482437.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009204.hg.1 | ENST00000482437.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009204.hg.1 | ENST00000482437.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009204.hg.1 | ENST00000493651.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009204.hg.1 | ENST00000493651.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009204.hg.1 | ENST00000493651.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009204.hg.1 | ENST00000493651.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009204.hg.1 | NM_019083.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009204.hg.1 | NM_019083.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009204.hg.1 | NM_019083.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009204.hg.1 | NM_019083.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009204.hg.1 | NM_019083.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009204.hg.1 | NM_019083.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009204.hg.1 | NM_019083.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009204.hg.1 | NM_019083.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009204.hg.1 | NM_019083.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009204.hg.1 | NM_019083.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009204.hg.1 | NM_019083.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009204.hg.1 | NM_019083.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009204.hg.1 | NM_019083.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009204.hg.1 | NM_019083.2 | Refseq |

TRMT13
Genomic Map