Assay Details
| Gene Symbol: MROH2A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001287395.1 | NP_001274324.1 | 26 - 27 | 2958 | 110 | |
| XM_011511075.2 | XP_011509377.1 | 26 - 27 | 3180 | 110 | ||
| XM_011511076.2 | XP_011509378.1 | 26 - 27 | 3180 | 110 | ||
| XM_011511077.2 | XP_011509379.1 | 26 - 27 | 3180 | 110 | ||
| XM_011511078.2 | XP_011509380.1 | 26 - 27 | 3141 | 110 | ||
| XM_011511079.2 | XP_011509381.1 | 26 - 27 | 3135 | 110 | ||
| XM_011511080.2 | XP_011509382.1 | 26 - 27 | 4651 | 110 | ||
| XM_011511082.2 | XP_011509384.1 | 26 - 27 | 3180 | 110 | ||
| XM_011511083.2 | XP_011509385.1 | 26 - 27 | 3180 | 110 | ||
| XM_011511084.2 | XP_011509386.1 | 25 - 26 | 2982 | 110 | ||
| XM_011511085.2 | XP_011509387.1 | 20 - 21 | 2250 | 110 | ||
More Information
Updated Mapping Information
Set Membership: |
|

MROH2A
Genomic Map