Assay Details
| Gene Symbol: MAN1B1-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_027447.1 | 1 | 141 | |||
| GenBank mRNA | AK023312.1 | - | 1 | 141 | ||
| BC020858.1 | - | 1 | 697 | 141 | ||
More Information
Updated Mapping Information
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Human Genome Microarray 4x44K v2 | A_24_P333567 | - | - |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_44k_24_P333567 | - | - |
| Agilent Whole Human Genome Oligo Microarray 4x44K | 6940 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 84 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_24_P333567 | - | - |
| SurePrint G3 Human GE Microarray G4851A | A_24_P333567 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07011557 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27363370 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07011558 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07011558 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07011557 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27363370 | - | - |
| Whole Human Genome Oligo Microarray G4112A | A_24_P333567 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P02195149 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07011513 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P16278852 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P16278889 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07011525 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07011530 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07011520 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P16278870 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07011516 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07011511 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07011528 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07011558 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07011532 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07011508 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P16278856 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07011509 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P16278894 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P16278892 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07011514 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P16278850 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07011507 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07011518 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07011523 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P16278885 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07011517 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07011534 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix HT HG-U133+ PM Array Plate | 232420_PM_x_at | - | - |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008247.hg.1 | ENST00000366110.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008247.hg.1 | ENST00000366110.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008247.hg.1 | ENST00000366110.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008247.hg.1 | ENST00000366110.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008247.hg.1 | ENST00000366110.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008247.hg.1 | ENST00000366110.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008247.hg.1 | ENST00000366110.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008247.hg.1 | ENST00000366110.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008247.hg.1 | ENST00000366110.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008247.hg.1 | ENST00000366110.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008247.hg.1 | ENST00000366110.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008247.hg.1 | ENST00000366110.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008247.hg.1 | ENST00000366110.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008247.hg.1 | ENST00000366110.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008247.hg.1 | NR_038941.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008247.hg.1 | NR_038941.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008247.hg.1 | NR_038941.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008247.hg.1 | NR_038941.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008247.hg.1 | NR_038941.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008247.hg.1 | NR_038941.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008247.hg.1 | NR_038941.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012031.hg.1 | ENST00000596585.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012031.hg.1 | ENST00000596585.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012031.hg.1 | NR_027447.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012031.hg.1 | NR_027447.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008247.hg.1 | ENST00000366110.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008247.hg.1 | ENST00000366110.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008247.hg.1 | ENST00000366110.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008247.hg.1 | ENST00000366110.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008247.hg.1 | ENST00000366110.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008247.hg.1 | ENST00000366110.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008247.hg.1 | ENST00000366110.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008247.hg.1 | ENST00000366110.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008247.hg.1 | ENST00000366110.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008247.hg.1 | ENST00000366110.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008247.hg.1 | ENST00000366110.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008247.hg.1 | ENST00000366110.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008247.hg.1 | ENST00000366110.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008247.hg.1 | ENST00000366110.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008247.hg.1 | NR_038941.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008247.hg.1 | NR_038941.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008247.hg.1 | NR_038941.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008247.hg.1 | NR_038941.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008247.hg.1 | NR_038941.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008247.hg.1 | NR_038941.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008247.hg.1 | NR_038941.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900182757.hg.1 | ENST00000596585.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900182757.hg.1 | NR_027447.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900182757.hg.1 | ENST00000596585.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900182757.hg.1 | NR_027447.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900182757.hg.1 | TC0900012031.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0900012031.hg.1 | ENST00000596585.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012031.hg.1 | ENST00000596585.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012031.hg.1 | NR_027447.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012031.hg.1 | NR_027447.1 | Refseq |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A038256 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A038256 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 231011 | - | - |

MAN1B1-AS1
Genomic Map