Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR04023318.hg.1 |
TC04001673.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR04023318.hg.1 |
ENST00000422544.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04023318.hg.1 |
ENST00000502773.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04023318.hg.1 |
ENST00000543489.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04023318.hg.1 |
NM_016205.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR04023318.hg.1 |
NR_036641.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400183989.hg.1 |
ENST00000274071.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400183989.hg.1 |
ENST00000422544.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400183989.hg.1 |
ENST00000502773.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400183989.hg.1 |
ENST00000512711.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400183989.hg.1 |
NM_016205.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400183989.hg.1 |
NR_036641.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400183989.hg.1 |
ENST00000274071.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400183989.hg.1 |
ENST00000422544.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400183989.hg.1 |
ENST00000502773.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400183989.hg.1 |
ENST00000512711.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400183989.hg.1 |
NM_016205.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400183989.hg.1 |
NR_036641.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400183989.hg.1 |
TC0400012222.hg.1 |
Clariom D TC |