Assay Details
| Gene Symbol: MED28 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF317678.1 | - | 4 - 5 | 532 | 116 | |
More Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0400006990.hg.1 | ENST00000237380.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006990.hg.1 | ENST00000237380.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006990.hg.1 | ENST00000237380.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006990.hg.1 | ENST00000237380.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006990.hg.1 | ENST00000237380.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006990.hg.1 | ENST00000237380.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006990.hg.1 | ENST00000237380.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006990.hg.1 | ENST00000237380.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006990.hg.1 | ENST00000237380.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006990.hg.1 | ENST00000237380.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006990.hg.1 | ENST00000237380.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006990.hg.1 | ENST00000237380.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006990.hg.1 | ENST00000237380.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006990.hg.1 | ENST00000237380.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006990.hg.1 | ENST00000237380.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006990.hg.1 | ENST00000237380.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006990.hg.1 | ENST00000237380.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006990.hg.1 | ENST00000237380.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006990.hg.1 | ENST00000237380.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006990.hg.1 | ENST00000499786.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006990.hg.1 | ENST00000499786.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006990.hg.1 | ENST00000503945.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006990.hg.1 | ENST00000503945.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006990.hg.1 | ENST00000503945.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006990.hg.1 | ENST00000503945.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006990.hg.1 | ENST00000503945.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006990.hg.1 | ENST00000503945.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006990.hg.1 | ENST00000506409.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006990.hg.1 | ENST00000506409.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006990.hg.1 | ENST00000506409.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006990.hg.1 | ENST00000612711.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006990.hg.1 | ENST00000612711.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006990.hg.1 | ENST00000612711.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006990.hg.1 | ENST00000612711.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006990.hg.1 | ENST00000612711.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006990.hg.1 | NM_025205.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006990.hg.1 | NM_025205.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006990.hg.1 | NM_025205.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006990.hg.1 | NM_025205.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006990.hg.1 | NM_025205.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006990.hg.1 | NM_025205.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006990.hg.1 | NM_025205.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400006990.hg.1 | ENST00000237380.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006990.hg.1 | ENST00000237380.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006990.hg.1 | ENST00000237380.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006990.hg.1 | ENST00000237380.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006990.hg.1 | ENST00000237380.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006990.hg.1 | ENST00000237380.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006990.hg.1 | ENST00000237380.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006990.hg.1 | ENST00000237380.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006990.hg.1 | ENST00000237380.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006990.hg.1 | ENST00000237380.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006990.hg.1 | ENST00000237380.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006990.hg.1 | ENST00000237380.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006990.hg.1 | ENST00000237380.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006990.hg.1 | ENST00000237380.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006990.hg.1 | ENST00000237380.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006990.hg.1 | ENST00000237380.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006990.hg.1 | ENST00000237380.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006990.hg.1 | ENST00000237380.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006990.hg.1 | ENST00000237380.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006990.hg.1 | ENST00000499786.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006990.hg.1 | ENST00000499786.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006990.hg.1 | ENST00000503945.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006990.hg.1 | ENST00000503945.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006990.hg.1 | ENST00000503945.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006990.hg.1 | ENST00000503945.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006990.hg.1 | ENST00000503945.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006990.hg.1 | ENST00000503945.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006990.hg.1 | ENST00000506409.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006990.hg.1 | ENST00000506409.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006990.hg.1 | ENST00000506409.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006990.hg.1 | ENST00000612711.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006990.hg.1 | ENST00000612711.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006990.hg.1 | ENST00000612711.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006990.hg.1 | ENST00000612711.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006990.hg.1 | ENST00000612711.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006990.hg.1 | NM_025205.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400006990.hg.1 | NM_025205.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400006990.hg.1 | NM_025205.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400006990.hg.1 | NM_025205.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400006990.hg.1 | NM_025205.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400006990.hg.1 | NM_025205.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400006990.hg.1 | NM_025205.4 | Refseq |

MED28
Genomic Map