Assay Details
| Gene Symbol: SEC31A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077206.3 | NP_001070674.1 | 15 - 16 | 1895 | 104 | |
| NM_001077207.3 | NP_001070675.1 | 15 - 16 | 1895 | 104 | ||
| NM_001077208.3 | NP_001070676.1 | 15 - 16 | 1895 | 104 | ||
| NM_001191049.2 | NP_001177978.1 | 14 - 15 | 1812 | 104 | ||
| NM_001300744.2 | NP_001287673.1 | 13 - 14 | 1778 | 104 | ||
| NM_001300745.2 | NP_001287674.1 | 13 - 14 | 1778 | 104 | ||
| NM_001318119.1 | NP_001305048.1 | 15 - 16 | 1892 | 104 | ||
| NM_001318120.1 | NP_001305049.1 | 17 - 18 | 2109 | 104 | ||
| NM_016211.4 | NP_057295.2 | 13 - 14 | 1778 | 104 | ||
| GenBank mRNA | AB018358.1 | - | 13 - 14 | 1634 | 104 | |
| AB018359.1 | - | 15 - 16 | 1751 | 104 | ||
| AB020712.1 | - | 17 - 18 | 2076 | 104 | ||
| AF139184.1 | - | 14 - 15 | 1698 | 104 | ||
| AK001014.1 | - | 4 - 5 | 208 | 104 | ||
| AK002161.1 | - | 15 - 16 | 1768 | 104 | ||
| AK125897.1 | - | 7 - 8 | 1382 | 104 | ||
| AK295810.1 | - | 14 - 15 | 1710 | 104 | ||
| BC084583.1 | - | 15 - 16 | 1734 | 6196402 | 104 | |
| BC117221.1 | - | 15 - 16 | 1763 | 40125772 | 104 | |
| BC143489.1 | - | 15 - 16 | 1760 | 9051997 | 104 | |
| BC143491.1 | - | 15 - 16 | 1763 | 9051999 | 104 | |
| BC143492.2 | - | 13 - 14 | 1646 | 9052000 | 104 | |
| CR933696.1 | - | 13 - 14 | 1651 | 104 | ||
More Information
Set Membership: |
|

SEC31A
Genomic Map