Assay Details
| Gene Symbol: MTFR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001099286.2 | NP_001092756.1 | 6 - 7 | 1283 | 121 | |
| NM_001318738.1 | NP_001305667.1 | 7 - 8 | 1095 | 121 | ||
| NM_138419.4 | NP_612428.2 | 6 - 7 | 995 | 121 | ||
| XM_011535410.2 | XP_011533712.1 | 7 - 8 | 1347 | 121 | ||
| XM_011535412.2 | XP_011533714.1 | 5 - 6 | 1088 | 121 | ||
| XM_011535413.2 | XP_011533715.1 | 5 - 6 | 4202 | 121 | ||
| GenBank mRNA | AK125758.1 | - | 7 - 8 | 1052 | 121 | |
| AK292303.1 | - | 6 - 7 | 1234 | 121 | ||
| BC011716.2 | - | 3 - 4 | 618 | 121 | ||
| BC063688.1 | - | 6 - 7 | 948 | 4455541 | 121 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC06002142.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR06026950.hg.1 | TC06002142.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR06026950.hg.1 | ENST00000420702.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06026950.hg.1 | ENST00000451457.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06026950.hg.1 | NM_001099286.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06026950.hg.1 | NM_138419.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06026949.hg.1 | TC06002142.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR06026949.hg.1 | ENST00000420702.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06026949.hg.1 | ENST00000451457.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06026949.hg.1 | NM_001099286.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06026949.hg.1 | NM_138419.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | ENST00000367784.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | ENST00000367784.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | ENST00000367784.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | ENST00000367784.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | ENST00000367784.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | ENST00000367784.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | ENST00000367784.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | ENST00000367784.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | ENST00000367784.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | ENST00000367784.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | ENST00000367784.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | ENST00000367784.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | ENST00000418509.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | ENST00000418509.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | ENST00000418509.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | ENST00000418509.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | ENST00000418509.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | ENST00000418509.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | ENST00000420702.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | ENST00000420702.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | ENST00000420702.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | ENST00000420702.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | ENST00000420702.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | ENST00000420702.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | ENST00000420702.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | ENST00000420702.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | ENST00000420702.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | ENST00000420702.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | ENST00000420702.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | ENST00000420702.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | ENST00000445767.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | ENST00000445767.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | ENST00000445767.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | ENST00000445767.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | ENST00000445767.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | ENST00000451457.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | ENST00000451457.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | ENST00000451457.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | ENST00000451457.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | ENST00000451457.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | ENST00000451457.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | ENST00000451457.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | ENST00000451457.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | ENST00000451457.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | ENST00000451457.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | ENST00000451457.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | ENST00000527338.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | ENST00000527338.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | ENST00000527338.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | ENST00000527338.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | ENST00000532958.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | ENST00000532958.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | ENST00000532958.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | NM_001099286.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | NM_001099286.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | NM_001099286.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | NM_001099286.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | NM_001099286.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | NM_001099286.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | NM_001099286.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | NM_001099286.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | NM_001099286.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | NM_001099286.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | NM_001099286.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | NM_001099286.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | NM_138419.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | NM_138419.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | NM_138419.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | NM_138419.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | NM_138419.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | NM_138419.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | NM_138419.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | NM_138419.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | NM_138419.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | NM_138419.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013271.hg.1 | NM_138419.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600191782.hg.1 | ENST00000367784.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600191782.hg.1 | ENST00000418509.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600191782.hg.1 | ENST00000420702.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600191782.hg.1 | ENST00000451457.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600191782.hg.1 | ENST00000532958.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600191782.hg.1 | NM_001099286.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600191782.hg.1 | NM_138419.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600191782.hg.1 | ENST00000367784.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600191782.hg.1 | ENST00000418509.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600191782.hg.1 | ENST00000420702.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600191782.hg.1 | ENST00000451457.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600191782.hg.1 | ENST00000532958.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600191782.hg.1 | NM_001099286.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600191782.hg.1 | NM_138419.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600191782.hg.1 | TC0600013271.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | ENST00000367784.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | ENST00000367784.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | ENST00000367784.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | ENST00000367784.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | ENST00000367784.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | ENST00000367784.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | ENST00000367784.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | ENST00000367784.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | ENST00000367784.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | ENST00000367784.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | ENST00000367784.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | ENST00000367784.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | ENST00000418509.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | ENST00000418509.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | ENST00000418509.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | ENST00000418509.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | ENST00000418509.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | ENST00000418509.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | ENST00000420702.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | ENST00000420702.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | ENST00000420702.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | ENST00000420702.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | ENST00000420702.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | ENST00000420702.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | ENST00000420702.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | ENST00000420702.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | ENST00000420702.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | ENST00000420702.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | ENST00000420702.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | ENST00000420702.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | ENST00000445767.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | ENST00000445767.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | ENST00000445767.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | ENST00000445767.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | ENST00000445767.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | ENST00000451457.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | ENST00000451457.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | ENST00000451457.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | ENST00000451457.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | ENST00000451457.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | ENST00000451457.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | ENST00000451457.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | ENST00000451457.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | ENST00000451457.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | ENST00000451457.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | ENST00000451457.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | ENST00000527338.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | ENST00000527338.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | ENST00000527338.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | ENST00000527338.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | ENST00000532958.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | ENST00000532958.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | ENST00000532958.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | NM_001099286.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | NM_001099286.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | NM_001099286.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | NM_001099286.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | NM_001099286.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | NM_001099286.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | NM_001099286.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | NM_001099286.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | NM_001099286.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | NM_001099286.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | NM_001099286.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | NM_001099286.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | NM_138419.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | NM_138419.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | NM_138419.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | NM_138419.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | NM_138419.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | NM_138419.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | NM_138419.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | NM_138419.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | NM_138419.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | NM_138419.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013271.hg.1 | NM_138419.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600191780.hg.1 | ENST00000367784.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600191780.hg.1 | ENST00000420702.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600191780.hg.1 | ENST00000451457.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600191780.hg.1 | ENST00000532958.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600191780.hg.1 | NM_001099286.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600191780.hg.1 | NM_138419.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600191780.hg.1 | ENST00000367784.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600191780.hg.1 | ENST00000420702.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600191780.hg.1 | ENST00000451457.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600191780.hg.1 | ENST00000532958.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600191780.hg.1 | NM_001099286.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600191780.hg.1 | NM_138419.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600191780.hg.1 | TC0600013271.hg.1 | Clariom D TC |

MTFR2
Genomic Map