Assay Details
| Gene Symbol: ADAMTS8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007037.5 | NP_008968.4 | 1 - 2 | 1426 | 64 | |
| XM_017017145.1 | XP_016872634.1 | 1 - 2 | 946 | 64 | ||
| GenBank mRNA | AF060153.1 | - | 1 - 2 | 1427 | 64 | |
| AF175283.1 | - | 1 - 2 | 142 | 64 | ||
| BC089435.1 | - | 2 - 3 | 806 | 64 | ||
| BC113874.1 | - | 2 - 3 | 81 | 64 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC11002452.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR11029453.hg.1 | ENST00000257359.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11029453.hg.1 | ENST00000414575.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11029453.hg.1 | NM_007037.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11029453.hg.1 | TC11002452.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR11029456.hg.1 | ENST00000257359.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11029456.hg.1 | NM_007037.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11029456.hg.1 | TC11002452.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012834.hg.1 | ENST00000257359.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012834.hg.1 | ENST00000257359.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012834.hg.1 | ENST00000257359.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012834.hg.1 | ENST00000257359.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012834.hg.1 | ENST00000257359.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012834.hg.1 | ENST00000257359.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012834.hg.1 | ENST00000257359.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012834.hg.1 | ENST00000257359.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012834.hg.1 | ENST00000257359.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012834.hg.1 | ENST00000257359.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012834.hg.1 | ENST00000257359.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012834.hg.1 | ENST00000257359.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012834.hg.1 | ENST00000257359.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012834.hg.1 | ENST00000257359.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012834.hg.1 | ENST00000531752.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012834.hg.1 | ENST00000531752.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012834.hg.1 | ENST00000531752.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012834.hg.1 | ENST00000531752.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012834.hg.1 | ENST00000531752.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012834.hg.1 | ENST00000531752.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012834.hg.1 | ENST00000531752.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012834.hg.1 | ENST00000531752.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012834.hg.1 | NM_007037.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012834.hg.1 | NM_007037.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012834.hg.1 | NM_007037.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012834.hg.1 | NM_007037.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012834.hg.1 | NM_007037.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012834.hg.1 | NM_007037.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012834.hg.1 | NM_007037.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012834.hg.1 | NM_007037.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012834.hg.1 | NM_007037.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012834.hg.1 | NM_007037.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012834.hg.1 | NM_007037.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012834.hg.1 | NM_007037.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012834.hg.1 | NM_007037.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11029454.hg.1 | ENST00000257359.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11029454.hg.1 | NM_007037.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11029454.hg.1 | TC11002452.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR11029451.hg.1 | ENST00000257359.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11029451.hg.1 | ENST00000414575.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11029451.hg.1 | NM_007037.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11029451.hg.1 | TC11002452.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR11029455.hg.1 | ENST00000257359.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11029455.hg.1 | ENST00000414575.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11029455.hg.1 | NM_007037.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11029455.hg.1 | TC11002452.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR11029452.hg.1 | ENST00000257359.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11029452.hg.1 | ENST00000414575.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11029452.hg.1 | NM_007037.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11029452.hg.1 | TC11002452.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR11029450.hg.1 | ENST00000257359.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11029450.hg.1 | ENST00000414575.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11029450.hg.1 | NM_007037.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11029450.hg.1 | TC11002452.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1100198507.hg.1 | ENST00000257359.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100198507.hg.1 | NM_007037.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100198507.hg.1 | ENST00000257359.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100198507.hg.1 | NM_007037.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100198507.hg.1 | TC1100012834.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1100198508.hg.1 | ENST00000257359.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100198508.hg.1 | NM_007037.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100198508.hg.1 | ENST00000257359.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100198508.hg.1 | NM_007037.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100198508.hg.1 | TC1100012834.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1100012834.hg.1 | ENST00000257359.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012834.hg.1 | ENST00000257359.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012834.hg.1 | ENST00000257359.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012834.hg.1 | ENST00000257359.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012834.hg.1 | ENST00000257359.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012834.hg.1 | ENST00000257359.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012834.hg.1 | ENST00000257359.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012834.hg.1 | ENST00000257359.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012834.hg.1 | ENST00000257359.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012834.hg.1 | ENST00000257359.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012834.hg.1 | ENST00000257359.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012834.hg.1 | ENST00000257359.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012834.hg.1 | ENST00000257359.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012834.hg.1 | ENST00000257359.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012834.hg.1 | ENST00000531752.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012834.hg.1 | ENST00000531752.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012834.hg.1 | ENST00000531752.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012834.hg.1 | ENST00000531752.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012834.hg.1 | ENST00000531752.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012834.hg.1 | ENST00000531752.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012834.hg.1 | ENST00000531752.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012834.hg.1 | ENST00000531752.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012834.hg.1 | NM_007037.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012834.hg.1 | NM_007037.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012834.hg.1 | NM_007037.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012834.hg.1 | NM_007037.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012834.hg.1 | NM_007037.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012834.hg.1 | NM_007037.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012834.hg.1 | NM_007037.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012834.hg.1 | NM_007037.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012834.hg.1 | NM_007037.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012834.hg.1 | NM_007037.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012834.hg.1 | NM_007037.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012834.hg.1 | NM_007037.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012834.hg.1 | NM_007037.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100198506.hg.1 | ENST00000257359.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100198506.hg.1 | NM_007037.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100198506.hg.1 | ENST00000257359.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100198506.hg.1 | NM_007037.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100198506.hg.1 | TC1100012834.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1100198510.hg.1 | ENST00000257359.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100198510.hg.1 | NM_007037.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100198510.hg.1 | ENST00000257359.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100198510.hg.1 | NM_007037.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100198510.hg.1 | TC1100012834.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1100198509.hg.1 | ENST00000257359.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100198509.hg.1 | NM_007037.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100198509.hg.1 | ENST00000257359.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100198509.hg.1 | NM_007037.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100198509.hg.1 | TC1100012834.hg.1 | Clariom D TC |

ADAMTS8
Genomic Map