| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11016761.hg.1 |
ENST00000454892.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11016761.hg.1 |
NR_001589.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11016761.hg.1 |
TC11001332.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11016762.hg.1 |
NR_001589.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11016762.hg.1 |
TC11001332.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11016763.hg.1 |
NR_001589.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11016763.hg.1 |
TC11001332.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009919.hg.1 |
ENST00000433329.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009919.hg.1 |
ENST00000433329.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009919.hg.1 |
ENST00000433329.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009919.hg.1 |
ENST00000433329.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009919.hg.1 |
ENST00000454892.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009919.hg.1 |
ENST00000454892.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009919.hg.1 |
ENST00000454892.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009919.hg.1 |
NR_001589.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009919.hg.1 |
NR_001589.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009919.hg.1 |
NR_001589.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009919.hg.1 |
NR_001589.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009919.hg.1 |
NR_001589.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009919.hg.1 |
NR_001589.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100174870.hg.1 |
ENST00000433329.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100174870.hg.1 |
ENST00000454892.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100174870.hg.1 |
NR_001589.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100174870.hg.1 |
ENST00000433329.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100174870.hg.1 |
ENST00000454892.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100174870.hg.1 |
NR_001589.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100174870.hg.1 |
TC1100009919.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1100174871.hg.1 |
ENST00000433329.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100174871.hg.1 |
NR_001589.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100174871.hg.1 |
ENST00000433329.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100174871.hg.1 |
NR_001589.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100174871.hg.1 |
TC1100009919.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1100174872.hg.1 |
ENST00000433329.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100174872.hg.1 |
NR_001589.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100174872.hg.1 |
ENST00000433329.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100174872.hg.1 |
NR_001589.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100174872.hg.1 |
TC1100009919.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC1100009919.hg.1 |
ENST00000433329.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009919.hg.1 |
ENST00000433329.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009919.hg.1 |
ENST00000433329.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009919.hg.1 |
ENST00000433329.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009919.hg.1 |
ENST00000454892.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009919.hg.1 |
ENST00000454892.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009919.hg.1 |
ENST00000454892.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009919.hg.1 |
NR_001589.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100009919.hg.1 |
NR_001589.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100009919.hg.1 |
NR_001589.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100009919.hg.1 |
NR_001589.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100009919.hg.1 |
NR_001589.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100009919.hg.1 |
NR_001589.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100174873.hg.1 |
NR_001589.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100174873.hg.1 |
NR_001589.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100174873.hg.1 |
TC1100009919.hg.1 |
Clariom D TC |