Assay Details
Gene Symbol: OR52I2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005170.2 | NP_001005170.1 | 1 | 593 | 78 | |
GenBank mRNA | BC136921.1 | - | 1 | 593 | 9020912 | 78 |
BC151144.1 | - | 1 | 593 | 9020913 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Human Whole Genome ChIP Tiling Array | A_17_P07610338 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P28009401 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P28009404 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P07610355 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P07610351 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P28009386 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P07610369 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P07610361 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P07610337 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P28009400 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P28009384 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P07610352 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P28009387 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P28009403 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P07610371 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P28009383 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P07610339 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P28009406 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P28009389 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P16415332 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P07610358 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P07610353 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P07610357 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P07610372 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P07610366 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P16415343 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P28009385 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P28009402 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P07610360 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P07610349 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3230374 | - | - |
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 26040 | - | - |
Agilent Human Genome CGH + SNP Microarray | A_16_P32831561 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3230374 | - | - |
SurePrint G3 Human GE Microarray G4851A | A_33_P3230374 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28009389 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07610360 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07610338 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28009400 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07610358 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07610339 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07610371 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28009393 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07610357 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16415332 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07610337 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28009387 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28009401 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07610349 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28009402 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07610372 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16415343 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07610355 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28009403 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28009404 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28009386 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28009384 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28009406 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07610352 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28009385 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28009406 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07610358 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28009384 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28009400 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07610337 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07610357 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28009404 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07610349 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07610372 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07610338 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07610339 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28009386 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28009389 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28009402 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28009393 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07610371 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16415343 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28009401 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07610360 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28009403 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28009385 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07610352 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28009387 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07610355 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16415332 | - | - |
Agilent Human Genome CGH Microarray 105A (G4412A) | A_16_P02383787 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P07610372 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P07610371 | - | - |
Agilent Human Genome 244K CGH Microarray (Alpha Test) | A_16_P02383787 | - | - |
Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 | 90120 | - | - |
Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 | 102512 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P02383787 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07610316 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07610314 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07610405 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07610339 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07610341 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07610304 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07610358 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07610343 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07610319 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07610366 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07610360 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07610312 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07610326 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07610308 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07610400 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07610327 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P16415355 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07610337 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P16415350 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07610419 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07610362 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07610403 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07610396 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07610371 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07610345 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07610335 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P16415360 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07610350 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07610421 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P16415359 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07610369 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07610333 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07610301 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07610323 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07610325 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07610328 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07610352 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07610414 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07610412 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07610338 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07610354 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07610416 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07610298 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07610306 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07610424 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07610409 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07610398 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07610364 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07610368 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07610348 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07610330 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07610402 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07610297 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07610310 | - | - |
Agilent SurePrint G3 Human CGH Microarray 2x400K | A_16_P02383787 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Gene 1.0 ST Array | NM_001005170_at | - | - |
Affymetrix Human Gene 1.0 ST Array | 7937947 | - | - |
Affymetrix Human Gene 1.1 ST Array | 7937946 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A046032 | - | - |
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A046032 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Human Genome Survey Microarray Version 2.0 | 176284 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
HumanHT-12 v4 Expression BeadChip | ILMN_1768064 | BC151144.1 | human_transcriptome |
HumanHT-12 v4 Expression BeadChip | ILMN_1768064 | NM_001005170.2 | human_transcriptome |
HumanHT-12 v4 Expression BeadChip | ILMN_1768064 | BC136921.1 | human_transcriptome |
Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1768064 | BC151144.1 | human_transcriptome |
Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1768064 | NM_001005170.2 | human_transcriptome |
Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1768064 | BC136921.1 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_1768064 | BC151144.1 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_1768064 | NM_001005170.2 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_1768064 | BC136921.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1768064 | BC151144.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1768064 | NM_001005170.2 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1768064 | BC136921.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1768064 | BC151144.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1768064 | NM_001005170.2 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1768064 | BC136921.1 | human_transcriptome |
Illumina HumanRef-8 v2 Expression BeadChip | GI_3014-S | - | - |
Illumina human-6 v2.0 expression beadchip | ILMN_1768064 | BC151144.1 | human_transcriptome |
Illumina human-6 v2.0 expression beadchip | ILMN_1768064 | NM_001005170.2 | human_transcriptome |
Illumina human-6 v2.0 expression beadchip | ILMN_1768064 | BC136921.1 | human_transcriptome |
Illumina humanRef-8 v2.0 expression beadchip | ILMN_1768064 | BC151144.1 | human_transcriptome |
Illumina humanRef-8 v2.0 expression beadchip | ILMN_1768064 | NM_001005170.2 | human_transcriptome |
Illumina humanRef-8 v2.0 expression beadchip | ILMN_1768064 | BC136921.1 | human_transcriptome |
Illumina Sentrix Human-6 v2 Expression BeadChip | 5130546 | - | - |
Illumina HumanRef-6 v2.0 expression beadchip | ILMN_3014 | - | - |
Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1768064 | BC151144.1 | human_transcriptome |
Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1768064 | NM_001005170.2 | human_transcriptome |
Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1768064 | BC136921.1 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1768064 | BC151144.1 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1768064 | NM_001005170.2 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1768064 | BC136921.1 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1768064 | BC151144.1 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1768064 | NM_001005170.2 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1768064 | BC136921.1 | human_transcriptome |
Illumina HumanWG-6_V2_0_R2 | ILMN_1768064 | BC151144.1 | human_transcriptome |
Illumina HumanWG-6_V2_0_R2 | ILMN_1768064 | NM_001005170.2 | human_transcriptome |
Illumina HumanWG-6_V2_0_R2 | ILMN_1768064 | BC136921.1 | human_transcriptome |
Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1768064 | BC151144.1 | human_transcriptome |
Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1768064 | NM_001005170.2 | human_transcriptome |
Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1768064 | BC136921.1 | human_transcriptome |
