Assay Details
| Gene Symbol: DNA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080449.2 | NP_001073918.2 | 12 - 13 | 1979 | 110 | |
| NR_102264.1 | 13 - 14 | 1959 | 110 | |||
| XM_006717680.3 | XP_006717743.1 | 13 - 14 | 2359 | 110 | ||
| XM_011539417.1 | XP_011537719.1 | 6 - 7 | 903 | 110 | ||
| XM_017015799.1 | XP_016871288.1 | 9 - 10 | 1368 | 110 | ||
| GenBank mRNA | AK308333.1 | - | 12 - 13 | 1979 | 110 | |
| AK308358.1 | - | 10 - 11 | 1620 | 110 | ||
| BC053574.1 | - | 2 - 3 | 173 | 110 | ||
| BC063664.1 | - | 12 - 13 | 1892 | 110 | ||
| BC111740.1 | - | 13 - 14 | 1959 | 40038149 | 110 | |
| D42046.1 | - | 12 - 13 | 1921 | 110 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC10001347.hg.1
Set Membership: |
|
Gene Ontology Categories:
Function(s)
Process(es)
DNA replication checkpoint
telomere maintenance via recombination
telomere maintenance
DNA double-strand break processing
DNA synthesis involved in DNA repair
strand displacement
DNA replication
mitochondrial DNA replication
base-excision repair
DNA duplex unwinding
DNA replication, Okazaki fragment processing
DNA replication, removal of RNA primer
mitochondrial DNA repair
positive regulation of DNA replication
nucleic acid phosphodiester bond hydrolysis
regulation of signal transduction by p53 class mediator
mitotic telomere maintenance via semi-conservative replication
DNA binding
DNA helicase activity
helicase activity
nuclease activity
protein binding
ATP binding
ATPase activity
site-specific endodeoxyribonuclease activity, specific for altered base
5'-flap endonuclease activity
5'-3' DNA helicase activity
single-stranded DNA-dependent ATPase activity
metal ion binding
4 iron, 4 sulfur cluster binding
DNA replication checkpoint
telomere maintenance via recombination
telomere maintenance
DNA double-strand break processing
DNA synthesis involved in DNA repair
strand displacement
DNA replication
mitochondrial DNA replication
base-excision repair
DNA duplex unwinding
DNA replication, Okazaki fragment processing
DNA replication, removal of RNA primer
mitochondrial DNA repair
positive regulation of DNA replication
nucleic acid phosphodiester bond hydrolysis
regulation of signal transduction by p53 class mediator
mitotic telomere maintenance via semi-conservative replication
DNA binding
DNA helicase activity
helicase activity
nuclease activity
protein binding
ATP binding
ATPase activity
site-specific endodeoxyribonuclease activity, specific for altered base
5'-flap endonuclease activity
5'-3' DNA helicase activity
single-stranded DNA-dependent ATPase activity
metal ion binding
4 iron, 4 sulfur cluster binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1000010847.hg.1 | ENST00000358410.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010847.hg.1 | ENST00000358410.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010847.hg.1 | ENST00000358410.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010847.hg.1 | ENST00000358410.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010847.hg.1 | ENST00000358410.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010847.hg.1 | ENST00000358410.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010847.hg.1 | ENST00000358410.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010847.hg.1 | ENST00000358410.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010847.hg.1 | ENST00000358410.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010847.hg.1 | ENST00000358410.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010847.hg.1 | ENST00000358410.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010847.hg.1 | ENST00000358410.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010847.hg.1 | ENST00000358410.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010847.hg.1 | ENST00000358410.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010847.hg.1 | ENST00000358410.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010847.hg.1 | ENST00000358410.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010847.hg.1 | ENST00000358410.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010847.hg.1 | ENST00000358410.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010847.hg.1 | ENST00000358410.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010847.hg.1 | ENST00000358410.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010847.hg.1 | ENST00000358410.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010847.hg.1 | ENST00000358410.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010847.hg.1 | ENST00000358410.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010847.hg.1 | ENST00000358410.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010847.hg.1 | ENST00000358410.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010847.hg.1 | ENST00000358410.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010847.hg.1 | ENST00000358410.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010847.hg.1 | ENST00000358410.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010847.hg.1 | ENST00000399179.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010847.hg.1 | ENST00000399179.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010847.hg.1 | ENST00000399179.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010847.hg.1 | ENST00000399179.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010847.hg.1 | ENST00000399179.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010847.hg.1 | ENST00000399179.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010847.hg.1 | ENST00000399179.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010847.hg.1 | ENST00000399179.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010847.hg.1 | ENST00000399179.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010847.hg.1 | ENST00000399179.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010847.hg.1 | ENST00000399179.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010847.hg.1 | ENST00000399179.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010847.hg.1 | ENST00000399179.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010847.hg.1 | ENST00000399179.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010847.hg.1 | ENST00000399179.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010847.hg.1 | ENST00000399179.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010847.hg.1 | ENST00000399179.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010847.hg.1 | ENST00000399179.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010847.hg.1 | ENST00000399179.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010847.hg.1 | ENST00000399179.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010847.hg.1 | ENST00000399179.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010847.hg.1 | ENST00000399179.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010847.hg.1 | ENST00000399179.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010847.hg.1 | ENST00000399179.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010847.hg.1 | ENST00000399179.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010847.hg.1 | ENST00000399180.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010847.hg.1 | ENST00000399180.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010847.hg.1 | ENST00000399180.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010847.hg.1 | ENST00000399180.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010847.hg.1 | ENST00000399180.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010847.hg.1 | ENST00000399180.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010847.hg.1 | ENST00000399180.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010847.hg.1 | ENST00000399180.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010847.hg.1 | ENST00000399180.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010847.hg.1 | ENST00000399180.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010847.hg.1 | ENST00000399180.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010847.hg.1 | ENST00000399180.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010847.hg.1 | ENST00000399180.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010847.hg.1 | ENST00000399180.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010847.hg.1 | ENST00000399180.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010847.hg.1 | ENST00000399180.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010847.hg.1 | ENST00000399180.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010847.hg.1 | ENST00000399180.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010847.hg.1 | ENST00000399180.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010847.hg.1 | ENST00000399180.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010847.hg.1 | ENST00000399180.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010847.hg.1 | ENST00000399180.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010847.hg.1 | ENST00000399180.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010847.hg.1 | ENST00000399180.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010847.hg.1 | ENST00000399180.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010847.hg.1 | ENST00000440722.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010847.hg.1 | ENST00000440722.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010847.hg.1 | ENST00000440722.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010847.hg.1 | ENST00000440722.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010847.hg.1 | ENST00000440722.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010847.hg.1 | ENST00000440722.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010847.hg.1 | ENST00000440722.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010847.hg.1 | ENST00000440722.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010847.hg.1 | ENST00000440722.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010847.hg.1 | ENST00000478029.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010847.hg.1 | ENST00000478029.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010847.hg.1 | ENST00000478029.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010847.hg.1 | ENST00000550357.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010847.hg.1 | ENST00000550357.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010847.hg.1 | ENST00000550357.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010847.hg.1 | ENST00000550357.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010847.hg.1 | ENST00000550545.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010847.hg.1 | ENST00000550545.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010847.hg.1 | ENST00000550545.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010847.hg.1 | ENST00000551118.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010847.hg.1 | ENST00000551118.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010847.hg.1 | ENST00000551118.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010847.hg.1 | ENST00000551118.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010847.hg.1 | ENST00000551118.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010847.hg.1 | ENST00000551118.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010847.hg.1 | ENST00000551118.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010847.hg.1 | ENST00000551118.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010847.hg.1 | ENST00000551118.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010847.hg.1 | ENST00000551118.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010847.hg.1 | ENST00000551118.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010847.hg.1 | ENST00000551118.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010847.hg.1 | ENST00000551118.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010847.hg.1 | ENST00000551118.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010847.hg.1 | ENST00000551118.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010847.hg.1 | ENST00000551118.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010847.hg.1 | ENST00000551118.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010847.hg.1 | ENST00000551118.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010847.hg.1 | ENST00000551118.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010847.hg.1 | ENST00000551118.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010847.hg.1 | ENST00000551118.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010847.hg.1 | NM_001080449.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010847.hg.1 | NM_001080449.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010847.hg.1 | NM_001080449.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010847.hg.1 | NM_001080449.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010847.hg.1 | NM_001080449.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010847.hg.1 | NM_001080449.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010847.hg.1 | NM_001080449.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010847.hg.1 | NM_001080449.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010847.hg.1 | NM_001080449.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010847.hg.1 | NM_001080449.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010847.hg.1 | NM_001080449.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010847.hg.1 | NM_001080449.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010847.hg.1 | NM_001080449.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010847.hg.1 | NM_001080449.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010847.hg.1 | NM_001080449.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010847.hg.1 | NM_001080449.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010847.hg.1 | NM_001080449.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010847.hg.1 | NM_001080449.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010847.hg.1 | NM_001080449.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010847.hg.1 | NM_001080449.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010847.hg.1 | NM_001080449.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010847.hg.1 | NM_001080449.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010847.hg.1 | NM_001080449.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010847.hg.1 | NM_001080449.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010847.hg.1 | NM_001080449.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010847.hg.1 | NM_001080449.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010847.hg.1 | NM_001080449.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010847.hg.1 | NM_001080449.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010847.hg.1 | NM_001080449.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010847.hg.1 | NR_102264.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010847.hg.1 | NR_102264.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010847.hg.1 | NR_102264.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010847.hg.1 | NR_102264.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010847.hg.1 | NR_102264.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010847.hg.1 | NR_102264.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010847.hg.1 | NR_102264.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010847.hg.1 | NR_102264.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010847.hg.1 | NR_102264.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010847.hg.1 | NR_102264.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010847.hg.1 | NR_102264.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010847.hg.1 | NR_102264.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010847.hg.1 | NR_102264.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010847.hg.1 | NR_102264.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010847.hg.1 | NR_102264.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010847.hg.1 | NR_102264.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010847.hg.1 | NR_102264.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010847.hg.1 | NR_102264.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010847.hg.1 | NR_102264.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010847.hg.1 | NR_102264.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010847.hg.1 | NR_102264.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010847.hg.1 | NR_102264.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010847.hg.1 | NR_102264.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010847.hg.1 | NR_102264.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010847.hg.1 | NR_102264.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010847.hg.1 | NR_102264.1 | Refseq |

DNA2
Genomic Map