Assay Details
| Gene Symbol: OSMR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001323505.1 | NP_001310434.1 | 16 - 17 | 2573 | 76 | |
| NM_001323506.1 | NP_001310435.1 | 16 - 17 | 2576 | 76 | ||
| NM_003999.2 | NP_003990.1 | 16 - 17 | 2693 | 76 | ||
| XM_005248384.1 | XP_005248441.1 | 16 - 17 | 2683 | 76 | ||
| XM_011514161.2 | XP_011512463.1 | 16 - 17 | 2683 | 76 | ||
| XM_017010019.1 | XP_016865508.1 | 16 - 17 | 2680 | 76 | ||
| XM_017010020.1 | XP_016865509.1 | 10 - 11 | 1383 | 76 | ||
| GenBank mRNA | BC125209.1 | - | 16 - 17 | 2463 | 40123501 | 76 |
| BC125210.1 | - | 16 - 17 | 2463 | 40123502 | 76 | |
| U60805.1 | - | 16 - 17 | 2658 | 76 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC05000179.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR05002114.hg.1 | TC05000179.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR05002114.hg.1 | ENST00000274276.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05002114.hg.1 | NM_003999.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007201.hg.1 | ENST00000274276.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007201.hg.1 | ENST00000274276.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007201.hg.1 | ENST00000274276.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007201.hg.1 | ENST00000274276.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007201.hg.1 | ENST00000274276.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007201.hg.1 | ENST00000274276.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007201.hg.1 | ENST00000274276.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007201.hg.1 | ENST00000274276.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007201.hg.1 | ENST00000274276.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007201.hg.1 | ENST00000274276.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007201.hg.1 | ENST00000274276.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007201.hg.1 | ENST00000274276.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007201.hg.1 | ENST00000274276.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007201.hg.1 | ENST00000274276.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007201.hg.1 | ENST00000274276.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007201.hg.1 | ENST00000274276.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007201.hg.1 | ENST00000274276.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007201.hg.1 | ENST00000274276.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007201.hg.1 | ENST00000274276.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007201.hg.1 | ENST00000274276.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007201.hg.1 | ENST00000274276.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007201.hg.1 | ENST00000274276.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007201.hg.1 | ENST00000274276.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007201.hg.1 | ENST00000502536.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007201.hg.1 | ENST00000502536.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007201.hg.1 | ENST00000502536.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007201.hg.1 | ENST00000502536.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007201.hg.1 | ENST00000502536.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007201.hg.1 | ENST00000502536.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007201.hg.1 | ENST00000502536.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007201.hg.1 | ENST00000502536.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007201.hg.1 | ENST00000502536.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007201.hg.1 | ENST00000502536.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007201.hg.1 | ENST00000508882.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007201.hg.1 | ENST00000508882.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007201.hg.1 | ENST00000508882.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007201.hg.1 | ENST00000509237.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007201.hg.1 | ENST00000509237.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007201.hg.1 | ENST00000509237.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007201.hg.1 | ENST00000509237.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007201.hg.1 | ENST00000513831.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007201.hg.1 | ENST00000513831.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007201.hg.1 | ENST00000513831.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007201.hg.1 | ENST00000513831.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007201.hg.1 | NM_001168355.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007201.hg.1 | NM_001168355.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007201.hg.1 | NM_001168355.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007201.hg.1 | NM_001168355.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007201.hg.1 | NM_001168355.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007201.hg.1 | NM_001168355.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007201.hg.1 | NM_001168355.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007201.hg.1 | NM_001168355.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007201.hg.1 | NM_001168355.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007201.hg.1 | NM_001168355.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007201.hg.1 | NM_003999.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007201.hg.1 | NM_003999.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007201.hg.1 | NM_003999.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007201.hg.1 | NM_003999.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007201.hg.1 | NM_003999.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007201.hg.1 | NM_003999.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007201.hg.1 | NM_003999.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007201.hg.1 | NM_003999.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007201.hg.1 | NM_003999.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007201.hg.1 | NM_003999.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007201.hg.1 | NM_003999.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007201.hg.1 | NM_003999.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007201.hg.1 | NM_003999.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007201.hg.1 | NM_003999.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007201.hg.1 | NM_003999.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007201.hg.1 | NM_003999.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007201.hg.1 | NM_003999.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007201.hg.1 | NM_003999.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007201.hg.1 | NM_003999.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007201.hg.1 | NM_003999.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007201.hg.1 | NM_003999.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007201.hg.1 | NM_003999.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007201.hg.1 | NM_003999.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500148742.hg.1 | ENST00000274276.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500148742.hg.1 | ENST00000508882.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500148742.hg.1 | ENST00000509237.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500148742.hg.1 | NM_003999.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500148742.hg.1 | ENST00000274276.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500148742.hg.1 | ENST00000508882.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500148742.hg.1 | ENST00000509237.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500148742.hg.1 | NM_003999.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500148742.hg.1 | TC0500007201.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0500007201.hg.1 | ENST00000274276.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007201.hg.1 | ENST00000274276.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007201.hg.1 | ENST00000274276.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007201.hg.1 | ENST00000274276.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007201.hg.1 | ENST00000274276.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007201.hg.1 | ENST00000274276.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007201.hg.1 | ENST00000274276.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007201.hg.1 | ENST00000274276.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007201.hg.1 | ENST00000274276.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007201.hg.1 | ENST00000274276.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007201.hg.1 | ENST00000274276.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007201.hg.1 | ENST00000274276.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007201.hg.1 | ENST00000274276.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007201.hg.1 | ENST00000274276.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007201.hg.1 | ENST00000274276.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007201.hg.1 | ENST00000274276.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007201.hg.1 | ENST00000274276.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007201.hg.1 | ENST00000274276.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007201.hg.1 | ENST00000274276.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007201.hg.1 | ENST00000274276.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007201.hg.1 | ENST00000274276.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007201.hg.1 | ENST00000274276.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007201.hg.1 | ENST00000274276.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007201.hg.1 | ENST00000502536.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007201.hg.1 | ENST00000502536.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007201.hg.1 | ENST00000502536.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007201.hg.1 | ENST00000502536.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007201.hg.1 | ENST00000502536.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007201.hg.1 | ENST00000502536.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007201.hg.1 | ENST00000502536.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007201.hg.1 | ENST00000502536.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007201.hg.1 | ENST00000502536.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007201.hg.1 | ENST00000502536.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007201.hg.1 | ENST00000508882.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007201.hg.1 | ENST00000508882.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007201.hg.1 | ENST00000508882.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007201.hg.1 | ENST00000509237.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007201.hg.1 | ENST00000509237.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007201.hg.1 | ENST00000509237.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007201.hg.1 | ENST00000509237.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007201.hg.1 | ENST00000513831.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007201.hg.1 | ENST00000513831.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007201.hg.1 | ENST00000513831.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007201.hg.1 | ENST00000513831.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007201.hg.1 | NM_001168355.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007201.hg.1 | NM_001168355.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007201.hg.1 | NM_001168355.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007201.hg.1 | NM_001168355.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007201.hg.1 | NM_001168355.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007201.hg.1 | NM_001168355.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007201.hg.1 | NM_001168355.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007201.hg.1 | NM_001168355.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007201.hg.1 | NM_001168355.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007201.hg.1 | NM_001168355.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007201.hg.1 | NM_003999.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007201.hg.1 | NM_003999.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007201.hg.1 | NM_003999.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007201.hg.1 | NM_003999.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007201.hg.1 | NM_003999.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007201.hg.1 | NM_003999.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007201.hg.1 | NM_003999.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007201.hg.1 | NM_003999.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007201.hg.1 | NM_003999.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007201.hg.1 | NM_003999.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007201.hg.1 | NM_003999.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007201.hg.1 | NM_003999.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007201.hg.1 | NM_003999.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007201.hg.1 | NM_003999.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007201.hg.1 | NM_003999.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007201.hg.1 | NM_003999.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007201.hg.1 | NM_003999.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007201.hg.1 | NM_003999.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007201.hg.1 | NM_003999.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007201.hg.1 | NM_003999.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007201.hg.1 | NM_003999.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007201.hg.1 | NM_003999.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007201.hg.1 | NM_003999.2 | Refseq |

OSMR
Genomic Map