Assay Details
| Gene Symbol: CA9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001216.2 | NP_001207.2 | 5 - 6 | 884 | 56 | |
| GenBank mRNA | AJ010588.1 | - | 5 - 6 | 851 | 56 | |
| BC014950.1 | - | 5 - 6 | 946 | 4865275 | 56 | |
| EF122496.1 | - | 5 - 6 | 884 | 56 | ||
| KU177952.1 | - | 5 - 6 | 842 | 56 | ||
| X66839.1 | - | 5 - 6 | 884 | 56 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC09000191.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR09002137.hg.1 | TC09000191.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR09002137.hg.1 | ENST00000378357.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09002137.hg.1 | ENST00000493245.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09002137.hg.1 | ENST00000544074.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09002137.hg.1 | NM_001216.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007094.hg.1 | ENST00000378357.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007094.hg.1 | ENST00000378357.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007094.hg.1 | ENST00000378357.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007094.hg.1 | ENST00000378357.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007094.hg.1 | ENST00000378357.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007094.hg.1 | ENST00000378357.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007094.hg.1 | ENST00000378357.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007094.hg.1 | ENST00000378357.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007094.hg.1 | ENST00000378357.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007094.hg.1 | ENST00000378357.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007094.hg.1 | ENST00000378357.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007094.hg.1 | ENST00000378357.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007094.hg.1 | ENST00000378357.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007094.hg.1 | ENST00000378357.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007094.hg.1 | ENST00000378357.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007094.hg.1 | ENST00000485665.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007094.hg.1 | ENST00000485665.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007094.hg.1 | ENST00000485665.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007094.hg.1 | ENST00000485665.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007094.hg.1 | ENST00000493245.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007094.hg.1 | ENST00000493245.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007094.hg.1 | ENST00000493245.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007094.hg.1 | ENST00000493245.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007094.hg.1 | ENST00000493245.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007094.hg.1 | ENST00000493245.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007094.hg.1 | ENST00000493245.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007094.hg.1 | ENST00000493245.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007094.hg.1 | ENST00000617161.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007094.hg.1 | ENST00000617161.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007094.hg.1 | ENST00000617161.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007094.hg.1 | ENST00000617161.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007094.hg.1 | ENST00000617161.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007094.hg.1 | ENST00000617161.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007094.hg.1 | ENST00000617161.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007094.hg.1 | ENST00000617161.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007094.hg.1 | ENST00000617161.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007094.hg.1 | ENST00000617161.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007094.hg.1 | ENST00000617161.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007094.hg.1 | ENST00000617161.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007094.hg.1 | NM_001216.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007094.hg.1 | NM_001216.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007094.hg.1 | NM_001216.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007094.hg.1 | NM_001216.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007094.hg.1 | NM_001216.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007094.hg.1 | NM_001216.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007094.hg.1 | NM_001216.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007094.hg.1 | NM_001216.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007094.hg.1 | NM_001216.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007094.hg.1 | NM_001216.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007094.hg.1 | NM_001216.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007094.hg.1 | NM_001216.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007094.hg.1 | NM_001216.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007094.hg.1 | NM_001216.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900007094.hg.1 | ENST00000378357.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007094.hg.1 | ENST00000378357.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007094.hg.1 | ENST00000378357.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007094.hg.1 | ENST00000378357.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007094.hg.1 | ENST00000378357.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007094.hg.1 | ENST00000378357.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007094.hg.1 | ENST00000378357.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007094.hg.1 | ENST00000378357.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007094.hg.1 | ENST00000378357.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007094.hg.1 | ENST00000378357.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007094.hg.1 | ENST00000378357.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007094.hg.1 | ENST00000378357.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007094.hg.1 | ENST00000378357.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007094.hg.1 | ENST00000378357.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007094.hg.1 | ENST00000378357.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007094.hg.1 | ENST00000485665.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007094.hg.1 | ENST00000485665.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007094.hg.1 | ENST00000485665.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007094.hg.1 | ENST00000485665.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007094.hg.1 | ENST00000493245.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007094.hg.1 | ENST00000493245.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007094.hg.1 | ENST00000493245.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007094.hg.1 | ENST00000493245.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007094.hg.1 | ENST00000493245.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007094.hg.1 | ENST00000493245.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007094.hg.1 | ENST00000493245.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007094.hg.1 | ENST00000493245.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007094.hg.1 | ENST00000617161.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007094.hg.1 | ENST00000617161.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007094.hg.1 | ENST00000617161.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007094.hg.1 | ENST00000617161.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007094.hg.1 | ENST00000617161.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007094.hg.1 | ENST00000617161.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007094.hg.1 | ENST00000617161.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007094.hg.1 | ENST00000617161.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007094.hg.1 | ENST00000617161.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007094.hg.1 | ENST00000617161.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007094.hg.1 | ENST00000617161.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007094.hg.1 | ENST00000617161.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007094.hg.1 | NM_001216.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900007094.hg.1 | NM_001216.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900007094.hg.1 | NM_001216.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900007094.hg.1 | NM_001216.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900007094.hg.1 | NM_001216.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900007094.hg.1 | NM_001216.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900007094.hg.1 | NM_001216.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900007094.hg.1 | NM_001216.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900007094.hg.1 | NM_001216.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900007094.hg.1 | NM_001216.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900007094.hg.1 | NM_001216.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900007094.hg.1 | NM_001216.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900007094.hg.1 | NM_001216.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900007094.hg.1 | NM_001216.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900148211.hg.1 | ENST00000378357.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900148211.hg.1 | ENST00000493245.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900148211.hg.1 | ENST00000617161.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900148211.hg.1 | NM_001216.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900148211.hg.1 | ENST00000378357.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900148211.hg.1 | ENST00000493245.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900148211.hg.1 | ENST00000617161.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900148211.hg.1 | NM_001216.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900148211.hg.1 | TC0900007094.hg.1 | Clariom D TC |

CA9
Genomic Map