Assay Details
| Gene Symbol: PGM2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_018290.3 | NP_060760.2 | 12 - 13 | 1678 | 85 | |
| XM_011513711.2 | XP_011512013.1 | 13 - 14 | 1781 | 85 | ||
| GenBank mRNA | AF109360.1 | - | 12 - 13 | 1618 | 85 | |
| AK001845.1 | - | 12 - 13 | 1621 | 85 | ||
| AK223237.1 | - | 12 - 13 | 1620 | 85 | ||
| AK297752.1 | - | 10 - 11 | 1415 | 85 | ||
| AL136705.1 | - | 12 - 13 | 1635 | 85 | ||
| BC010087.2 | - | 12 - 13 | 1618 | 2964390 | 85 | |
| CR457274.1 | - | 12 - 13 | 1605 | 85 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC04000213.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000381967.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000381967.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000381967.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000381967.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000381967.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000381967.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000381967.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000381967.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000381967.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000381967.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000381967.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000381967.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000381967.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000381967.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000381967.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000381967.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000381967.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000381967.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000381967.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000381967.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000381967.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000505986.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000505986.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000505986.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000505986.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000505986.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000505986.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000505986.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000505986.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000505986.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000505986.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000505986.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000505986.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000505986.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000505986.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000510084.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000510084.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000510084.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000510084.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000510084.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000512556.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000512556.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000512556.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000512556.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000512556.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000512556.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000512556.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000512556.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000512556.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000512556.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000512556.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000512556.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000512556.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000512556.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000512556.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000512556.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000515668.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000515668.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000515668.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000515668.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000515668.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000515668.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000515668.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000515668.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000515668.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000515668.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000515668.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000515668.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000515668.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000625312.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000625312.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000625312.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000625312.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000625312.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000625312.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | NM_018290.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | NM_018290.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | NM_018290.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | NM_018290.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | NM_018290.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | NM_018290.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | NM_018290.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | NM_018290.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | NM_018290.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | NM_018290.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | NM_018290.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | NM_018290.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | NM_018290.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | NM_018290.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | NM_018290.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | NM_018290.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | NM_018290.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | NM_018290.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | NM_018290.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | NM_018290.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR04003154.hg.1 | TC04000213.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR04003154.hg.1 | ENST00000381967.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04003154.hg.1 | ENST00000537241.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04003154.hg.1 | NM_018290.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400149497.hg.1 | ENST00000381967.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400149497.hg.1 | ENST00000510084.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400149497.hg.1 | ENST00000512556.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400149497.hg.1 | NM_018290.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400149497.hg.1 | ENST00000381967.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400149497.hg.1 | ENST00000510084.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400149497.hg.1 | ENST00000512556.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400149497.hg.1 | NM_018290.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400149497.hg.1 | TC0400007243.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000381967.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000381967.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000381967.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000381967.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000381967.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000381967.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000381967.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000381967.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000381967.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000381967.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000381967.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000381967.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000381967.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000381967.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000381967.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000381967.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000381967.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000381967.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000381967.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000381967.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000381967.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000505986.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000505986.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000505986.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000505986.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000505986.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000505986.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000505986.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000505986.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000505986.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000505986.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000505986.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000505986.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000505986.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000505986.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000510084.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000510084.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000510084.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000510084.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000510084.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000512556.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000512556.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000512556.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000512556.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000512556.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000512556.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000512556.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000512556.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000512556.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000512556.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000512556.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000512556.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000512556.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000512556.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000512556.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000512556.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000515668.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000515668.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000515668.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000515668.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000515668.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000515668.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000515668.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000515668.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000515668.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000515668.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000515668.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000515668.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000515668.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000625312.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000625312.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000625312.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000625312.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000625312.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000625312.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | NM_018290.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | NM_018290.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | NM_018290.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | NM_018290.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | NM_018290.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | NM_018290.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | NM_018290.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | NM_018290.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | NM_018290.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | NM_018290.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | NM_018290.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | NM_018290.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | NM_018290.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | NM_018290.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | NM_018290.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | NM_018290.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | NM_018290.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | NM_018290.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | NM_018290.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | NM_018290.3 | Refseq |

PGM2
Genomic Map