Assay Details
| Gene Symbol: DUSP9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001395.3 | NP_001386.1 | 1 - 2 | 171 | 62 | |
| GenBank mRNA | AK314250.1 | - | 1 - 2 | 171 | 62 | |
| Y08302.1 | - | 1 - 2 | 80 | 62 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC0X000753.hg.1
Additional Information:
This assay is in the 5'UTR region of the AK314250.1,NM_001395.3,Y08302.1 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
MAPK cascade
activation of MAPK activity
inactivation of MAPK activity
protein dephosphorylation
JNK cascade
peptidyl-tyrosine dephosphorylation
phosphoprotein phosphatase activity
protein tyrosine phosphatase activity
protein binding
protein tyrosine/serine/threonine phosphatase activity
MAP kinase tyrosine/serine/threonine phosphatase activity
MAPK cascade
activation of MAPK activity
inactivation of MAPK activity
protein dephosphorylation
JNK cascade
peptidyl-tyrosine dephosphorylation
phosphoprotein phosphatase activity
protein tyrosine phosphatase activity
protein binding
protein tyrosine/serine/threonine phosphatase activity
MAP kinase tyrosine/serine/threonine phosphatase activity
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008790.hg.1 | ENST00000342782.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008790.hg.1 | ENST00000342782.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008790.hg.1 | ENST00000342782.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008790.hg.1 | ENST00000342782.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008790.hg.1 | ENST00000342782.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008790.hg.1 | ENST00000342782.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008790.hg.1 | ENST00000342782.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008790.hg.1 | ENST00000342782.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008790.hg.1 | ENST00000342782.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008790.hg.1 | ENST00000370167.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008790.hg.1 | ENST00000370167.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008790.hg.1 | ENST00000370167.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008790.hg.1 | ENST00000370167.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008790.hg.1 | ENST00000370167.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008790.hg.1 | ENST00000370167.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008790.hg.1 | ENST00000370167.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008790.hg.1 | ENST00000370167.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008790.hg.1 | ENST00000370167.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008790.hg.1 | ENST00000477033.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008790.hg.1 | ENST00000477033.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008790.hg.1 | ENST00000477033.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008790.hg.1 | ENST00000477033.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008790.hg.1 | ENST00000477033.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008790.hg.1 | NM_001395.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008790.hg.1 | NM_001395.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008790.hg.1 | NM_001395.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008790.hg.1 | NM_001395.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008790.hg.1 | NM_001395.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008790.hg.1 | NM_001395.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008790.hg.1 | NM_001395.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008790.hg.1 | NM_001395.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008790.hg.1 | NM_001395.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008790.hg.1 | NM_001395.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008790.hg.1 | NM_001395.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X009806.hg.1 | TC0X000753.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X009806.hg.1 | ENST00000370167.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X009806.hg.1 | ENST00000477033.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X009806.hg.1 | NM_001395.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X009805.hg.1 | TC0X000753.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X009805.hg.1 | ENST00000477033.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X009805.hg.1 | NM_001395.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X009804.hg.1 | TC0X000753.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X009804.hg.1 | NM_001395.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X009811.hg.1 | TC0X000753.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X009811.hg.1 | ENST00000342782.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X009811.hg.1 | ENST00000370167.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X009811.hg.1 | ENST00000477033.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X009811.hg.1 | NM_001395.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X009812.hg.1 | TC0X000753.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X009812.hg.1 | ENST00000342782.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X009812.hg.1 | ENST00000370167.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X009812.hg.1 | NM_001395.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00159849.hg.1 | ENST00000370167.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00159849.hg.1 | ENST00000477033.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00159849.hg.1 | NM_001395.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00159849.hg.1 | ENST00000370167.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00159849.hg.1 | ENST00000477033.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00159849.hg.1 | NM_001395.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00159849.hg.1 | TC0X00008790.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0X00159854.hg.1 | ENST00000342782.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00159854.hg.1 | ENST00000370167.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00159854.hg.1 | ENST00000477033.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00159854.hg.1 | NM_001395.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00159854.hg.1 | ENST00000342782.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00159854.hg.1 | ENST00000370167.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00159854.hg.1 | ENST00000477033.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00159854.hg.1 | NM_001395.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00159854.hg.1 | TC0X00008790.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0X00159855.hg.1 | ENST00000342782.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00159855.hg.1 | ENST00000370167.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00159855.hg.1 | NM_001395.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00159855.hg.1 | ENST00000342782.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00159855.hg.1 | ENST00000370167.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00159855.hg.1 | NM_001395.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00159855.hg.1 | TC0X00008790.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0X00159847.hg.1 | NM_001395.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00159847.hg.1 | NM_001395.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00159847.hg.1 | TC0X00008790.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0X00159848.hg.1 | ENST00000477033.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00159848.hg.1 | NM_001395.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00159848.hg.1 | ENST00000477033.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00159848.hg.1 | NM_001395.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00159848.hg.1 | TC0X00008790.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0X00008790.hg.1 | ENST00000342782.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008790.hg.1 | ENST00000342782.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008790.hg.1 | ENST00000342782.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008790.hg.1 | ENST00000342782.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008790.hg.1 | ENST00000342782.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008790.hg.1 | ENST00000342782.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008790.hg.1 | ENST00000342782.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008790.hg.1 | ENST00000342782.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008790.hg.1 | ENST00000342782.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008790.hg.1 | ENST00000370167.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008790.hg.1 | ENST00000370167.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008790.hg.1 | ENST00000370167.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008790.hg.1 | ENST00000370167.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008790.hg.1 | ENST00000370167.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008790.hg.1 | ENST00000370167.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008790.hg.1 | ENST00000370167.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008790.hg.1 | ENST00000370167.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008790.hg.1 | ENST00000370167.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008790.hg.1 | ENST00000477033.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008790.hg.1 | ENST00000477033.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008790.hg.1 | ENST00000477033.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008790.hg.1 | ENST00000477033.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008790.hg.1 | ENST00000477033.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008790.hg.1 | NM_001395.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008790.hg.1 | NM_001395.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008790.hg.1 | NM_001395.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008790.hg.1 | NM_001395.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008790.hg.1 | NM_001395.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008790.hg.1 | NM_001395.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008790.hg.1 | NM_001395.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008790.hg.1 | NM_001395.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008790.hg.1 | NM_001395.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008790.hg.1 | NM_001395.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008790.hg.1 | NM_001395.3 | Refseq |

DUSP9
Genomic Map