Assay Details
| Gene Symbol: WEE2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001105558.1 | NP_001099028.1 | 1 - 2 | 747 | 75 | |
| GenBank mRNA | AK131218.1 | - | 1 - 2 | 747 | 75 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC07000890.hg.1
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
mitotic nuclear division
female meiotic division
peptidyl-tyrosine phosphorylation
female pronucleus assembly
negative regulation of cyclin-dependent protein serine/threonine kinase activity
regulation of meiosis I
negative regulation of oocyte maturation
magnesium ion binding
non-membrane spanning protein tyrosine kinase activity
ATP binding
mitotic nuclear division
female meiotic division
peptidyl-tyrosine phosphorylation
female pronucleus assembly
negative regulation of cyclin-dependent protein serine/threonine kinase activity
regulation of meiosis I
negative regulation of oocyte maturation
magnesium ion binding
non-membrane spanning protein tyrosine kinase activity
ATP binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR07013851.hg.1 | TC07000890.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR07013851.hg.1 | ENST00000397541.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07013851.hg.1 | NM_001105558.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR07013852.hg.1 | TC07000890.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR07013852.hg.1 | ENST00000397541.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07013852.hg.1 | NM_001105558.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR07013853.hg.1 | TC07000890.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR07013853.hg.1 | ENST00000397541.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07013853.hg.1 | NM_001105558.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009395.hg.1 | ENST00000397541.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009395.hg.1 | ENST00000397541.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009395.hg.1 | ENST00000397541.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009395.hg.1 | ENST00000397541.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009395.hg.1 | ENST00000397541.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009395.hg.1 | ENST00000397541.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009395.hg.1 | ENST00000397541.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009395.hg.1 | ENST00000397541.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009395.hg.1 | ENST00000397541.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009395.hg.1 | ENST00000397541.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009395.hg.1 | ENST00000397541.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009395.hg.1 | ENST00000397541.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009395.hg.1 | ENST00000397541.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009395.hg.1 | ENST00000397541.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009395.hg.1 | ENST00000397541.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009395.hg.1 | ENST00000397541.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009395.hg.1 | ENST00000493845.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009395.hg.1 | ENST00000493845.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009395.hg.1 | ENST00000493845.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009395.hg.1 | ENST00000493845.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009395.hg.1 | ENST00000493845.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009395.hg.1 | ENST00000493845.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009395.hg.1 | NM_001105558.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009395.hg.1 | NM_001105558.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009395.hg.1 | NM_001105558.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009395.hg.1 | NM_001105558.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009395.hg.1 | NM_001105558.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009395.hg.1 | NM_001105558.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009395.hg.1 | NM_001105558.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009395.hg.1 | NM_001105558.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009395.hg.1 | NM_001105558.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009395.hg.1 | NM_001105558.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009395.hg.1 | NM_001105558.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009395.hg.1 | NM_001105558.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009395.hg.1 | NM_001105558.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009395.hg.1 | NM_001105558.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009395.hg.1 | NM_001105558.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009395.hg.1 | NM_001105558.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR07013850.hg.1 | TC07000890.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR07013850.hg.1 | ENST00000397541.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07013850.hg.1 | NM_001105558.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010541.hg.1 | ENST00000529830.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700166656.hg.1 | ENST00000397541.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700166656.hg.1 | NM_001105558.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700166656.hg.1 | ENST00000397541.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700166656.hg.1 | NM_001105558.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700166656.hg.1 | TC0700009395.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0700166657.hg.1 | ENST00000397541.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700166657.hg.1 | NM_001105558.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700166657.hg.1 | ENST00000397541.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700166657.hg.1 | NM_001105558.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700166657.hg.1 | TC0700009395.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0700166655.hg.1 | ENST00000397541.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700166655.hg.1 | NM_001105558.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700166655.hg.1 | ENST00000397541.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700166655.hg.1 | NM_001105558.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700166655.hg.1 | TC0700009395.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1100010541.hg.1 | ENST00000529830.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009395.hg.1 | ENST00000397541.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009395.hg.1 | ENST00000397541.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009395.hg.1 | ENST00000397541.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009395.hg.1 | ENST00000397541.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009395.hg.1 | ENST00000397541.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009395.hg.1 | ENST00000397541.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009395.hg.1 | ENST00000397541.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009395.hg.1 | ENST00000397541.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009395.hg.1 | ENST00000397541.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009395.hg.1 | ENST00000397541.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009395.hg.1 | ENST00000397541.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009395.hg.1 | ENST00000397541.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009395.hg.1 | ENST00000397541.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009395.hg.1 | ENST00000397541.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009395.hg.1 | ENST00000397541.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009395.hg.1 | ENST00000397541.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009395.hg.1 | ENST00000493845.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009395.hg.1 | ENST00000493845.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009395.hg.1 | ENST00000493845.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009395.hg.1 | ENST00000493845.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009395.hg.1 | ENST00000493845.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009395.hg.1 | ENST00000493845.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009395.hg.1 | NM_001105558.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009395.hg.1 | NM_001105558.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009395.hg.1 | NM_001105558.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009395.hg.1 | NM_001105558.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009395.hg.1 | NM_001105558.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009395.hg.1 | NM_001105558.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009395.hg.1 | NM_001105558.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009395.hg.1 | NM_001105558.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009395.hg.1 | NM_001105558.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009395.hg.1 | NM_001105558.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009395.hg.1 | NM_001105558.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009395.hg.1 | NM_001105558.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009395.hg.1 | NM_001105558.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009395.hg.1 | NM_001105558.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009395.hg.1 | NM_001105558.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009395.hg.1 | NM_001105558.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700166654.hg.1 | ENST00000397541.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700166654.hg.1 | NM_001105558.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700166654.hg.1 | ENST00000397541.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700166654.hg.1 | NM_001105558.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700166654.hg.1 | TC0700009395.hg.1 | Clariom D TC |

WEE2
Genomic Map