Assay Details
| Gene Symbol: SRRD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001013694.2 | NP_001013716.2 | 4 - 5 | 612 | 61 | |
| XM_011530178.2 | XP_011528480.1 | 4 - 5 | 675 | 61 | ||
| GenBank mRNA | AK293916.1 | - | 5 - 6 | 592 | 61 | |
| BC066962.1 | - | 5 - 6 | 595 | 4820477 | 61 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC22000170.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR22003229.hg.1 | ENST00000215917.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22003229.hg.1 | NM_001013694.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR22003229.hg.1 | TC22000170.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006950.hg.1 | ENST00000215917.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006950.hg.1 | ENST00000215917.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006950.hg.1 | ENST00000215917.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006950.hg.1 | ENST00000215917.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006950.hg.1 | ENST00000215917.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006950.hg.1 | ENST00000215917.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006950.hg.1 | ENST00000215917.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006950.hg.1 | ENST00000215917.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006950.hg.1 | ENST00000215917.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006950.hg.1 | ENST00000215917.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006950.hg.1 | ENST00000215917.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006950.hg.1 | ENST00000215917.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006950.hg.1 | ENST00000215917.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006950.hg.1 | ENST00000215917.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006950.hg.1 | ENST00000471799.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006950.hg.1 | ENST00000471799.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006950.hg.1 | ENST00000471799.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006950.hg.1 | ENST00000471799.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006950.hg.1 | ENST00000477945.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006950.hg.1 | ENST00000477945.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006950.hg.1 | ENST00000477945.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006950.hg.1 | ENST00000477945.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006950.hg.1 | ENST00000477945.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006950.hg.1 | ENST00000613968.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006950.hg.1 | ENST00000613968.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006950.hg.1 | ENST00000613968.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006950.hg.1 | ENST00000613968.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006950.hg.1 | ENST00000613968.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006950.hg.1 | ENST00000613968.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006950.hg.1 | ENST00000613968.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006950.hg.1 | ENST00000613968.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006950.hg.1 | ENST00000613968.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006950.hg.1 | ENST00000613968.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006950.hg.1 | NM_001013694.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006950.hg.1 | NM_001013694.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006950.hg.1 | NM_001013694.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006950.hg.1 | NM_001013694.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006950.hg.1 | NM_001013694.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006950.hg.1 | NM_001013694.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006950.hg.1 | NM_001013694.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006950.hg.1 | NM_001013694.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006950.hg.1 | NM_001013694.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006950.hg.1 | NM_001013694.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200006950.hg.1 | ENST00000215917.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006950.hg.1 | ENST00000215917.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006950.hg.1 | ENST00000215917.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006950.hg.1 | ENST00000215917.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006950.hg.1 | ENST00000215917.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006950.hg.1 | ENST00000215917.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006950.hg.1 | ENST00000215917.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006950.hg.1 | ENST00000215917.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006950.hg.1 | ENST00000215917.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006950.hg.1 | ENST00000215917.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006950.hg.1 | ENST00000215917.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006950.hg.1 | ENST00000215917.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006950.hg.1 | ENST00000215917.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006950.hg.1 | ENST00000215917.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006950.hg.1 | ENST00000471799.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006950.hg.1 | ENST00000471799.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006950.hg.1 | ENST00000471799.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006950.hg.1 | ENST00000471799.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006950.hg.1 | ENST00000477945.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006950.hg.1 | ENST00000477945.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006950.hg.1 | ENST00000477945.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006950.hg.1 | ENST00000477945.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006950.hg.1 | ENST00000477945.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006950.hg.1 | ENST00000613968.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006950.hg.1 | ENST00000613968.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006950.hg.1 | ENST00000613968.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006950.hg.1 | ENST00000613968.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006950.hg.1 | ENST00000613968.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006950.hg.1 | ENST00000613968.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006950.hg.1 | ENST00000613968.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006950.hg.1 | ENST00000613968.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006950.hg.1 | ENST00000613968.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006950.hg.1 | ENST00000613968.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006950.hg.1 | NM_001013694.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200006950.hg.1 | NM_001013694.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200006950.hg.1 | NM_001013694.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200006950.hg.1 | NM_001013694.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200006950.hg.1 | NM_001013694.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200006950.hg.1 | NM_001013694.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200006950.hg.1 | NM_001013694.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200006950.hg.1 | NM_001013694.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200006950.hg.1 | NM_001013694.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200006950.hg.1 | NM_001013694.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200148324.hg.1 | TC2200006950.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2200148324.hg.1 | ENST00000215917.11 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200148324.hg.1 | ENST00000477945.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200148324.hg.1 | ENST00000613968.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200148324.hg.1 | NM_001013694.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200148324.hg.1 | ENST00000215917.11 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200148324.hg.1 | ENST00000477945.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200148324.hg.1 | ENST00000613968.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200148324.hg.1 | NM_001013694.2 | Refseq |

SRRD
Genomic Map