Assay Details
Gene Symbol: ZNF516 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_014643.3 | NP_055458.1 | 1 - 2 | 44 | 66 | |
GenBank mRNA | D86975.1 | - | 1 - 2 | 44 | 66 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC18000581.hg.1
Additional Information:
This assay is in the 5'UTR region of the D86975.1,NM_014643.3 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
transcription from RNA polymerase II promoter
signal transduction
multicellular organism development
response to cold
positive regulation of transcription, DNA-templated
brown fat cell differentiation
adipose tissue development
core promoter proximal region sequence-specific DNA binding
transcription factor activity, sequence-specific DNA binding
activating transcription factor binding
sequence-specific DNA binding
transcription regulatory region DNA binding
metal ion binding
transcription from RNA polymerase II promoter
signal transduction
multicellular organism development
response to cold
positive regulation of transcription, DNA-templated
brown fat cell differentiation
adipose tissue development
core promoter proximal region sequence-specific DNA binding
transcription factor activity, sequence-specific DNA binding
activating transcription factor binding
sequence-specific DNA binding
transcription regulatory region DNA binding
metal ion binding
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR18006105.hg.1 | ENST00000443185.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR18006105.hg.1 | ENST00000524431.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR18006105.hg.1 | NM_014643.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR18006105.hg.1 | TC18000581.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR18006104.hg.1 | ENST00000443185.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR18006104.hg.1 | ENST00000524431.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR18006104.hg.1 | NM_014643.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR18006104.hg.1 | TC18000581.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC1800009086.hg.1 | ENST00000443185.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800009086.hg.1 | ENST00000443185.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800009086.hg.1 | ENST00000443185.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800009086.hg.1 | ENST00000443185.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800009086.hg.1 | ENST00000443185.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800009086.hg.1 | ENST00000443185.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800009086.hg.1 | ENST00000443185.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800009086.hg.1 | ENST00000443185.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800009086.hg.1 | ENST00000443185.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800009086.hg.1 | ENST00000443185.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800009086.hg.1 | ENST00000443185.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800009086.hg.1 | ENST00000443185.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800009086.hg.1 | ENST00000443185.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800009086.hg.1 | ENST00000443185.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800009086.hg.1 | ENST00000443185.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800009086.hg.1 | ENST00000443185.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800009086.hg.1 | ENST00000443185.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800009086.hg.1 | ENST00000443185.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800009086.hg.1 | ENST00000532857.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800009086.hg.1 | ENST00000532857.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800009086.hg.1 | ENST00000532857.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800009086.hg.1 | ENST00000542818.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800009086.hg.1 | ENST00000542818.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800009086.hg.1 | ENST00000542818.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800009086.hg.1 | ENST00000543743.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800009086.hg.1 | ENST00000543743.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800009086.hg.1 | ENST00000617840.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800009086.hg.1 | ENST00000617840.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800009086.hg.1 | ENST00000617840.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800009086.hg.1 | ENST00000617840.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800009086.hg.1 | ENST00000617840.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800009086.hg.1 | ENST00000617840.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800009086.hg.1 | NM_014643.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800009086.hg.1 | NM_014643.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800009086.hg.1 | NM_014643.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800009086.hg.1 | NM_014643.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800009086.hg.1 | NM_014643.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800009086.hg.1 | NM_014643.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800009086.hg.1 | NM_014643.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800009086.hg.1 | NM_014643.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800009086.hg.1 | NM_014643.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800009086.hg.1 | NM_014643.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800009086.hg.1 | NM_014643.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800009086.hg.1 | NM_014643.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800009086.hg.1 | NM_014643.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800009086.hg.1 | NM_014643.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800009086.hg.1 | NM_014643.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800009086.hg.1 | NM_014643.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800009086.hg.1 | NM_014643.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800009086.hg.1 | NM_014643.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800009086.hg.1 | NM_014643.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1800162246.hg.1 | ENST00000443185.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800162246.hg.1 | NM_014643.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1800162246.hg.1 | TC1800009086.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1800162246.hg.1 | ENST00000443185.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800162246.hg.1 | NM_014643.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1800162248.hg.1 | ENST00000443185.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800162248.hg.1 | NM_014643.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1800162248.hg.1 | TC1800009086.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1800162248.hg.1 | ENST00000443185.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800162248.hg.1 | NM_014643.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1800009086.hg.1 | ENST00000443185.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800009086.hg.1 | ENST00000443185.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800009086.hg.1 | ENST00000443185.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800009086.hg.1 | ENST00000443185.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800009086.hg.1 | ENST00000443185.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800009086.hg.1 | ENST00000443185.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800009086.hg.1 | ENST00000443185.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800009086.hg.1 | ENST00000443185.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800009086.hg.1 | ENST00000443185.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800009086.hg.1 | ENST00000443185.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800009086.hg.1 | ENST00000443185.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800009086.hg.1 | ENST00000443185.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800009086.hg.1 | ENST00000443185.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800009086.hg.1 | ENST00000443185.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800009086.hg.1 | ENST00000443185.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800009086.hg.1 | ENST00000443185.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800009086.hg.1 | ENST00000443185.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800009086.hg.1 | ENST00000443185.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800009086.hg.1 | ENST00000532857.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800009086.hg.1 | ENST00000532857.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800009086.hg.1 | ENST00000532857.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800009086.hg.1 | ENST00000542818.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800009086.hg.1 | ENST00000542818.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800009086.hg.1 | ENST00000542818.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800009086.hg.1 | ENST00000543743.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800009086.hg.1 | ENST00000543743.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800009086.hg.1 | ENST00000617840.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800009086.hg.1 | ENST00000617840.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800009086.hg.1 | ENST00000617840.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800009086.hg.1 | ENST00000617840.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800009086.hg.1 | ENST00000617840.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800009086.hg.1 | ENST00000617840.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800009086.hg.1 | NM_014643.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1800009086.hg.1 | NM_014643.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1800009086.hg.1 | NM_014643.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1800009086.hg.1 | NM_014643.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1800009086.hg.1 | NM_014643.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1800009086.hg.1 | NM_014643.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1800009086.hg.1 | NM_014643.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1800009086.hg.1 | NM_014643.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1800009086.hg.1 | NM_014643.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1800009086.hg.1 | NM_014643.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1800009086.hg.1 | NM_014643.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1800009086.hg.1 | NM_014643.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1800009086.hg.1 | NM_014643.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1800009086.hg.1 | NM_014643.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1800009086.hg.1 | NM_014643.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1800009086.hg.1 | NM_014643.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1800009086.hg.1 | NM_014643.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1800009086.hg.1 | NM_014643.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1800009086.hg.1 | NM_014643.3 | Refseq |
