Assay Details
| Gene Symbol: CPA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001868.3 | NP_001859.1 | 4 - 5 | 635 | 66 | |
| GenBank mRNA | AK131054.1 | - | 66 | |||
| AK291493.1 | - | 4 - 5 | 512 | 66 | ||
| AK293366.1 | - | 4 - 5 | 447 | 66 | ||
| BC005279.1 | - | 4 - 5 | 507 | 3949850 | 66 | |
| BT007313.1 | - | 4 - 5 | 485 | 66 | ||
| X67318.1 | - | 4 - 5 | 66 | |||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC07000814.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0700009140.hg.1 | ENST00000011292.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009140.hg.1 | ENST00000011292.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009140.hg.1 | ENST00000011292.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009140.hg.1 | ENST00000011292.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009140.hg.1 | ENST00000011292.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009140.hg.1 | ENST00000011292.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009140.hg.1 | ENST00000011292.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009140.hg.1 | ENST00000011292.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009140.hg.1 | ENST00000011292.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009140.hg.1 | ENST00000011292.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009140.hg.1 | ENST00000011292.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009140.hg.1 | ENST00000011292.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009140.hg.1 | ENST00000011292.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009140.hg.1 | ENST00000011292.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009140.hg.1 | ENST00000011292.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009140.hg.1 | ENST00000011292.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009140.hg.1 | ENST00000470838.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009140.hg.1 | ENST00000470838.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009140.hg.1 | ENST00000470838.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009140.hg.1 | ENST00000476062.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009140.hg.1 | ENST00000476062.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009140.hg.1 | ENST00000476062.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009140.hg.1 | ENST00000476062.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009140.hg.1 | ENST00000476062.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009140.hg.1 | ENST00000476062.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009140.hg.1 | ENST00000476062.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009140.hg.1 | ENST00000476062.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009140.hg.1 | ENST00000476062.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009140.hg.1 | ENST00000476062.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009140.hg.1 | ENST00000476062.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009140.hg.1 | ENST00000476062.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009140.hg.1 | ENST00000478096.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009140.hg.1 | ENST00000478096.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009140.hg.1 | ENST00000478096.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009140.hg.1 | ENST00000478096.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009140.hg.1 | ENST00000478096.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009140.hg.1 | ENST00000478096.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009140.hg.1 | ENST00000479106.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009140.hg.1 | ENST00000479106.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009140.hg.1 | ENST00000479106.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009140.hg.1 | ENST00000479106.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009140.hg.1 | ENST00000479106.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009140.hg.1 | ENST00000479106.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009140.hg.1 | ENST00000479106.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009140.hg.1 | ENST00000479106.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009140.hg.1 | ENST00000479106.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009140.hg.1 | ENST00000479106.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009140.hg.1 | ENST00000481342.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009140.hg.1 | ENST00000481342.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009140.hg.1 | ENST00000481342.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009140.hg.1 | ENST00000481342.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009140.hg.1 | ENST00000481342.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009140.hg.1 | ENST00000481342.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009140.hg.1 | ENST00000481342.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009140.hg.1 | ENST00000481342.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009140.hg.1 | ENST00000484324.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009140.hg.1 | ENST00000484324.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009140.hg.1 | ENST00000484324.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009140.hg.1 | ENST00000484324.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009140.hg.1 | ENST00000484324.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009140.hg.1 | ENST00000484324.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009140.hg.1 | ENST00000484324.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009140.hg.1 | ENST00000484324.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009140.hg.1 | ENST00000484324.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009140.hg.1 | ENST00000484324.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009140.hg.1 | ENST00000484324.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009140.hg.1 | ENST00000484324.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009140.hg.1 | ENST00000484324.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009140.hg.1 | ENST00000491460.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009140.hg.1 | ENST00000491460.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009140.hg.1 | ENST00000491460.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009140.hg.1 | ENST00000491460.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009140.hg.1 | ENST00000491460.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009140.hg.1 | ENST00000491460.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009140.hg.1 | ENST00000491460.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009140.hg.1 | ENST00000491460.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009140.hg.1 | ENST00000604896.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009140.hg.1 | ENST00000604896.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009140.hg.1 | ENST00000604896.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009140.hg.1 | ENST00000604896.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009140.hg.1 | ENST00000604896.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009140.hg.1 | ENST00000604896.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009140.hg.1 | NM_001868.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009140.hg.1 | NM_001868.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009140.hg.1 | NM_001868.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009140.hg.1 | NM_001868.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009140.hg.1 | NM_001868.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009140.hg.1 | NM_001868.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009140.hg.1 | NM_001868.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009140.hg.1 | NM_001868.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009140.hg.1 | NM_001868.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009140.hg.1 | NM_001868.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009140.hg.1 | NM_001868.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009140.hg.1 | NM_001868.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009140.hg.1 | NM_001868.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009140.hg.1 | NM_001868.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009140.hg.1 | NM_001868.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009140.hg.1 | NM_001868.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR07012830.hg.1 | TC07000814.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR07012830.hg.1 | ENST00000011292.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07012830.hg.1 | ENST00000484324.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07012830.hg.1 | NM_001868.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700165037.hg.1 | ENST00000011292.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700165037.hg.1 | ENST00000476062.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700165037.hg.1 | ENST00000481342.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700165037.hg.1 | ENST00000484324.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700165037.hg.1 | ENST00000491460.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700165037.hg.1 | NM_001868.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700165037.hg.1 | ENST00000011292.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700165037.hg.1 | ENST00000476062.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700165037.hg.1 | ENST00000481342.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700165037.hg.1 | ENST00000484324.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700165037.hg.1 | ENST00000491460.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700165037.hg.1 | NM_001868.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700165037.hg.1 | TC0700009140.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0700009140.hg.1 | ENST00000011292.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009140.hg.1 | ENST00000011292.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009140.hg.1 | ENST00000011292.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009140.hg.1 | ENST00000011292.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009140.hg.1 | ENST00000011292.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009140.hg.1 | ENST00000011292.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009140.hg.1 | ENST00000011292.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009140.hg.1 | ENST00000011292.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009140.hg.1 | ENST00000011292.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009140.hg.1 | ENST00000011292.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009140.hg.1 | ENST00000011292.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009140.hg.1 | ENST00000011292.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009140.hg.1 | ENST00000011292.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009140.hg.1 | ENST00000011292.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009140.hg.1 | ENST00000011292.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009140.hg.1 | ENST00000011292.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009140.hg.1 | ENST00000470838.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009140.hg.1 | ENST00000470838.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009140.hg.1 | ENST00000470838.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009140.hg.1 | ENST00000476062.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009140.hg.1 | ENST00000476062.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009140.hg.1 | ENST00000476062.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009140.hg.1 | ENST00000476062.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009140.hg.1 | ENST00000476062.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009140.hg.1 | ENST00000476062.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009140.hg.1 | ENST00000476062.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009140.hg.1 | ENST00000476062.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009140.hg.1 | ENST00000476062.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009140.hg.1 | ENST00000476062.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009140.hg.1 | ENST00000476062.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009140.hg.1 | ENST00000476062.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009140.hg.1 | ENST00000478096.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009140.hg.1 | ENST00000478096.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009140.hg.1 | ENST00000478096.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009140.hg.1 | ENST00000478096.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009140.hg.1 | ENST00000478096.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009140.hg.1 | ENST00000478096.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009140.hg.1 | ENST00000479106.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009140.hg.1 | ENST00000479106.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009140.hg.1 | ENST00000479106.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009140.hg.1 | ENST00000479106.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009140.hg.1 | ENST00000479106.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009140.hg.1 | ENST00000479106.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009140.hg.1 | ENST00000479106.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009140.hg.1 | ENST00000479106.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009140.hg.1 | ENST00000479106.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009140.hg.1 | ENST00000479106.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009140.hg.1 | ENST00000481342.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009140.hg.1 | ENST00000481342.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009140.hg.1 | ENST00000481342.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009140.hg.1 | ENST00000481342.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009140.hg.1 | ENST00000481342.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009140.hg.1 | ENST00000481342.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009140.hg.1 | ENST00000481342.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009140.hg.1 | ENST00000481342.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009140.hg.1 | ENST00000484324.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009140.hg.1 | ENST00000484324.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009140.hg.1 | ENST00000484324.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009140.hg.1 | ENST00000484324.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009140.hg.1 | ENST00000484324.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009140.hg.1 | ENST00000484324.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009140.hg.1 | ENST00000484324.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009140.hg.1 | ENST00000484324.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009140.hg.1 | ENST00000484324.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009140.hg.1 | ENST00000484324.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009140.hg.1 | ENST00000484324.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009140.hg.1 | ENST00000484324.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009140.hg.1 | ENST00000484324.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009140.hg.1 | ENST00000491460.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009140.hg.1 | ENST00000491460.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009140.hg.1 | ENST00000491460.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009140.hg.1 | ENST00000491460.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009140.hg.1 | ENST00000491460.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009140.hg.1 | ENST00000491460.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009140.hg.1 | ENST00000491460.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009140.hg.1 | ENST00000491460.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009140.hg.1 | ENST00000604896.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009140.hg.1 | ENST00000604896.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009140.hg.1 | ENST00000604896.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009140.hg.1 | ENST00000604896.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009140.hg.1 | ENST00000604896.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009140.hg.1 | ENST00000604896.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009140.hg.1 | NM_001868.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009140.hg.1 | NM_001868.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009140.hg.1 | NM_001868.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009140.hg.1 | NM_001868.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009140.hg.1 | NM_001868.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009140.hg.1 | NM_001868.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009140.hg.1 | NM_001868.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009140.hg.1 | NM_001868.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009140.hg.1 | NM_001868.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009140.hg.1 | NM_001868.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009140.hg.1 | NM_001868.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009140.hg.1 | NM_001868.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009140.hg.1 | NM_001868.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009140.hg.1 | NM_001868.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009140.hg.1 | NM_001868.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009140.hg.1 | NM_001868.3 | Refseq |

CPA1
Genomic Map