Assay Details
| Gene Symbol: TDRD12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_011527471.2 | XP_011525773.1 | 15 - 16 | 1696 | 119 | |
| XM_011527472.2 | XP_011525774.1 | 15 - 16 | 1696 | 119 | ||
| XM_011527473.2 | XP_011525775.1 | 15 - 16 | 1696 | 119 | ||
| XM_011527474.2 | XP_011525776.1 | 14 - 15 | 1576 | 119 | ||
| XM_011527475.2 | XP_011525777.1 | 15 - 16 | 1696 | 119 | ||
| XM_011527476.2 | XP_011525778.1 | 15 - 16 | 1696 | 119 | ||
| XM_017027458.1 | XP_016882947.1 | 15 - 16 | 1696 | 119 | ||
| XM_017027459.1 | XP_016882948.1 | 5 - 6 | 679 | 119 | ||
| GenBank mRNA | BC049000.1 | - | 15 - 16 | 1936 | 119 | |
More Information
Set Membership: |
|

TDRD12
Genomic Map