Assay Details
| Gene Symbol: NQO2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_000904.4 | NP_000895.2 | 5 - 6 | 770 | 79 | |
| NM_001290221.1 | NP_001277150.1 | 8 - 9 | 1285 | 79 | ||
| NM_001318940.1 | NP_001305869.1 | 5 - 6 | 710 | 79 | ||
| GenBank mRNA | AK056981.1 | - | 8 - 9 | 1127 | 79 | |
| AK311746.1 | - | 5 - 6 | 729 | 79 | ||
| BC006096.2 | - | 4 - 5 | 501 | 3507945 | 79 | |
| J02888.1 | - | 5 - 6 | 590 | 79 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC06004062.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR06000153.hg.1 | TC06004062.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR06000153.hg.1 | ENST00000338130.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06000153.hg.1 | ENST00000380430.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06000153.hg.1 | ENST00000380441.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06000153.hg.1 | ENST00000380454.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06000153.hg.1 | ENST00000380455.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06000153.hg.1 | NM_000904.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014055.hg.1 | ENST00000338130.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014055.hg.1 | ENST00000338130.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014055.hg.1 | ENST00000338130.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014055.hg.1 | ENST00000338130.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014055.hg.1 | ENST00000338130.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014055.hg.1 | ENST00000338130.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014055.hg.1 | ENST00000338130.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014055.hg.1 | ENST00000338130.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014055.hg.1 | ENST00000338130.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014055.hg.1 | ENST00000338130.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014055.hg.1 | ENST00000338130.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014055.hg.1 | ENST00000338130.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014055.hg.1 | ENST00000380430.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014055.hg.1 | ENST00000380430.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014055.hg.1 | ENST00000380430.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014055.hg.1 | ENST00000380430.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014055.hg.1 | ENST00000380430.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014055.hg.1 | ENST00000380430.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014055.hg.1 | ENST00000380430.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014055.hg.1 | ENST00000380430.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014055.hg.1 | ENST00000380430.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014055.hg.1 | ENST00000380441.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014055.hg.1 | ENST00000380441.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014055.hg.1 | ENST00000380441.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014055.hg.1 | ENST00000380441.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014055.hg.1 | ENST00000380441.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014055.hg.1 | ENST00000380441.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014055.hg.1 | ENST00000380441.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014055.hg.1 | ENST00000380441.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014055.hg.1 | ENST00000380441.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014055.hg.1 | ENST00000380454.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014055.hg.1 | ENST00000380454.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014055.hg.1 | ENST00000380454.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014055.hg.1 | ENST00000380454.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014055.hg.1 | ENST00000380454.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014055.hg.1 | ENST00000380454.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014055.hg.1 | ENST00000380454.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014055.hg.1 | ENST00000380454.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014055.hg.1 | ENST00000397717.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014055.hg.1 | ENST00000397717.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014055.hg.1 | ENST00000397717.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014055.hg.1 | ENST00000397717.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014055.hg.1 | ENST00000397717.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014055.hg.1 | ENST00000397717.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014055.hg.1 | ENST00000397717.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014055.hg.1 | ENST00000397717.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014055.hg.1 | ENST00000606474.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014055.hg.1 | ENST00000606474.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014055.hg.1 | ENST00000606474.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014055.hg.1 | ENST00000606474.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014055.hg.1 | NM_000904.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014055.hg.1 | NM_000904.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014055.hg.1 | NM_000904.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014055.hg.1 | NM_000904.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014055.hg.1 | NM_000904.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014055.hg.1 | NM_000904.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014055.hg.1 | NM_000904.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014055.hg.1 | NM_000904.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014055.hg.1 | NM_000904.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014055.hg.1 | NM_000904.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014055.hg.1 | NM_000904.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014055.hg.1 | NM_000904.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014055.hg.1 | NM_001290221.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014055.hg.1 | NM_001290221.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014055.hg.1 | NM_001290221.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014055.hg.1 | NM_001290221.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014055.hg.1 | NM_001290221.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014055.hg.1 | NM_001290221.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014055.hg.1 | NM_001290221.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014055.hg.1 | NM_001290221.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014055.hg.1 | NM_001290221.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014055.hg.1 | NM_001290221.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014055.hg.1 | NM_001290221.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014055.hg.1 | NM_001290221.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014055.hg.1 | NM_001290221.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014055.hg.1 | NM_001290221.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014055.hg.1 | NM_001290221.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014055.hg.1 | NM_001290222.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014055.hg.1 | NM_001290222.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014055.hg.1 | NM_001290222.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014055.hg.1 | NM_001290222.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014055.hg.1 | NM_001290222.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014055.hg.1 | NM_001290222.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014055.hg.1 | NM_001290222.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014055.hg.1 | NM_001290222.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014055.hg.1 | NM_001290222.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014055.hg.1 | NM_001290222.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014055.hg.1 | NM_001290222.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014055.hg.1 | ENST00000338130.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014055.hg.1 | ENST00000338130.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014055.hg.1 | ENST00000338130.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014055.hg.1 | ENST00000338130.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014055.hg.1 | ENST00000338130.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014055.hg.1 | ENST00000338130.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014055.hg.1 | ENST00000338130.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014055.hg.1 | ENST00000338130.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014055.hg.1 | ENST00000338130.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014055.hg.1 | ENST00000338130.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014055.hg.1 | ENST00000338130.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014055.hg.1 | ENST00000338130.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014055.hg.1 | ENST00000380430.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014055.hg.1 | ENST00000380430.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014055.hg.1 | ENST00000380430.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014055.hg.1 | ENST00000380430.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014055.hg.1 | ENST00000380430.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014055.hg.1 | ENST00000380430.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014055.hg.1 | ENST00000380430.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014055.hg.1 | ENST00000380430.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014055.hg.1 | ENST00000380430.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014055.hg.1 | ENST00000380441.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014055.hg.1 | ENST00000380441.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014055.hg.1 | ENST00000380441.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014055.hg.1 | ENST00000380441.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014055.hg.1 | ENST00000380441.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014055.hg.1 | ENST00000380441.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014055.hg.1 | ENST00000380441.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014055.hg.1 | ENST00000380441.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014055.hg.1 | ENST00000380441.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014055.hg.1 | ENST00000380454.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014055.hg.1 | ENST00000380454.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014055.hg.1 | ENST00000380454.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014055.hg.1 | ENST00000380454.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014055.hg.1 | ENST00000380454.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014055.hg.1 | ENST00000380454.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014055.hg.1 | ENST00000380454.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014055.hg.1 | ENST00000380454.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014055.hg.1 | ENST00000397717.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014055.hg.1 | ENST00000397717.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014055.hg.1 | ENST00000397717.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014055.hg.1 | ENST00000397717.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014055.hg.1 | ENST00000397717.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014055.hg.1 | ENST00000397717.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014055.hg.1 | ENST00000397717.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014055.hg.1 | ENST00000397717.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014055.hg.1 | ENST00000606474.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014055.hg.1 | ENST00000606474.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014055.hg.1 | ENST00000606474.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014055.hg.1 | ENST00000606474.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014055.hg.1 | NM_000904.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014055.hg.1 | NM_000904.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014055.hg.1 | NM_000904.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014055.hg.1 | NM_000904.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014055.hg.1 | NM_000904.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014055.hg.1 | NM_000904.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014055.hg.1 | NM_000904.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014055.hg.1 | NM_000904.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014055.hg.1 | NM_000904.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014055.hg.1 | NM_000904.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014055.hg.1 | NM_000904.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014055.hg.1 | NM_000904.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014055.hg.1 | NM_001290221.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014055.hg.1 | NM_001290221.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014055.hg.1 | NM_001290221.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014055.hg.1 | NM_001290221.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014055.hg.1 | NM_001290221.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014055.hg.1 | NM_001290221.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014055.hg.1 | NM_001290221.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014055.hg.1 | NM_001290221.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014055.hg.1 | NM_001290221.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014055.hg.1 | NM_001290221.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014055.hg.1 | NM_001290221.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014055.hg.1 | NM_001290221.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014055.hg.1 | NM_001290221.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014055.hg.1 | NM_001290221.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014055.hg.1 | NM_001290221.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014055.hg.1 | NM_001290222.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014055.hg.1 | NM_001290222.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014055.hg.1 | NM_001290222.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014055.hg.1 | NM_001290222.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014055.hg.1 | NM_001290222.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014055.hg.1 | NM_001290222.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014055.hg.1 | NM_001290222.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014055.hg.1 | NM_001290222.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014055.hg.1 | NM_001290222.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014055.hg.1 | NM_001290222.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014055.hg.1 | NM_001290222.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600197187.hg.1 | ENST00000338130.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600197187.hg.1 | ENST00000380430.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600197187.hg.1 | ENST00000380441.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600197187.hg.1 | ENST00000380454.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600197187.hg.1 | NM_000904.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600197187.hg.1 | NM_001290221.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600197187.hg.1 | NM_001290222.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600197187.hg.1 | ENST00000338130.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600197187.hg.1 | ENST00000380430.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600197187.hg.1 | ENST00000380441.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600197187.hg.1 | ENST00000380454.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600197187.hg.1 | NM_000904.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600197187.hg.1 | NM_001290221.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600197187.hg.1 | NM_001290222.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600197187.hg.1 | TC0600014055.hg.1 | Clariom D TC |

NQO2
Genomic Map