Assay Details
| Gene Symbol: WRNIP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_020135.2 | NP_064520.2 | 4 - 5 | 1680 | 66 | |
| NM_130395.2 | NP_569079.1 | 4 - 5 | 1605 | 66 | ||
| GenBank mRNA | AB056152.1 | - | 4 - 5 | 1681 | 66 | |
| AB209723.1 | - | 4 - 5 | 1433 | 66 | ||
| AF218313.1 | - | 4 - 5 | 1067 | 66 | ||
| AK026179.1 | - | 3 - 4 | 585 | 66 | ||
| AK094823.1 | - | 4 - 5 | 1640 | 66 | ||
| AK315047.1 | - | 4 - 5 | 1772 | 66 | ||
| AK315471.1 | - | 4 - 5 | 1574 | 66 | ||
| BC018923.2 | - | 4 - 5 | 1566 | 4109718 | 66 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC06000018.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR06000107.hg.1 | TC06000018.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR06000107.hg.1 | ENST00000380764.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06000107.hg.1 | ENST00000380769.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06000107.hg.1 | ENST00000380771.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06000107.hg.1 | ENST00000380773.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06000107.hg.1 | NM_020135.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06000107.hg.1 | NM_130395.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006611.hg.1 | ENST00000380764.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006611.hg.1 | ENST00000380764.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006611.hg.1 | ENST00000380764.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006611.hg.1 | ENST00000380764.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006611.hg.1 | ENST00000380764.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006611.hg.1 | ENST00000380764.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006611.hg.1 | ENST00000380764.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006611.hg.1 | ENST00000380764.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006611.hg.1 | ENST00000380769.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006611.hg.1 | ENST00000380769.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006611.hg.1 | ENST00000380769.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006611.hg.1 | ENST00000380769.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006611.hg.1 | ENST00000380769.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006611.hg.1 | ENST00000380769.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006611.hg.1 | ENST00000380769.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006611.hg.1 | ENST00000380769.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006611.hg.1 | ENST00000380769.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006611.hg.1 | ENST00000380769.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006611.hg.1 | ENST00000380769.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006611.hg.1 | ENST00000380771.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006611.hg.1 | ENST00000380771.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006611.hg.1 | ENST00000380771.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006611.hg.1 | ENST00000380771.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006611.hg.1 | ENST00000380771.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006611.hg.1 | ENST00000380771.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006611.hg.1 | ENST00000380771.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006611.hg.1 | ENST00000380771.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006611.hg.1 | ENST00000380771.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006611.hg.1 | ENST00000380771.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006611.hg.1 | ENST00000380771.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006611.hg.1 | ENST00000380771.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006611.hg.1 | ENST00000380773.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006611.hg.1 | ENST00000380773.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006611.hg.1 | ENST00000380773.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006611.hg.1 | ENST00000380773.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006611.hg.1 | ENST00000380773.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006611.hg.1 | ENST00000380773.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006611.hg.1 | ENST00000380773.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006611.hg.1 | ENST00000380773.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006611.hg.1 | ENST00000380773.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006611.hg.1 | ENST00000380773.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006611.hg.1 | ENST00000380773.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006611.hg.1 | ENST00000380773.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006611.hg.1 | ENST00000380773.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006611.hg.1 | ENST00000618555.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006611.hg.1 | ENST00000618555.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006611.hg.1 | ENST00000618555.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006611.hg.1 | ENST00000618555.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006611.hg.1 | ENST00000618555.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006611.hg.1 | ENST00000618555.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006611.hg.1 | ENST00000618555.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006611.hg.1 | ENST00000618555.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006611.hg.1 | ENST00000618555.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006611.hg.1 | ENST00000618555.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006611.hg.1 | ENST00000618555.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006611.hg.1 | ENST00000618555.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006611.hg.1 | ENST00000618555.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006611.hg.1 | NM_020135.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006611.hg.1 | NM_020135.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006611.hg.1 | NM_020135.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006611.hg.1 | NM_020135.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006611.hg.1 | NM_020135.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006611.hg.1 | NM_020135.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006611.hg.1 | NM_020135.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006611.hg.1 | NM_020135.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006611.hg.1 | NM_020135.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006611.hg.1 | NM_020135.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006611.hg.1 | NM_020135.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006611.hg.1 | NM_020135.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006611.hg.1 | NM_020135.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006611.hg.1 | NM_130395.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006611.hg.1 | NM_130395.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006611.hg.1 | NM_130395.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006611.hg.1 | NM_130395.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006611.hg.1 | NM_130395.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006611.hg.1 | NM_130395.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006611.hg.1 | NM_130395.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006611.hg.1 | NM_130395.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006611.hg.1 | NM_130395.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006611.hg.1 | NM_130395.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006611.hg.1 | NM_130395.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006611.hg.1 | NM_130395.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600006611.hg.1 | ENST00000380764.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600006611.hg.1 | ENST00000380764.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600006611.hg.1 | ENST00000380764.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600006611.hg.1 | ENST00000380764.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600006611.hg.1 | ENST00000380764.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600006611.hg.1 | ENST00000380764.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600006611.hg.1 | ENST00000380764.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600006611.hg.1 | ENST00000380764.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600006611.hg.1 | ENST00000380769.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600006611.hg.1 | ENST00000380769.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600006611.hg.1 | ENST00000380769.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600006611.hg.1 | ENST00000380769.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600006611.hg.1 | ENST00000380769.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600006611.hg.1 | ENST00000380769.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600006611.hg.1 | ENST00000380769.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600006611.hg.1 | ENST00000380769.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600006611.hg.1 | ENST00000380769.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600006611.hg.1 | ENST00000380769.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600006611.hg.1 | ENST00000380769.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600006611.hg.1 | ENST00000380771.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600006611.hg.1 | ENST00000380771.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600006611.hg.1 | ENST00000380771.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600006611.hg.1 | ENST00000380771.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600006611.hg.1 | ENST00000380771.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600006611.hg.1 | ENST00000380771.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600006611.hg.1 | ENST00000380771.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600006611.hg.1 | ENST00000380771.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600006611.hg.1 | ENST00000380771.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600006611.hg.1 | ENST00000380771.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600006611.hg.1 | ENST00000380771.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600006611.hg.1 | ENST00000380771.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600006611.hg.1 | ENST00000380773.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600006611.hg.1 | ENST00000380773.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600006611.hg.1 | ENST00000380773.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600006611.hg.1 | ENST00000380773.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600006611.hg.1 | ENST00000380773.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600006611.hg.1 | ENST00000380773.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600006611.hg.1 | ENST00000380773.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600006611.hg.1 | ENST00000380773.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600006611.hg.1 | ENST00000380773.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600006611.hg.1 | ENST00000380773.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600006611.hg.1 | ENST00000380773.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600006611.hg.1 | ENST00000380773.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600006611.hg.1 | ENST00000380773.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600006611.hg.1 | ENST00000618555.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600006611.hg.1 | ENST00000618555.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600006611.hg.1 | ENST00000618555.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600006611.hg.1 | ENST00000618555.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600006611.hg.1 | ENST00000618555.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600006611.hg.1 | ENST00000618555.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600006611.hg.1 | ENST00000618555.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600006611.hg.1 | ENST00000618555.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600006611.hg.1 | ENST00000618555.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600006611.hg.1 | ENST00000618555.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600006611.hg.1 | ENST00000618555.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600006611.hg.1 | ENST00000618555.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600006611.hg.1 | ENST00000618555.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600006611.hg.1 | NM_020135.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600006611.hg.1 | NM_020135.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600006611.hg.1 | NM_020135.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600006611.hg.1 | NM_020135.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600006611.hg.1 | NM_020135.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600006611.hg.1 | NM_020135.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600006611.hg.1 | NM_020135.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600006611.hg.1 | NM_020135.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600006611.hg.1 | NM_020135.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600006611.hg.1 | NM_020135.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600006611.hg.1 | NM_020135.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600006611.hg.1 | NM_020135.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600006611.hg.1 | NM_020135.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600006611.hg.1 | NM_130395.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600006611.hg.1 | NM_130395.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600006611.hg.1 | NM_130395.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600006611.hg.1 | NM_130395.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600006611.hg.1 | NM_130395.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600006611.hg.1 | NM_130395.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600006611.hg.1 | NM_130395.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600006611.hg.1 | NM_130395.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600006611.hg.1 | NM_130395.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600006611.hg.1 | NM_130395.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600006611.hg.1 | NM_130395.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600006611.hg.1 | NM_130395.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600145105.hg.1 | ENST00000380764.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600145105.hg.1 | ENST00000380769.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600145105.hg.1 | ENST00000380771.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600145105.hg.1 | ENST00000380773.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600145105.hg.1 | ENST00000618555.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600145105.hg.1 | NM_020135.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600145105.hg.1 | NM_130395.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600145105.hg.1 | ENST00000380764.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600145105.hg.1 | ENST00000380769.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600145105.hg.1 | ENST00000380771.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600145105.hg.1 | ENST00000380773.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600145105.hg.1 | ENST00000618555.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600145105.hg.1 | NM_020135.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600145105.hg.1 | NM_130395.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600145105.hg.1 | TC0600006611.hg.1 | Clariom D TC |

WRNIP1
Genomic Map