Assay Details
| Gene Symbol: CTR9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_014633.4 | NP_055448.1 | 19 - 20 | 2870 | 57 | |
| GenBank mRNA | AK291794.1 | - | 19 - 20 | 2606 | 57 | |
| BC058914.1 | - | 19 - 20 | 2617 | 6527835 | 57 | |
| D63875.1 | - | 19 - 20 | 2530 | 57 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC11000187.hg.1
Set Membership: |
|

CTR9
Genomic Map