Assay Details
| Gene Symbol: KIFC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145754.3 | NP_665697.1 | 10 - 11 | 1200 | 88 | |
| XM_005272357.3 | XP_005272414.1 | 10 - 11 | 1290 | 88 | ||
| XM_011517362.2 | XP_011515664.1 | 10 - 11 | 1291 | 88 | ||
| GenBank mRNA | AK055045.1 | - | 7 - 8 | 578 | 88 | |
| BC017311.2 | - | 11 - 12 | 1182 | 5090622 | 88 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC08000854.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0800009262.hg.1 | ENST00000301332.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009262.hg.1 | ENST00000301332.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009262.hg.1 | ENST00000301332.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009262.hg.1 | ENST00000301332.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009262.hg.1 | ENST00000301332.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009262.hg.1 | ENST00000301332.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009262.hg.1 | ENST00000301332.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009262.hg.1 | ENST00000301332.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009262.hg.1 | ENST00000301332.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009262.hg.1 | ENST00000301332.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009262.hg.1 | ENST00000301332.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009262.hg.1 | ENST00000301332.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009262.hg.1 | ENST00000301332.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009262.hg.1 | ENST00000301332.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009262.hg.1 | ENST00000301332.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009262.hg.1 | ENST00000301332.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009262.hg.1 | ENST00000301332.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009262.hg.1 | ENST00000301332.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009262.hg.1 | ENST00000301332.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009262.hg.1 | ENST00000301332.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009262.hg.1 | ENST00000301332.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009262.hg.1 | ENST00000301332.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009262.hg.1 | ENST00000301332.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009262.hg.1 | ENST00000301332.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009262.hg.1 | ENST00000301332.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009262.hg.1 | ENST00000529644.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009262.hg.1 | ENST00000529644.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009262.hg.1 | ENST00000529644.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009262.hg.1 | ENST00000529644.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009262.hg.1 | ENST00000529644.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009262.hg.1 | ENST00000529644.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009262.hg.1 | ENST00000529644.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009262.hg.1 | ENST00000529864.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009262.hg.1 | ENST00000529864.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009262.hg.1 | ENST00000529864.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009262.hg.1 | ENST00000529864.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009262.hg.1 | ENST00000531423.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009262.hg.1 | ENST00000531423.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009262.hg.1 | ENST00000531423.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009262.hg.1 | ENST00000531423.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009262.hg.1 | ENST00000531423.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009262.hg.1 | ENST00000531423.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009262.hg.1 | ENST00000531425.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009262.hg.1 | ENST00000531425.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009262.hg.1 | ENST00000531425.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009262.hg.1 | ENST00000531425.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009262.hg.1 | ENST00000531425.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009262.hg.1 | ENST00000531425.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009262.hg.1 | ENST00000531425.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009262.hg.1 | ENST00000533114.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009262.hg.1 | ENST00000533114.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009262.hg.1 | ENST00000533114.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009262.hg.1 | ENST00000533114.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009262.hg.1 | ENST00000533114.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009262.hg.1 | NM_145754.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009262.hg.1 | NM_145754.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009262.hg.1 | NM_145754.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009262.hg.1 | NM_145754.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009262.hg.1 | NM_145754.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009262.hg.1 | NM_145754.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009262.hg.1 | NM_145754.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009262.hg.1 | NM_145754.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009262.hg.1 | NM_145754.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009262.hg.1 | NM_145754.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009262.hg.1 | NM_145754.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009262.hg.1 | NM_145754.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009262.hg.1 | NM_145754.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009262.hg.1 | NM_145754.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009262.hg.1 | NM_145754.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009262.hg.1 | NM_145754.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009262.hg.1 | NM_145754.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009262.hg.1 | NM_145754.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009262.hg.1 | NM_145754.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009262.hg.1 | NM_145754.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009262.hg.1 | NM_145754.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009262.hg.1 | NM_145754.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009262.hg.1 | NM_145754.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR08011397.hg.1 | TC08000854.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR08011397.hg.1 | ENST00000301331.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08011397.hg.1 | ENST00000301332.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08011397.hg.1 | NM_145754.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR08011396.hg.1 | TC08000854.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR08011396.hg.1 | ENST00000301331.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08011396.hg.1 | ENST00000301332.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08011396.hg.1 | NM_145754.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009262.hg.1 | ENST00000301332.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009262.hg.1 | ENST00000301332.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009262.hg.1 | ENST00000301332.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009262.hg.1 | ENST00000301332.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009262.hg.1 | ENST00000301332.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009262.hg.1 | ENST00000301332.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009262.hg.1 | ENST00000301332.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009262.hg.1 | ENST00000301332.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009262.hg.1 | ENST00000301332.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009262.hg.1 | ENST00000301332.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009262.hg.1 | ENST00000301332.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009262.hg.1 | ENST00000301332.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009262.hg.1 | ENST00000301332.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009262.hg.1 | ENST00000301332.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009262.hg.1 | ENST00000301332.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009262.hg.1 | ENST00000301332.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009262.hg.1 | ENST00000301332.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009262.hg.1 | ENST00000301332.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009262.hg.1 | ENST00000301332.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009262.hg.1 | ENST00000301332.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009262.hg.1 | ENST00000301332.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009262.hg.1 | ENST00000301332.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009262.hg.1 | ENST00000301332.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009262.hg.1 | ENST00000301332.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009262.hg.1 | ENST00000301332.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009262.hg.1 | ENST00000529644.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009262.hg.1 | ENST00000529644.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009262.hg.1 | ENST00000529644.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009262.hg.1 | ENST00000529644.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009262.hg.1 | ENST00000529644.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009262.hg.1 | ENST00000529644.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009262.hg.1 | ENST00000529644.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009262.hg.1 | ENST00000529864.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009262.hg.1 | ENST00000529864.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009262.hg.1 | ENST00000529864.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009262.hg.1 | ENST00000529864.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009262.hg.1 | ENST00000531423.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009262.hg.1 | ENST00000531423.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009262.hg.1 | ENST00000531423.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009262.hg.1 | ENST00000531423.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009262.hg.1 | ENST00000531423.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009262.hg.1 | ENST00000531423.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009262.hg.1 | ENST00000531425.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009262.hg.1 | ENST00000531425.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009262.hg.1 | ENST00000531425.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009262.hg.1 | ENST00000531425.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009262.hg.1 | ENST00000531425.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009262.hg.1 | ENST00000531425.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009262.hg.1 | ENST00000531425.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009262.hg.1 | ENST00000533114.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009262.hg.1 | ENST00000533114.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009262.hg.1 | ENST00000533114.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009262.hg.1 | ENST00000533114.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009262.hg.1 | ENST00000533114.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009262.hg.1 | NM_145754.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009262.hg.1 | NM_145754.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009262.hg.1 | NM_145754.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009262.hg.1 | NM_145754.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009262.hg.1 | NM_145754.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009262.hg.1 | NM_145754.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009262.hg.1 | NM_145754.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009262.hg.1 | NM_145754.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009262.hg.1 | NM_145754.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009262.hg.1 | NM_145754.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009262.hg.1 | NM_145754.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009262.hg.1 | NM_145754.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009262.hg.1 | NM_145754.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009262.hg.1 | NM_145754.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009262.hg.1 | NM_145754.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009262.hg.1 | NM_145754.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009262.hg.1 | NM_145754.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009262.hg.1 | NM_145754.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009262.hg.1 | NM_145754.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009262.hg.1 | NM_145754.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009262.hg.1 | NM_145754.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009262.hg.1 | NM_145754.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009262.hg.1 | NM_145754.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800163474.hg.1 | ENST00000301332.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800163474.hg.1 | ENST00000529864.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800163474.hg.1 | NM_145754.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800163474.hg.1 | ENST00000301332.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800163474.hg.1 | ENST00000529864.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800163474.hg.1 | NM_145754.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800163474.hg.1 | TC0800009262.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0800163476.hg.1 | ENST00000301332.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800163476.hg.1 | NM_145754.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800163476.hg.1 | ENST00000301332.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800163476.hg.1 | NM_145754.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800163476.hg.1 | TC0800009262.hg.1 | Clariom D TC |

KIFC2
Genomic Map