Assay Details
| Gene Symbol: KIAA1614 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_020950.1 | NP_066001.1 | 5 - 6 | 2829 | 58 | |
| GenBank mRNA | AB046834.1 | - | 5 - 6 | 2817 | 58 | |
| AK090668.1 | - | 2 - 3 | 1984 | 58 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01001566.hg.1
Set Membership: |
|
Panther Classification:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0100010810.hg.1 | ENST00000367587.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010810.hg.1 | ENST00000367587.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010810.hg.1 | ENST00000367587.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010810.hg.1 | ENST00000367587.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010810.hg.1 | ENST00000367587.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010810.hg.1 | ENST00000367587.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010810.hg.1 | ENST00000367587.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010810.hg.1 | ENST00000367587.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010810.hg.1 | ENST00000367587.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010810.hg.1 | ENST00000367587.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010810.hg.1 | ENST00000367587.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010810.hg.1 | ENST00000367587.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010810.hg.1 | ENST00000367588.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010810.hg.1 | ENST00000367588.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010810.hg.1 | ENST00000367588.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010810.hg.1 | ENST00000367588.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010810.hg.1 | ENST00000367588.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010810.hg.1 | ENST00000367588.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010810.hg.1 | ENST00000367588.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010810.hg.1 | ENST00000367588.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010810.hg.1 | ENST00000367588.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010810.hg.1 | ENST00000367588.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010810.hg.1 | ENST00000367588.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010810.hg.1 | ENST00000367588.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010810.hg.1 | ENST00000367588.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010810.hg.1 | ENST00000367588.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010810.hg.1 | ENST00000367588.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010810.hg.1 | ENST00000367588.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010810.hg.1 | ENST00000367588.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010810.hg.1 | ENST00000367588.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010810.hg.1 | ENST00000367588.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010810.hg.1 | ENST00000461346.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010810.hg.1 | ENST00000461346.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010810.hg.1 | ENST00000461346.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010810.hg.1 | ENST00000461346.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010810.hg.1 | ENST00000461346.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010810.hg.1 | ENST00000483705.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010810.hg.1 | ENST00000483705.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010810.hg.1 | ENST00000483705.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010810.hg.1 | ENST00000483705.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010810.hg.1 | ENST00000483705.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010810.hg.1 | ENST00000496210.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010810.hg.1 | ENST00000496210.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010810.hg.1 | ENST00000496210.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010810.hg.1 | NM_020950.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010810.hg.1 | NM_020950.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010810.hg.1 | NM_020950.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010810.hg.1 | NM_020950.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010810.hg.1 | NM_020950.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010810.hg.1 | NM_020950.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010810.hg.1 | NM_020950.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010810.hg.1 | NM_020950.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010810.hg.1 | NM_020950.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010810.hg.1 | NM_020950.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010810.hg.1 | NM_020950.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010810.hg.1 | NM_020950.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010810.hg.1 | NM_020950.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010810.hg.1 | NM_020950.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010810.hg.1 | NM_020950.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01024200.hg.1 | TC01001566.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01024200.hg.1 | ENST00000367587.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01024200.hg.1 | ENST00000367588.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01024200.hg.1 | NM_020950.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100179451.hg.1 | ENST00000367587.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100179451.hg.1 | ENST00000367588.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100179451.hg.1 | ENST00000483705.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100179451.hg.1 | NM_020950.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100179451.hg.1 | ENST00000367587.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100179451.hg.1 | ENST00000367588.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100179451.hg.1 | ENST00000483705.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100179451.hg.1 | NM_020950.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100179451.hg.1 | TC0100010810.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0100010810.hg.1 | ENST00000367587.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010810.hg.1 | ENST00000367587.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010810.hg.1 | ENST00000367587.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010810.hg.1 | ENST00000367587.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010810.hg.1 | ENST00000367587.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010810.hg.1 | ENST00000367587.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010810.hg.1 | ENST00000367587.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010810.hg.1 | ENST00000367587.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010810.hg.1 | ENST00000367587.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010810.hg.1 | ENST00000367587.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010810.hg.1 | ENST00000367587.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010810.hg.1 | ENST00000367587.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010810.hg.1 | ENST00000367588.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010810.hg.1 | ENST00000367588.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010810.hg.1 | ENST00000367588.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010810.hg.1 | ENST00000367588.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010810.hg.1 | ENST00000367588.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010810.hg.1 | ENST00000367588.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010810.hg.1 | ENST00000367588.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010810.hg.1 | ENST00000367588.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010810.hg.1 | ENST00000367588.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010810.hg.1 | ENST00000367588.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010810.hg.1 | ENST00000367588.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010810.hg.1 | ENST00000367588.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010810.hg.1 | ENST00000367588.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010810.hg.1 | ENST00000367588.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010810.hg.1 | ENST00000367588.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010810.hg.1 | ENST00000367588.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010810.hg.1 | ENST00000367588.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010810.hg.1 | ENST00000367588.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010810.hg.1 | ENST00000367588.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010810.hg.1 | ENST00000461346.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010810.hg.1 | ENST00000461346.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010810.hg.1 | ENST00000461346.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010810.hg.1 | ENST00000461346.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010810.hg.1 | ENST00000461346.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010810.hg.1 | ENST00000483705.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010810.hg.1 | ENST00000483705.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010810.hg.1 | ENST00000483705.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010810.hg.1 | ENST00000483705.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010810.hg.1 | ENST00000483705.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010810.hg.1 | ENST00000496210.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010810.hg.1 | ENST00000496210.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010810.hg.1 | ENST00000496210.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010810.hg.1 | NM_020950.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010810.hg.1 | NM_020950.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010810.hg.1 | NM_020950.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010810.hg.1 | NM_020950.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010810.hg.1 | NM_020950.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010810.hg.1 | NM_020950.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010810.hg.1 | NM_020950.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010810.hg.1 | NM_020950.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010810.hg.1 | NM_020950.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010810.hg.1 | NM_020950.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010810.hg.1 | NM_020950.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010810.hg.1 | NM_020950.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010810.hg.1 | NM_020950.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010810.hg.1 | NM_020950.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010810.hg.1 | NM_020950.1 | Refseq |

KIAA1614
Genomic Map