Assay Details
| Gene Symbol: HLX | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021958.3 | NP_068777.1 | 1 - 2 | 1048 | 112 | |
| GenBank mRNA | AK056326.1 | - | 1 - 2 | 1045 | 112 | |
| AK127923.1 | - | 1 - 2 | 938 | 112 | ||
| AK313298.1 | - | 1 - 2 | 935 | 112 | ||
| BC007294.1 | - | 1 - 2 | 1033 | 3349581 | 112 | |
| BC033808.1 | - | 1 - 2 | 806 | 5184776 | 112 | |
| M60721.1 | - | 1 - 2 | 1004 | 112 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01001823.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
liver development
transcription, DNA-templated
regulation of transcription, DNA-templated
multicellular organism development
skeletal muscle tissue development
positive regulation of cell proliferation
cell differentiation
positive regulation of T-helper 1 cell differentiation
negative regulation of T-helper 2 cell differentiation
positive regulation of organ growth
enteric nervous system development
embryonic digestive tract morphogenesis
protein binding
sequence-specific DNA binding
liver development
transcription, DNA-templated
regulation of transcription, DNA-templated
multicellular organism development
skeletal muscle tissue development
positive regulation of cell proliferation
cell differentiation
positive regulation of T-helper 1 cell differentiation
negative regulation of T-helper 2 cell differentiation
positive regulation of organ growth
enteric nervous system development
embryonic digestive tract morphogenesis
protein binding
sequence-specific DNA binding
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR01028360.hg.1 | TC01001823.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01028360.hg.1 | ENST00000366903.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01028360.hg.1 | ENST00000549319.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01028360.hg.1 | NM_021958.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01028356.hg.1 | TC01001823.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01028356.hg.1 | ENST00000366903.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01028356.hg.1 | NM_021958.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01028357.hg.1 | TC01001823.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01028357.hg.1 | ENST00000366903.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01028357.hg.1 | NM_021958.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017398.hg.1 | ENST00000552026.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017398.hg.1 | ENST00000552026.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017398.hg.1 | ENST00000552026.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017398.hg.1 | NR_046901.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017398.hg.1 | NR_046901.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017398.hg.1 | NR_046901.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01028359.hg.1 | TC01001823.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01028359.hg.1 | ENST00000366903.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01028359.hg.1 | NM_021958.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011669.hg.1 | ENST00000366903.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011669.hg.1 | ENST00000366903.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011669.hg.1 | ENST00000366903.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011669.hg.1 | ENST00000366903.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011669.hg.1 | ENST00000366903.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011669.hg.1 | ENST00000366903.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011669.hg.1 | ENST00000366903.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011669.hg.1 | ENST00000366903.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011669.hg.1 | ENST00000427693.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011669.hg.1 | ENST00000427693.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011669.hg.1 | ENST00000427693.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011669.hg.1 | ENST00000427693.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011669.hg.1 | ENST00000549319.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011669.hg.1 | ENST00000549319.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011669.hg.1 | ENST00000549319.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011669.hg.1 | ENST00000549319.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011669.hg.1 | ENST00000549319.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011669.hg.1 | ENST00000549319.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011669.hg.1 | ENST00000549319.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011669.hg.1 | ENST00000549319.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011669.hg.1 | ENST00000549319.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011669.hg.1 | ENST00000549319.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011669.hg.1 | ENST00000549319.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011669.hg.1 | ENST00000549319.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011669.hg.1 | ENST00000549319.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011669.hg.1 | ENST00000549319.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011669.hg.1 | ENST00000549319.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011669.hg.1 | ENST00000549319.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011669.hg.1 | ENST00000549319.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011669.hg.1 | NM_021958.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011669.hg.1 | NM_021958.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011669.hg.1 | NM_021958.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011669.hg.1 | NM_021958.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011669.hg.1 | NM_021958.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011669.hg.1 | NM_021958.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011669.hg.1 | NM_021958.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011669.hg.1 | NM_021958.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100185916.hg.1 | ENST00000366903.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100185916.hg.1 | ENST00000549319.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100185916.hg.1 | NM_021958.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100185916.hg.1 | ENST00000366903.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100185916.hg.1 | ENST00000549319.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100185916.hg.1 | NM_021958.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100185916.hg.1 | TC0100011669.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0100017398.hg.1 | ENST00000552026.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017398.hg.1 | ENST00000552026.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017398.hg.1 | ENST00000552026.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017398.hg.1 | NR_046901.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017398.hg.1 | NR_046901.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017398.hg.1 | NR_046901.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100185915.hg.1 | ENST00000366903.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100185915.hg.1 | ENST00000549319.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100185915.hg.1 | NM_021958.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100185915.hg.1 | ENST00000366903.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100185915.hg.1 | ENST00000549319.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100185915.hg.1 | NM_021958.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100185915.hg.1 | TC0100011669.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100185921.hg.1 | ENST00000366903.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100185921.hg.1 | ENST00000427693.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100185921.hg.1 | ENST00000549319.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100185921.hg.1 | NM_021958.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100185921.hg.1 | ENST00000366903.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100185921.hg.1 | ENST00000427693.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100185921.hg.1 | ENST00000549319.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100185921.hg.1 | NM_021958.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100185921.hg.1 | TC0100011669.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0100011669.hg.1 | ENST00000366903.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011669.hg.1 | ENST00000366903.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011669.hg.1 | ENST00000366903.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011669.hg.1 | ENST00000366903.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011669.hg.1 | ENST00000366903.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011669.hg.1 | ENST00000366903.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011669.hg.1 | ENST00000366903.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011669.hg.1 | ENST00000366903.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011669.hg.1 | ENST00000427693.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011669.hg.1 | ENST00000427693.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011669.hg.1 | ENST00000427693.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011669.hg.1 | ENST00000427693.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011669.hg.1 | ENST00000549319.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011669.hg.1 | ENST00000549319.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011669.hg.1 | ENST00000549319.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011669.hg.1 | ENST00000549319.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011669.hg.1 | ENST00000549319.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011669.hg.1 | ENST00000549319.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011669.hg.1 | ENST00000549319.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011669.hg.1 | ENST00000549319.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011669.hg.1 | ENST00000549319.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011669.hg.1 | ENST00000549319.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011669.hg.1 | ENST00000549319.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011669.hg.1 | ENST00000549319.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011669.hg.1 | ENST00000549319.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011669.hg.1 | ENST00000549319.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011669.hg.1 | ENST00000549319.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011669.hg.1 | ENST00000549319.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011669.hg.1 | ENST00000549319.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011669.hg.1 | NM_021958.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011669.hg.1 | NM_021958.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011669.hg.1 | NM_021958.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011669.hg.1 | NM_021958.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011669.hg.1 | NM_021958.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011669.hg.1 | NM_021958.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011669.hg.1 | NM_021958.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011669.hg.1 | NM_021958.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100185917.hg.1 | ENST00000366903.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100185917.hg.1 | ENST00000549319.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100185917.hg.1 | NM_021958.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100185917.hg.1 | ENST00000366903.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100185917.hg.1 | ENST00000549319.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100185917.hg.1 | NM_021958.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100185917.hg.1 | TC0100011669.hg.1 | Clariom D TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 197242 | - | - |

HLX
Genomic Map