| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009004.hg.1 |
ENST00000369221.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009004.hg.1 |
ENST00000369221.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009004.hg.1 |
ENST00000369221.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009004.hg.1 |
ENST00000369221.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009004.hg.1 |
ENST00000369221.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009004.hg.1 |
ENST00000369221.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009004.hg.1 |
ENST00000369221.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009004.hg.1 |
ENST00000369221.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009004.hg.1 |
ENST00000369221.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009004.hg.1 |
ENST00000369221.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009004.hg.1 |
ENST00000369221.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009004.hg.1 |
ENST00000369221.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009004.hg.1 |
ENST00000369221.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009004.hg.1 |
ENST00000369221.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009004.hg.1 |
ENST00000470562.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009004.hg.1 |
ENST00000470562.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009004.hg.1 |
ENST00000470562.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009004.hg.1 |
ENST00000470562.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009004.hg.1 |
ENST00000470562.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009004.hg.1 |
NM_000936.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009004.hg.1 |
NM_000936.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009004.hg.1 |
NM_000936.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009004.hg.1 |
NM_000936.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009004.hg.1 |
NM_000936.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009004.hg.1 |
NM_000936.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009004.hg.1 |
NM_000936.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009004.hg.1 |
NM_000936.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009004.hg.1 |
NM_000936.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009004.hg.1 |
NM_000936.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009004.hg.1 |
NM_000936.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009004.hg.1 |
NM_000936.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009004.hg.1 |
NM_000936.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009004.hg.1 |
NM_000936.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000161636.hg.1 |
ENST00000369221.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000161636.hg.1 |
NM_000936.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000161636.hg.1 |
ENST00000369221.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000161636.hg.1 |
NM_000936.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000161636.hg.1 |
TC1000009004.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC1000009004.hg.1 |
ENST00000369221.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000009004.hg.1 |
ENST00000369221.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000009004.hg.1 |
ENST00000369221.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000009004.hg.1 |
ENST00000369221.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000009004.hg.1 |
ENST00000369221.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000009004.hg.1 |
ENST00000369221.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000009004.hg.1 |
ENST00000369221.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000009004.hg.1 |
ENST00000369221.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000009004.hg.1 |
ENST00000369221.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000009004.hg.1 |
ENST00000369221.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000009004.hg.1 |
ENST00000369221.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000009004.hg.1 |
ENST00000369221.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000009004.hg.1 |
ENST00000369221.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000009004.hg.1 |
ENST00000369221.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000009004.hg.1 |
ENST00000470562.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000009004.hg.1 |
ENST00000470562.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000009004.hg.1 |
ENST00000470562.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000009004.hg.1 |
ENST00000470562.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000009004.hg.1 |
ENST00000470562.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000009004.hg.1 |
NM_000936.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000009004.hg.1 |
NM_000936.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000009004.hg.1 |
NM_000936.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000009004.hg.1 |
NM_000936.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000009004.hg.1 |
NM_000936.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000009004.hg.1 |
NM_000936.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000009004.hg.1 |
NM_000936.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000009004.hg.1 |
NM_000936.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000009004.hg.1 |
NM_000936.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000009004.hg.1 |
NM_000936.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000009004.hg.1 |
NM_000936.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000009004.hg.1 |
NM_000936.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000009004.hg.1 |
NM_000936.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000009004.hg.1 |
NM_000936.3 |
Refseq |