Assay Details
| Gene Symbol: C16orf72 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_014117.2 | NP_054836.2 | 1 - 2 | 596 | 109 | |
| XM_011522462.2 | XP_011520764.1 | 1 - 2 | 947 | 109 | ||
| GenBank mRNA | BC113570.1 | - | 1 - 2 | 496 | 8322622 | 109 |
| CB959956.1 | - | 1 - 2 | 416 | 109 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC16000150.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR16002336.hg.1 | ENST00000327827.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16002336.hg.1 | NM_014117.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16002336.hg.1 | TC16000150.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR16002337.hg.1 | ENST00000327827.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16002337.hg.1 | NM_014117.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16002337.hg.1 | TC16000150.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR16002335.hg.1 | ENST00000327827.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16002335.hg.1 | NM_014117.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16002335.hg.1 | TC16000150.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006842.hg.1 | ENST00000327827.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006842.hg.1 | ENST00000327827.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006842.hg.1 | ENST00000327827.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006842.hg.1 | ENST00000327827.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006842.hg.1 | ENST00000327827.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006842.hg.1 | ENST00000327827.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006842.hg.1 | ENST00000327827.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006842.hg.1 | ENST00000327827.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006842.hg.1 | ENST00000327827.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006842.hg.1 | ENST00000327827.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006842.hg.1 | ENST00000327827.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006842.hg.1 | ENST00000574285.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006842.hg.1 | ENST00000574285.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006842.hg.1 | ENST00000574285.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006842.hg.1 | ENST00000574285.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006842.hg.1 | ENST00000574285.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006842.hg.1 | ENST00000574285.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006842.hg.1 | ENST00000574285.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006842.hg.1 | ENST00000574285.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006842.hg.1 | ENST00000574285.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006842.hg.1 | ENST00000574285.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006842.hg.1 | ENST00000574285.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006842.hg.1 | ENST00000574285.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006842.hg.1 | ENST00000574285.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006842.hg.1 | ENST00000574285.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006842.hg.1 | ENST00000574616.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006842.hg.1 | ENST00000574616.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006842.hg.1 | ENST00000574616.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006842.hg.1 | ENST00000574616.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006842.hg.1 | ENST00000574616.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006842.hg.1 | ENST00000574616.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006842.hg.1 | ENST00000574616.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006842.hg.1 | ENST00000574616.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006842.hg.1 | ENST00000574616.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006842.hg.1 | ENST00000574616.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006842.hg.1 | NM_014117.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006842.hg.1 | NM_014117.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006842.hg.1 | NM_014117.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006842.hg.1 | NM_014117.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006842.hg.1 | NM_014117.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006842.hg.1 | NM_014117.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006842.hg.1 | NM_014117.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006842.hg.1 | NM_014117.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006842.hg.1 | NM_014117.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600148762.hg.1 | ENST00000327827.11 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600148762.hg.1 | ENST00000574285.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600148762.hg.1 | NM_014117.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600148762.hg.1 | ENST00000327827.11 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600148762.hg.1 | ENST00000574285.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600148762.hg.1 | NM_014117.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600148762.hg.1 | TC1600006842.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1600006842.hg.1 | ENST00000327827.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006842.hg.1 | ENST00000327827.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006842.hg.1 | ENST00000327827.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006842.hg.1 | ENST00000327827.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006842.hg.1 | ENST00000327827.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006842.hg.1 | ENST00000327827.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006842.hg.1 | ENST00000327827.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006842.hg.1 | ENST00000327827.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006842.hg.1 | ENST00000327827.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006842.hg.1 | ENST00000327827.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006842.hg.1 | ENST00000327827.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006842.hg.1 | ENST00000574285.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006842.hg.1 | ENST00000574285.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006842.hg.1 | ENST00000574285.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006842.hg.1 | ENST00000574285.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006842.hg.1 | ENST00000574285.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006842.hg.1 | ENST00000574285.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006842.hg.1 | ENST00000574285.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006842.hg.1 | ENST00000574285.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006842.hg.1 | ENST00000574285.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006842.hg.1 | ENST00000574285.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006842.hg.1 | ENST00000574285.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006842.hg.1 | ENST00000574285.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006842.hg.1 | ENST00000574285.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006842.hg.1 | ENST00000574285.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006842.hg.1 | ENST00000574616.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006842.hg.1 | ENST00000574616.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006842.hg.1 | ENST00000574616.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006842.hg.1 | ENST00000574616.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006842.hg.1 | ENST00000574616.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006842.hg.1 | ENST00000574616.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006842.hg.1 | ENST00000574616.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006842.hg.1 | ENST00000574616.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006842.hg.1 | ENST00000574616.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006842.hg.1 | ENST00000574616.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006842.hg.1 | NM_014117.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600006842.hg.1 | NM_014117.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600006842.hg.1 | NM_014117.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600006842.hg.1 | NM_014117.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600006842.hg.1 | NM_014117.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600006842.hg.1 | NM_014117.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600006842.hg.1 | NM_014117.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600006842.hg.1 | NM_014117.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600006842.hg.1 | NM_014117.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600148758.hg.1 | ENST00000327827.11 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600148758.hg.1 | NM_014117.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600148758.hg.1 | ENST00000327827.11 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600148758.hg.1 | NM_014117.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600148758.hg.1 | TC1600006842.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1600148759.hg.1 | ENST00000327827.11 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600148759.hg.1 | NM_014117.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600148759.hg.1 | ENST00000327827.11 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600148759.hg.1 | NM_014117.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600148759.hg.1 | TC1600006842.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1600148761.hg.1 | ENST00000327827.11 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600148761.hg.1 | ENST00000574285.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600148761.hg.1 | NM_014117.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600148761.hg.1 | ENST00000327827.11 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600148761.hg.1 | ENST00000574285.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600148761.hg.1 | NM_014117.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600148761.hg.1 | TC1600006842.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1600148760.hg.1 | ENST00000327827.11 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600148760.hg.1 | NM_014117.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600148760.hg.1 | ENST00000327827.11 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600148760.hg.1 | NM_014117.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600148760.hg.1 | TC1600006842.hg.1 | Clariom D TC |

C16orf72
Genomic Map