Assay Details
| Gene Symbol: MAGEA11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001011544.1 | NP_001011544.1 | 2 - 3 | 197 | 70 | |
| GenBank mRNA | BX337921.2 | - | 2 - 3 | 206 | 70 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC0X000713.hg.1
Additional Information:
This assay is in the 5'UTR region of the NM_001011544.1 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008689.hg.1 | ENST00000333104.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008689.hg.1 | ENST00000333104.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008689.hg.1 | ENST00000333104.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008689.hg.1 | ENST00000333104.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008689.hg.1 | ENST00000333104.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008689.hg.1 | ENST00000333104.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008689.hg.1 | ENST00000333104.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008689.hg.1 | ENST00000355220.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008689.hg.1 | ENST00000355220.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008689.hg.1 | ENST00000355220.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008689.hg.1 | ENST00000355220.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008689.hg.1 | ENST00000355220.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008689.hg.1 | ENST00000355220.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008689.hg.1 | ENST00000355220.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008689.hg.1 | ENST00000412632.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008689.hg.1 | ENST00000412632.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008689.hg.1 | ENST00000412632.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008689.hg.1 | ENST00000412632.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008689.hg.1 | ENST00000412632.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008689.hg.1 | ENST00000412632.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008689.hg.1 | ENST00000518694.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008689.hg.1 | ENST00000518694.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008689.hg.1 | ENST00000518694.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008689.hg.1 | ENST00000518694.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008689.hg.1 | ENST00000518694.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008689.hg.1 | ENST00000518694.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008689.hg.1 | ENST00000518694.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008689.hg.1 | ENST00000518694.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008689.hg.1 | ENST00000518694.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008689.hg.1 | ENST00000518694.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008689.hg.1 | ENST00000518694.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008689.hg.1 | ENST00000518694.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008689.hg.1 | NM_001011544.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008689.hg.1 | NM_001011544.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008689.hg.1 | NM_001011544.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008689.hg.1 | NM_001011544.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008689.hg.1 | NM_001011544.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008689.hg.1 | NM_001011544.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008689.hg.1 | NM_001011544.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008689.hg.1 | NM_001011544.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008689.hg.1 | NM_005366.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008689.hg.1 | NM_005366.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008689.hg.1 | NM_005366.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008689.hg.1 | NM_005366.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008689.hg.1 | NM_005366.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008689.hg.1 | NM_005366.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008689.hg.1 | NM_005366.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X009154.hg.1 | TC0X000713.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X009154.hg.1 | ENST00000333104.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X009154.hg.1 | NM_001011544.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X009152.hg.1 | TC0X000713.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X009152.hg.1 | NM_001011544.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X009155.hg.1 | TC0X000713.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X009155.hg.1 | ENST00000333104.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X009155.hg.1 | NM_001011544.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00158894.hg.1 | ENST00000333104.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00158894.hg.1 | ENST00000412632.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00158894.hg.1 | NM_001011544.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00158894.hg.1 | ENST00000333104.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00158894.hg.1 | ENST00000412632.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00158894.hg.1 | NM_001011544.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00158894.hg.1 | TC0X00008689.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0X00158895.hg.1 | ENST00000333104.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00158895.hg.1 | ENST00000412632.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00158895.hg.1 | ENST00000518694.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00158895.hg.1 | NM_001011544.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00158895.hg.1 | ENST00000333104.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00158895.hg.1 | ENST00000412632.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00158895.hg.1 | ENST00000518694.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00158895.hg.1 | NM_001011544.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00158895.hg.1 | TC0X00008689.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0X00008689.hg.1 | ENST00000333104.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008689.hg.1 | ENST00000333104.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008689.hg.1 | ENST00000333104.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008689.hg.1 | ENST00000333104.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008689.hg.1 | ENST00000333104.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008689.hg.1 | ENST00000333104.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008689.hg.1 | ENST00000333104.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008689.hg.1 | ENST00000355220.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008689.hg.1 | ENST00000355220.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008689.hg.1 | ENST00000355220.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008689.hg.1 | ENST00000355220.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008689.hg.1 | ENST00000355220.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008689.hg.1 | ENST00000355220.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008689.hg.1 | ENST00000355220.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008689.hg.1 | ENST00000412632.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008689.hg.1 | ENST00000412632.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008689.hg.1 | ENST00000412632.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008689.hg.1 | ENST00000412632.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008689.hg.1 | ENST00000412632.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008689.hg.1 | ENST00000412632.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008689.hg.1 | ENST00000518694.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008689.hg.1 | ENST00000518694.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008689.hg.1 | ENST00000518694.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008689.hg.1 | ENST00000518694.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008689.hg.1 | ENST00000518694.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008689.hg.1 | ENST00000518694.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008689.hg.1 | ENST00000518694.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008689.hg.1 | ENST00000518694.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008689.hg.1 | ENST00000518694.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008689.hg.1 | ENST00000518694.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008689.hg.1 | ENST00000518694.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008689.hg.1 | ENST00000518694.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008689.hg.1 | NM_001011544.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008689.hg.1 | NM_001011544.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008689.hg.1 | NM_001011544.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008689.hg.1 | NM_001011544.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008689.hg.1 | NM_001011544.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008689.hg.1 | NM_001011544.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008689.hg.1 | NM_001011544.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008689.hg.1 | NM_001011544.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008689.hg.1 | NM_005366.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008689.hg.1 | NM_005366.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008689.hg.1 | NM_005366.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008689.hg.1 | NM_005366.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008689.hg.1 | NM_005366.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008689.hg.1 | NM_005366.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008689.hg.1 | NM_005366.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00158892.hg.1 | ENST00000412632.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00158892.hg.1 | ENST00000518694.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00158892.hg.1 | NM_001011544.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00158892.hg.1 | ENST00000412632.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00158892.hg.1 | ENST00000518694.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00158892.hg.1 | NM_001011544.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00158892.hg.1 | TC0X00008689.hg.1 | Clariom D TC |

MAGEA11
Genomic Map