| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008689.hg.1 |
ENST00000333104.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008689.hg.1 |
ENST00000333104.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008689.hg.1 |
ENST00000333104.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008689.hg.1 |
ENST00000333104.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008689.hg.1 |
ENST00000333104.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008689.hg.1 |
ENST00000333104.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008689.hg.1 |
ENST00000333104.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008689.hg.1 |
ENST00000355220.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008689.hg.1 |
ENST00000355220.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008689.hg.1 |
ENST00000355220.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008689.hg.1 |
ENST00000355220.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008689.hg.1 |
ENST00000355220.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008689.hg.1 |
ENST00000355220.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008689.hg.1 |
ENST00000355220.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008689.hg.1 |
ENST00000412632.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008689.hg.1 |
ENST00000412632.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008689.hg.1 |
ENST00000412632.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008689.hg.1 |
ENST00000412632.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008689.hg.1 |
ENST00000412632.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008689.hg.1 |
ENST00000412632.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008689.hg.1 |
ENST00000518694.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008689.hg.1 |
ENST00000518694.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008689.hg.1 |
ENST00000518694.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008689.hg.1 |
ENST00000518694.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008689.hg.1 |
ENST00000518694.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008689.hg.1 |
ENST00000518694.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008689.hg.1 |
ENST00000518694.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008689.hg.1 |
ENST00000518694.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008689.hg.1 |
ENST00000518694.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008689.hg.1 |
ENST00000518694.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008689.hg.1 |
ENST00000518694.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008689.hg.1 |
ENST00000518694.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008689.hg.1 |
NM_001011544.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008689.hg.1 |
NM_001011544.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008689.hg.1 |
NM_001011544.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008689.hg.1 |
NM_001011544.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008689.hg.1 |
NM_001011544.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008689.hg.1 |
NM_001011544.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008689.hg.1 |
NM_001011544.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008689.hg.1 |
NM_001011544.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008689.hg.1 |
NM_005366.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008689.hg.1 |
NM_005366.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008689.hg.1 |
NM_005366.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008689.hg.1 |
NM_005366.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008689.hg.1 |
NM_005366.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008689.hg.1 |
NM_005366.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008689.hg.1 |
NM_005366.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X009163.hg.1 |
TC0X000713.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X009163.hg.1 |
ENST00000333104.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X009163.hg.1 |
ENST00000355220.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X009163.hg.1 |
NM_001011544.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X009163.hg.1 |
NM_005366.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00158904.hg.1 |
ENST00000333104.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00158904.hg.1 |
ENST00000355220.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00158904.hg.1 |
ENST00000412632.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00158904.hg.1 |
ENST00000518694.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00158904.hg.1 |
NM_001011544.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00158904.hg.1 |
NM_005366.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00158904.hg.1 |
ENST00000333104.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00158904.hg.1 |
ENST00000355220.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00158904.hg.1 |
ENST00000412632.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00158904.hg.1 |
ENST00000518694.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00158904.hg.1 |
NM_001011544.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00158904.hg.1 |
NM_005366.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00158904.hg.1 |
TC0X00008689.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0X00008689.hg.1 |
ENST00000333104.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008689.hg.1 |
ENST00000333104.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008689.hg.1 |
ENST00000333104.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008689.hg.1 |
ENST00000333104.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008689.hg.1 |
ENST00000333104.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008689.hg.1 |
ENST00000333104.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008689.hg.1 |
ENST00000333104.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008689.hg.1 |
ENST00000355220.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008689.hg.1 |
ENST00000355220.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008689.hg.1 |
ENST00000355220.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008689.hg.1 |
ENST00000355220.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008689.hg.1 |
ENST00000355220.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008689.hg.1 |
ENST00000355220.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008689.hg.1 |
ENST00000355220.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008689.hg.1 |
ENST00000412632.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008689.hg.1 |
ENST00000412632.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008689.hg.1 |
ENST00000412632.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008689.hg.1 |
ENST00000412632.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008689.hg.1 |
ENST00000412632.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008689.hg.1 |
ENST00000412632.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008689.hg.1 |
ENST00000518694.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008689.hg.1 |
ENST00000518694.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008689.hg.1 |
ENST00000518694.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008689.hg.1 |
ENST00000518694.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008689.hg.1 |
ENST00000518694.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008689.hg.1 |
ENST00000518694.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008689.hg.1 |
ENST00000518694.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008689.hg.1 |
ENST00000518694.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008689.hg.1 |
ENST00000518694.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008689.hg.1 |
ENST00000518694.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008689.hg.1 |
ENST00000518694.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008689.hg.1 |
ENST00000518694.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008689.hg.1 |
NM_001011544.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00008689.hg.1 |
NM_001011544.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00008689.hg.1 |
NM_001011544.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00008689.hg.1 |
NM_001011544.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00008689.hg.1 |
NM_001011544.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00008689.hg.1 |
NM_001011544.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00008689.hg.1 |
NM_001011544.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00008689.hg.1 |
NM_001011544.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00008689.hg.1 |
NM_005366.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00008689.hg.1 |
NM_005366.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00008689.hg.1 |
NM_005366.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00008689.hg.1 |
NM_005366.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00008689.hg.1 |
NM_005366.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00008689.hg.1 |
NM_005366.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00008689.hg.1 |
NM_005366.4 |
Refseq |