Assay Details
| Gene Symbol: ACMSD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_138326.2 | NP_612199.2 | 4 - 5 | 320 | 74 | |
| XM_005263589.4 | XP_005263646.1 | 5 - 6 | 728 | 74 | ||
| GenBank mRNA | AB071418.1 | - | 4 - 5 | 253 | 74 | |
| AK125008.1 | - | 4 - 5 | 320 | 74 | ||
| BC107420.1 | - | 4 - 5 | 260 | 4594027 | 74 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC02000877.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
tryptophan catabolic process
quinolinate metabolic process
protein oligomerization
regulation of quinolinate biosynthetic process
picolinic acid biosynthetic process
regulation of 'de novo' NAD biosynthetic process from tryptophan
aminocarboxymuconate-semialdehyde decarboxylase activity
protein binding
zinc ion binding
hydrolase activity
tryptophan catabolic process
quinolinate metabolic process
protein oligomerization
regulation of quinolinate biosynthetic process
picolinic acid biosynthetic process
regulation of 'de novo' NAD biosynthetic process from tryptophan
aminocarboxymuconate-semialdehyde decarboxylase activity
protein binding
zinc ion binding
hydrolase activity
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR02012942.hg.1 | TC02000877.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02012942.hg.1 | ENST00000283054.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02012942.hg.1 | ENST00000356140.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02012942.hg.1 | ENST00000392928.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02012942.hg.1 | NM_138326.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009424.hg.1 | ENST00000356140.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009424.hg.1 | ENST00000356140.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009424.hg.1 | ENST00000356140.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009424.hg.1 | ENST00000356140.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009424.hg.1 | ENST00000356140.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009424.hg.1 | ENST00000356140.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009424.hg.1 | ENST00000356140.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009424.hg.1 | ENST00000356140.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009424.hg.1 | ENST00000356140.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009424.hg.1 | ENST00000356140.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009424.hg.1 | ENST00000356140.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009424.hg.1 | ENST00000356140.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009424.hg.1 | ENST00000356140.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009424.hg.1 | ENST00000392928.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009424.hg.1 | ENST00000392928.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009424.hg.1 | ENST00000392928.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009424.hg.1 | ENST00000392928.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009424.hg.1 | ENST00000392928.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009424.hg.1 | ENST00000392928.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009424.hg.1 | ENST00000392928.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009424.hg.1 | ENST00000392928.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009424.hg.1 | ENST00000392928.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009424.hg.1 | ENST00000392928.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009424.hg.1 | ENST00000392928.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009424.hg.1 | ENST00000392928.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009424.hg.1 | ENST00000392928.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009424.hg.1 | ENST00000485893.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009424.hg.1 | ENST00000485893.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009424.hg.1 | ENST00000485893.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009424.hg.1 | ENST00000485893.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009424.hg.1 | ENST00000485893.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009424.hg.1 | ENST00000485893.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009424.hg.1 | ENST00000485893.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009424.hg.1 | ENST00000485893.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009424.hg.1 | ENST00000485893.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009424.hg.1 | ENST00000485893.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009424.hg.1 | ENST00000498093.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009424.hg.1 | ENST00000498093.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009424.hg.1 | ENST00000498093.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009424.hg.1 | ENST00000498093.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009424.hg.1 | NM_001307983.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009424.hg.1 | NM_001307983.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009424.hg.1 | NM_001307983.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009424.hg.1 | NM_001307983.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009424.hg.1 | NM_001307983.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009424.hg.1 | NM_001307983.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009424.hg.1 | NM_001307983.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009424.hg.1 | NM_001307983.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009424.hg.1 | NM_001307983.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009424.hg.1 | NM_001307983.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009424.hg.1 | NM_001307983.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009424.hg.1 | NM_001307983.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009424.hg.1 | NM_001307983.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009424.hg.1 | NM_138326.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009424.hg.1 | NM_138326.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009424.hg.1 | NM_138326.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009424.hg.1 | NM_138326.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009424.hg.1 | NM_138326.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009424.hg.1 | NM_138326.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009424.hg.1 | NM_138326.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009424.hg.1 | NM_138326.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009424.hg.1 | NM_138326.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009424.hg.1 | NM_138326.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009424.hg.1 | NM_138326.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009424.hg.1 | NM_138326.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200165888.hg.1 | ENST00000356140.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200165888.hg.1 | ENST00000392928.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200165888.hg.1 | ENST00000485893.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200165888.hg.1 | NM_001307983.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200165888.hg.1 | NM_138326.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200165888.hg.1 | ENST00000356140.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200165888.hg.1 | ENST00000392928.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200165888.hg.1 | ENST00000485893.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200165888.hg.1 | NM_001307983.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200165888.hg.1 | NM_138326.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200165888.hg.1 | TC0200009424.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0200009424.hg.1 | ENST00000356140.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009424.hg.1 | ENST00000356140.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009424.hg.1 | ENST00000356140.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009424.hg.1 | ENST00000356140.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009424.hg.1 | ENST00000356140.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009424.hg.1 | ENST00000356140.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009424.hg.1 | ENST00000356140.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009424.hg.1 | ENST00000356140.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009424.hg.1 | ENST00000356140.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009424.hg.1 | ENST00000356140.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009424.hg.1 | ENST00000356140.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009424.hg.1 | ENST00000356140.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009424.hg.1 | ENST00000356140.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009424.hg.1 | ENST00000392928.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009424.hg.1 | ENST00000392928.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009424.hg.1 | ENST00000392928.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009424.hg.1 | ENST00000392928.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009424.hg.1 | ENST00000392928.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009424.hg.1 | ENST00000392928.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009424.hg.1 | ENST00000392928.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009424.hg.1 | ENST00000392928.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009424.hg.1 | ENST00000392928.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009424.hg.1 | ENST00000392928.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009424.hg.1 | ENST00000392928.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009424.hg.1 | ENST00000392928.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009424.hg.1 | ENST00000392928.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009424.hg.1 | ENST00000485893.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009424.hg.1 | ENST00000485893.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009424.hg.1 | ENST00000485893.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009424.hg.1 | ENST00000485893.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009424.hg.1 | ENST00000485893.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009424.hg.1 | ENST00000485893.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009424.hg.1 | ENST00000485893.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009424.hg.1 | ENST00000485893.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009424.hg.1 | ENST00000485893.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009424.hg.1 | ENST00000485893.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009424.hg.1 | ENST00000498093.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009424.hg.1 | ENST00000498093.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009424.hg.1 | ENST00000498093.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009424.hg.1 | ENST00000498093.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009424.hg.1 | NM_001307983.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009424.hg.1 | NM_001307983.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009424.hg.1 | NM_001307983.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009424.hg.1 | NM_001307983.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009424.hg.1 | NM_001307983.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009424.hg.1 | NM_001307983.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009424.hg.1 | NM_001307983.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009424.hg.1 | NM_001307983.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009424.hg.1 | NM_001307983.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009424.hg.1 | NM_001307983.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009424.hg.1 | NM_001307983.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009424.hg.1 | NM_001307983.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009424.hg.1 | NM_001307983.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009424.hg.1 | NM_138326.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009424.hg.1 | NM_138326.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009424.hg.1 | NM_138326.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009424.hg.1 | NM_138326.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009424.hg.1 | NM_138326.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009424.hg.1 | NM_138326.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009424.hg.1 | NM_138326.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009424.hg.1 | NM_138326.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009424.hg.1 | NM_138326.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009424.hg.1 | NM_138326.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009424.hg.1 | NM_138326.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009424.hg.1 | NM_138326.2 | Refseq |

ACMSD
Genomic Map