Assay Details
Gene Symbol: ARHGAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_004308.3 | NP_004299.1 | 10 - 11 | 1060 | 61 | |
XM_011520095.1 | XP_011518397.1 | 10 - 11 | 1497 | 61 | ||
XM_017017720.1 | XP_016873209.1 | 8 - 9 | 813 | 61 | ||
GenBank mRNA | AK298562.1 | - | 10 - 11 | 1007 | 61 | |
BC018118.1 | - | 10 - 11 | 965 | 3896574 | 61 | |
U02570.1 | - | 10 - 11 | 967 | 61 | ||
Z23024.1 | - | 10 - 11 | 994 | 61 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC11001723.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
small GTPase mediated signal transduction
Rho protein signal transduction
positive regulation of signal transduction
positive regulation of GTPase activity
regulation of small GTPase mediated signal transduction
cell-cell adhesion
SH3/SH2 adaptor activity
GTPase activator activity
protein binding
SH3 domain binding
cadherin binding involved in cell-cell adhesion
small GTPase mediated signal transduction
Rho protein signal transduction
positive regulation of signal transduction
positive regulation of GTPase activity
regulation of small GTPase mediated signal transduction
cell-cell adhesion
SH3/SH2 adaptor activity
GTPase activator activity
protein binding
SH3 domain binding
cadherin binding involved in cell-cell adhesion
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1100010712.hg.1 | ENST00000311956.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010712.hg.1 | ENST00000311956.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010712.hg.1 | ENST00000311956.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010712.hg.1 | ENST00000311956.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010712.hg.1 | ENST00000311956.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010712.hg.1 | ENST00000311956.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010712.hg.1 | ENST00000311956.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010712.hg.1 | ENST00000311956.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010712.hg.1 | ENST00000311956.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010712.hg.1 | ENST00000311956.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010712.hg.1 | ENST00000311956.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010712.hg.1 | ENST00000311956.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010712.hg.1 | ENST00000311956.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010712.hg.1 | ENST00000311956.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010712.hg.1 | ENST00000311956.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010712.hg.1 | ENST00000311956.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010712.hg.1 | ENST00000311956.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010712.hg.1 | ENST00000524594.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010712.hg.1 | ENST00000524594.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010712.hg.1 | ENST00000524594.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010712.hg.1 | ENST00000524594.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010712.hg.1 | ENST00000524594.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010712.hg.1 | ENST00000525488.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010712.hg.1 | ENST00000525488.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010712.hg.1 | ENST00000525488.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010712.hg.1 | ENST00000525488.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010712.hg.1 | ENST00000525488.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010712.hg.1 | ENST00000525488.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010712.hg.1 | ENST00000525488.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010712.hg.1 | ENST00000525488.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010712.hg.1 | ENST00000526423.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010712.hg.1 | ENST00000526423.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010712.hg.1 | ENST00000526423.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010712.hg.1 | ENST00000526423.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010712.hg.1 | ENST00000526423.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010712.hg.1 | ENST00000526423.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010712.hg.1 | ENST00000526423.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010712.hg.1 | ENST00000526423.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010712.hg.1 | ENST00000526423.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010712.hg.1 | ENST00000526423.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010712.hg.1 | ENST00000526423.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010712.hg.1 | ENST00000526423.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010712.hg.1 | ENST00000527588.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010712.hg.1 | ENST00000527588.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010712.hg.1 | ENST00000527588.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010712.hg.1 | ENST00000527588.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010712.hg.1 | ENST00000527588.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010712.hg.1 | ENST00000527588.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010712.hg.1 | ENST00000527588.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010712.hg.1 | ENST00000527588.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010712.hg.1 | ENST00000528837.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010712.hg.1 | ENST00000528837.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010712.hg.1 | ENST00000528837.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010712.hg.1 | ENST00000528837.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010712.hg.1 | ENST00000528837.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010712.hg.1 | ENST00000528837.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010712.hg.1 | ENST00000528837.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010712.hg.1 | ENST00000528837.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010712.hg.1 | ENST00000528837.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010712.hg.1 | ENST00000528837.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010712.hg.1 | ENST00000528837.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010712.hg.1 | ENST00000529960.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010712.hg.1 | ENST00000529960.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010712.hg.1 | ENST00000529960.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010712.hg.1 | ENST00000529960.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010712.hg.1 | ENST00000529960.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010712.hg.1 | ENST00000529960.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010712.hg.1 | NM_004308.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100010712.hg.1 | NM_004308.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100010712.hg.1 | NM_004308.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100010712.hg.1 | NM_004308.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100010712.hg.1 | NM_004308.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100010712.hg.1 | NM_004308.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100010712.hg.1 | NM_004308.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100010712.hg.1 | NM_004308.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100010712.hg.1 | NM_004308.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100010712.hg.1 | NM_004308.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100010712.hg.1 | NM_004308.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100010712.hg.1 | NM_004308.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100010712.hg.1 | NM_004308.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100010712.hg.1 | NM_004308.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100010712.hg.1 | NM_004308.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100010712.hg.1 | NM_004308.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100010712.hg.1 | NM_004308.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100010712.hg.1 | NM_004308.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR11020026.hg.1 | ENST00000311956.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR11020026.hg.1 | ENST00000443332.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR11020026.hg.1 | NM_004308.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR11020026.hg.1 | TC11001723.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | TC1100010712.hg.1 | ENST00000311956.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010712.hg.1 | ENST00000311956.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010712.hg.1 | ENST00000311956.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010712.hg.1 | ENST00000311956.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010712.hg.1 | ENST00000311956.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010712.hg.1 | ENST00000311956.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010712.hg.1 | ENST00000311956.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010712.hg.1 | ENST00000311956.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010712.hg.1 | ENST00000311956.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010712.hg.1 | ENST00000311956.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010712.hg.1 | ENST00000311956.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010712.hg.1 | ENST00000311956.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010712.hg.1 | ENST00000311956.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010712.hg.1 | ENST00000311956.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010712.hg.1 | ENST00000311956.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010712.hg.1 | ENST00000311956.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010712.hg.1 | ENST00000311956.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010712.hg.1 | ENST00000524594.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010712.hg.1 | ENST00000524594.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010712.hg.1 | ENST00000524594.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010712.hg.1 | ENST00000524594.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010712.hg.1 | ENST00000524594.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010712.hg.1 | ENST00000525488.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010712.hg.1 | ENST00000525488.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010712.hg.1 | ENST00000525488.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010712.hg.1 | ENST00000525488.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010712.hg.1 | ENST00000525488.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010712.hg.1 | ENST00000525488.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010712.hg.1 | ENST00000525488.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010712.hg.1 | ENST00000525488.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010712.hg.1 | ENST00000526423.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010712.hg.1 | ENST00000526423.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010712.hg.1 | ENST00000526423.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010712.hg.1 | ENST00000526423.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010712.hg.1 | ENST00000526423.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010712.hg.1 | ENST00000526423.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010712.hg.1 | ENST00000526423.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010712.hg.1 | ENST00000526423.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010712.hg.1 | ENST00000526423.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010712.hg.1 | ENST00000526423.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010712.hg.1 | ENST00000526423.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010712.hg.1 | ENST00000526423.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010712.hg.1 | ENST00000527588.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010712.hg.1 | ENST00000527588.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010712.hg.1 | ENST00000527588.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010712.hg.1 | ENST00000527588.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010712.hg.1 | ENST00000527588.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010712.hg.1 | ENST00000527588.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010712.hg.1 | ENST00000527588.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010712.hg.1 | ENST00000527588.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010712.hg.1 | ENST00000528837.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010712.hg.1 | ENST00000528837.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010712.hg.1 | ENST00000528837.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010712.hg.1 | ENST00000528837.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010712.hg.1 | ENST00000528837.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010712.hg.1 | ENST00000528837.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010712.hg.1 | ENST00000528837.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010712.hg.1 | ENST00000528837.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010712.hg.1 | ENST00000528837.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010712.hg.1 | ENST00000528837.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010712.hg.1 | ENST00000528837.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010712.hg.1 | ENST00000529960.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010712.hg.1 | ENST00000529960.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010712.hg.1 | ENST00000529960.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010712.hg.1 | ENST00000529960.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010712.hg.1 | ENST00000529960.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010712.hg.1 | ENST00000529960.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010712.hg.1 | NM_004308.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100010712.hg.1 | NM_004308.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100010712.hg.1 | NM_004308.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100010712.hg.1 | NM_004308.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100010712.hg.1 | NM_004308.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100010712.hg.1 | NM_004308.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100010712.hg.1 | NM_004308.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100010712.hg.1 | NM_004308.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100010712.hg.1 | NM_004308.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100010712.hg.1 | NM_004308.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100010712.hg.1 | NM_004308.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100010712.hg.1 | NM_004308.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100010712.hg.1 | NM_004308.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100010712.hg.1 | NM_004308.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100010712.hg.1 | NM_004308.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100010712.hg.1 | NM_004308.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100010712.hg.1 | NM_004308.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100010712.hg.1 | NM_004308.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100180933.hg.1 | ENST00000311956.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100180933.hg.1 | ENST00000526423.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100180933.hg.1 | ENST00000528837.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100180933.hg.1 | NM_004308.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100180933.hg.1 | ENST00000311956.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100180933.hg.1 | ENST00000526423.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100180933.hg.1 | ENST00000528837.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100180933.hg.1 | NM_004308.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100180933.hg.1 | TC1100010712.hg.1 | Clariom D TC |
