Assay Details
| Gene Symbol: ARHGAP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_004308.3 | NP_004299.1 | 7 - 8 | 795 | 84 | |
| XM_011520095.1 | XP_011518397.1 | 7 - 8 | 1232 | 84 | ||
| XM_017017720.1 | XP_016873209.1 | 5 - 6 | 548 | 84 | ||
| GenBank mRNA | AK298562.1 | - | 7 - 8 | 742 | 84 | |
| BC018118.1 | - | 7 - 8 | 700 | 3896574 | 84 | |
| U02570.1 | - | 7 - 8 | 702 | 84 | ||
| U62794.1 | - | 2 - 3 | 319 | 84 | ||
| Z23024.1 | - | 7 - 8 | 729 | 84 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC11001723.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
small GTPase mediated signal transduction
Rho protein signal transduction
positive regulation of signal transduction
positive regulation of GTPase activity
regulation of small GTPase mediated signal transduction
cell-cell adhesion
SH3/SH2 adaptor activity
GTPase activator activity
protein binding
SH3 domain binding
cadherin binding involved in cell-cell adhesion
small GTPase mediated signal transduction
Rho protein signal transduction
positive regulation of signal transduction
positive regulation of GTPase activity
regulation of small GTPase mediated signal transduction
cell-cell adhesion
SH3/SH2 adaptor activity
GTPase activator activity
protein binding
SH3 domain binding
cadherin binding involved in cell-cell adhesion
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1100010712.hg.1 | ENST00000311956.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010712.hg.1 | ENST00000311956.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010712.hg.1 | ENST00000311956.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010712.hg.1 | ENST00000311956.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010712.hg.1 | ENST00000311956.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010712.hg.1 | ENST00000311956.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010712.hg.1 | ENST00000311956.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010712.hg.1 | ENST00000311956.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010712.hg.1 | ENST00000311956.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010712.hg.1 | ENST00000311956.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010712.hg.1 | ENST00000311956.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010712.hg.1 | ENST00000311956.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010712.hg.1 | ENST00000311956.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010712.hg.1 | ENST00000311956.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010712.hg.1 | ENST00000311956.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010712.hg.1 | ENST00000311956.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010712.hg.1 | ENST00000311956.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010712.hg.1 | ENST00000524594.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010712.hg.1 | ENST00000524594.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010712.hg.1 | ENST00000524594.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010712.hg.1 | ENST00000524594.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010712.hg.1 | ENST00000524594.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010712.hg.1 | ENST00000525488.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010712.hg.1 | ENST00000525488.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010712.hg.1 | ENST00000525488.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010712.hg.1 | ENST00000525488.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010712.hg.1 | ENST00000525488.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010712.hg.1 | ENST00000525488.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010712.hg.1 | ENST00000525488.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010712.hg.1 | ENST00000525488.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010712.hg.1 | ENST00000526423.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010712.hg.1 | ENST00000526423.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010712.hg.1 | ENST00000526423.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010712.hg.1 | ENST00000526423.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010712.hg.1 | ENST00000526423.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010712.hg.1 | ENST00000526423.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010712.hg.1 | ENST00000526423.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010712.hg.1 | ENST00000526423.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010712.hg.1 | ENST00000526423.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010712.hg.1 | ENST00000526423.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010712.hg.1 | ENST00000526423.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010712.hg.1 | ENST00000526423.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010712.hg.1 | ENST00000527588.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010712.hg.1 | ENST00000527588.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010712.hg.1 | ENST00000527588.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010712.hg.1 | ENST00000527588.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010712.hg.1 | ENST00000527588.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010712.hg.1 | ENST00000527588.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010712.hg.1 | ENST00000527588.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010712.hg.1 | ENST00000527588.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010712.hg.1 | ENST00000528837.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010712.hg.1 | ENST00000528837.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010712.hg.1 | ENST00000528837.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010712.hg.1 | ENST00000528837.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010712.hg.1 | ENST00000528837.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010712.hg.1 | ENST00000528837.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010712.hg.1 | ENST00000528837.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010712.hg.1 | ENST00000528837.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010712.hg.1 | ENST00000528837.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010712.hg.1 | ENST00000528837.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010712.hg.1 | ENST00000528837.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010712.hg.1 | ENST00000529960.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010712.hg.1 | ENST00000529960.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010712.hg.1 | ENST00000529960.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010712.hg.1 | ENST00000529960.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010712.hg.1 | ENST00000529960.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010712.hg.1 | ENST00000529960.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010712.hg.1 | NM_004308.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010712.hg.1 | NM_004308.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010712.hg.1 | NM_004308.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010712.hg.1 | NM_004308.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010712.hg.1 | NM_004308.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010712.hg.1 | NM_004308.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010712.hg.1 | NM_004308.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010712.hg.1 | NM_004308.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010712.hg.1 | NM_004308.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010712.hg.1 | NM_004308.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010712.hg.1 | NM_004308.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010712.hg.1 | NM_004308.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010712.hg.1 | NM_004308.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010712.hg.1 | NM_004308.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010712.hg.1 | NM_004308.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010712.hg.1 | NM_004308.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010712.hg.1 | NM_004308.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010712.hg.1 | NM_004308.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11020029.hg.1 | ENST00000311956.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11020029.hg.1 | ENST00000443332.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11020029.hg.1 | NM_004308.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11020029.hg.1 | TC11001723.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1100180938.hg.1 | ENST00000311956.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100180938.hg.1 | ENST00000526423.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100180938.hg.1 | ENST00000528837.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100180938.hg.1 | NM_004308.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100180938.hg.1 | ENST00000311956.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100180938.hg.1 | ENST00000526423.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100180938.hg.1 | ENST00000528837.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100180938.hg.1 | NM_004308.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100180938.hg.1 | TC1100010712.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1100180937.hg.1 | ENST00000311956.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100180937.hg.1 | ENST00000526423.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100180937.hg.1 | ENST00000528837.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100180937.hg.1 | NM_004308.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100180937.hg.1 | ENST00000311956.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100180937.hg.1 | ENST00000526423.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100180937.hg.1 | ENST00000528837.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100180937.hg.1 | NM_004308.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100180937.hg.1 | TC1100010712.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1100010712.hg.1 | ENST00000311956.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010712.hg.1 | ENST00000311956.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010712.hg.1 | ENST00000311956.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010712.hg.1 | ENST00000311956.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010712.hg.1 | ENST00000311956.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010712.hg.1 | ENST00000311956.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010712.hg.1 | ENST00000311956.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010712.hg.1 | ENST00000311956.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010712.hg.1 | ENST00000311956.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010712.hg.1 | ENST00000311956.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010712.hg.1 | ENST00000311956.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010712.hg.1 | ENST00000311956.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010712.hg.1 | ENST00000311956.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010712.hg.1 | ENST00000311956.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010712.hg.1 | ENST00000311956.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010712.hg.1 | ENST00000311956.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010712.hg.1 | ENST00000311956.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010712.hg.1 | ENST00000524594.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010712.hg.1 | ENST00000524594.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010712.hg.1 | ENST00000524594.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010712.hg.1 | ENST00000524594.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010712.hg.1 | ENST00000524594.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010712.hg.1 | ENST00000525488.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010712.hg.1 | ENST00000525488.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010712.hg.1 | ENST00000525488.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010712.hg.1 | ENST00000525488.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010712.hg.1 | ENST00000525488.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010712.hg.1 | ENST00000525488.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010712.hg.1 | ENST00000525488.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010712.hg.1 | ENST00000525488.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010712.hg.1 | ENST00000526423.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010712.hg.1 | ENST00000526423.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010712.hg.1 | ENST00000526423.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010712.hg.1 | ENST00000526423.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010712.hg.1 | ENST00000526423.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010712.hg.1 | ENST00000526423.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010712.hg.1 | ENST00000526423.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010712.hg.1 | ENST00000526423.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010712.hg.1 | ENST00000526423.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010712.hg.1 | ENST00000526423.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010712.hg.1 | ENST00000526423.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010712.hg.1 | ENST00000526423.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010712.hg.1 | ENST00000527588.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010712.hg.1 | ENST00000527588.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010712.hg.1 | ENST00000527588.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010712.hg.1 | ENST00000527588.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010712.hg.1 | ENST00000527588.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010712.hg.1 | ENST00000527588.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010712.hg.1 | ENST00000527588.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010712.hg.1 | ENST00000527588.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010712.hg.1 | ENST00000528837.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010712.hg.1 | ENST00000528837.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010712.hg.1 | ENST00000528837.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010712.hg.1 | ENST00000528837.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010712.hg.1 | ENST00000528837.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010712.hg.1 | ENST00000528837.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010712.hg.1 | ENST00000528837.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010712.hg.1 | ENST00000528837.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010712.hg.1 | ENST00000528837.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010712.hg.1 | ENST00000528837.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010712.hg.1 | ENST00000528837.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010712.hg.1 | ENST00000529960.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010712.hg.1 | ENST00000529960.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010712.hg.1 | ENST00000529960.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010712.hg.1 | ENST00000529960.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010712.hg.1 | ENST00000529960.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010712.hg.1 | ENST00000529960.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010712.hg.1 | NM_004308.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010712.hg.1 | NM_004308.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010712.hg.1 | NM_004308.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010712.hg.1 | NM_004308.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010712.hg.1 | NM_004308.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010712.hg.1 | NM_004308.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010712.hg.1 | NM_004308.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010712.hg.1 | NM_004308.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010712.hg.1 | NM_004308.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010712.hg.1 | NM_004308.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010712.hg.1 | NM_004308.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010712.hg.1 | NM_004308.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010712.hg.1 | NM_004308.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010712.hg.1 | NM_004308.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010712.hg.1 | NM_004308.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010712.hg.1 | NM_004308.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010712.hg.1 | NM_004308.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010712.hg.1 | NM_004308.3 | Refseq |

ARHGAP1
Genomic Map